Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 133228464-133228964 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WISP1
Entrez Gene ID
8840
Gene Name
WNT1 inducible signaling pathway protein 1
Gene Aliases
CCN4, WISP1c, WISP1i, WISP1tc
UniGene
Hs.492974
Gene Chromosome Location
Chr.8: 133191039-133231690 on Build GRCh38
Transcripts
NM_001204869.1 NM_001204870.1 NM_003882.3 NM_080838.2 NR_037944.1

SNP Detail

SNP ID Chromosome Location
rs12164193 Chr.8: 133228680-133228680 on Build GRCh38
rs540950347 Chr.8: 133228791-133228791 on Build GRCh38
rs559660325 Chr.8: 133228798-133228798 on Build GRCh38
rs923548019 Chr.8: 133228520-133228520 on Build GRCh38
rs965687782 Chr.8: 133228789-133228790 on Build GRCh38
rs978976959 Chr.8: 133228812-133228812 on Build GRCh38
rs1048831644 Chr.8: 133228922-133228922 on Build GRCh38
rs1015783840 Chr.8: 133228708-133228708 on Build GRCh38
rs4265166 Chr.8: 133228843-133228843 on Build GRCh38
rs1056855696 Chr.8: 133228891-133228891 on Build GRCh38
rs1055895975 Chr.8: 133228876-133228876 on Build GRCh38
rs962193267 Chr.8: 133228734-133228734 on Build GRCh38
rs533269074 Chr.8: 133228799-133228799 on Build GRCh38
rs888101569 Chr.8: 133228925-133228925 on Build GRCh38
rs1055843234 Chr.8: 133228576-133228576 on Build GRCh38
rs145960909 Chr.8: 133228689-133228689 on Build GRCh38
rs1014144709 Chr.8: 133228596-133228596 on Build GRCh38
rs751107346 Chr.8: 133228604-133228604 on Build GRCh38
rs1035499242 Chr.8: 133228668-133228668 on Build GRCh38
rs894375804 Chr.8: 133228672-133228672 on Build GRCh38
rs572089298 Chr.8: 133228566-133228566 on Build GRCh38
rs932186986 Chr.8: 133228793-133228793 on Build GRCh38
rs953173720 Chr.8: 133228692-133228692 on Build GRCh38
rs1012780542 Chr.8: 133228919-133228919 on Build GRCh38
rs1042232047 Chr.8: 133228606-133228606 on Build GRCh38
rs780194947 Chr.8: 133228580-133228580 on Build GRCh38
rs569907417 Chr.8: 133228870-133228870 on Build GRCh38
rs1002689567 Chr.8: 133228665-133228665 on Build GRCh38
rs893133641 Chr.8: 133228560-133228560 on Build GRCh38
rs182003497 Chr.8: 133228764-133228764 on Build GRCh38
rs923435402 Chr.8: 133228768-133228768 on Build GRCh38
rs765566205 Chr.8: 133228752-133228752 on Build GRCh38
rs947396557 Chr.8: 133228819-133228819 on Build GRCh38
rs2929970 Chr.8: 133228894-133228894 on Build GRCh38
rs73362554 Chr.8: 133228695-133228695 on Build GRCh38
rs1050408742 Chr.8: 133228554-133228554 on Build GRCh38
rs938784743 Chr.8: 133228867-133228867 on Build GRCh38
rs1041789563 Chr.8: 133228829-133228829 on Build GRCh38
rs763332091 Chr.8: 133228893-133228893 on Build GRCh38
rs558245735 Chr.8: 133228682-133228682 on Build GRCh38
rs932234735 Chr.8: 133228525-133228525 on Build GRCh38
rs986821782 Chr.8: 133228794-133228794 on Build GRCh38
rs967937376 Chr.8: 133228805-133228805 on Build GRCh38
rs902995216 Chr.8: 133228645-133228645 on Build GRCh38
rs1036658336 Chr.8: 133228915-133228915 on Build GRCh38
rs947485813 Chr.8: 133228597-133228597 on Build GRCh38

View Full Product Details