Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 133237475-133237978 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NDRG1
Entrez Gene ID
10397
Gene Name
N-myc downstream regulated 1
Gene Aliases
CAP43, CMT4D, DRG-1, DRG1, GC4, HMSNL, NDR1, NMSL, PROXY1, RIT42, RTP, TARG1, TDD5
UniGene
Hs.372914
Gene Chromosome Location
Chr.8: 133237171-133297587 on Build GRCh38
Transcripts
NM_001135242.1 NM_001258432.1 NM_001258433.1 NM_006096.3 XM_011516791.1 XM_011516792.1

SNP Detail

SNP ID Chromosome Location
rs920332363 Chr.8: 133237630-133237630 on Build GRCh38
rs973863842 Chr.8: 133237760-133237760 on Build GRCh38
rs899516376 Chr.8: 133237608-133237608 on Build GRCh38
rs569279826 Chr.8: 133237954-133237954 on Build GRCh38
rs770548372 Chr.8: 133237881-133237883 on Build GRCh38
rs868286576 Chr.8: 133237921-133237921 on Build GRCh38
rs1031086247 Chr.8: 133237693-133237693 on Build GRCh38
rs766707808 Chr.8: 133237791-133237791 on Build GRCh38
rs879441181 Chr.8: 133237705-133237705 on Build GRCh38
rs776885479 Chr.8: 133237906-133237906 on Build GRCh38
rs576876313 Chr.8: 133237558-133237558 on Build GRCh38
rs926267815 Chr.8: 133237579-133237579 on Build GRCh38
rs900066936 Chr.8: 133237929-133237929 on Build GRCh38
rs929059375 Chr.8: 133237634-133237634 on Build GRCh38
rs920917027 Chr.8: 133237773-133237773 on Build GRCh38
rs1019573490 Chr.8: 133237523-133237523 on Build GRCh38
rs144034977 Chr.8: 133237584-133237584 on Build GRCh38
rs747671981 Chr.8: 133237563-133237563 on Build GRCh38
rs1010089084 Chr.8: 133237737-133237737 on Build GRCh38
rs1049697 Chr.8: 133237922-133237922 on Build GRCh38
rs1008921666 Chr.8: 133237521-133237521 on Build GRCh38
rs973033664 Chr.8: 133237834-133237834 on Build GRCh38
rs973067702 Chr.8: 133237592-133237592 on Build GRCh38
rs554339360 Chr.8: 133237533-133237533 on Build GRCh38
rs929071157 Chr.8: 133237912-133237912 on Build GRCh38
rs912184966 Chr.8: 133237842-133237842 on Build GRCh38
rs879342771 Chr.8: 133237700-133237700 on Build GRCh38
rs1803463 Chr.8: 133237830-133237830 on Build GRCh38
rs550687871 Chr.8: 133237933-133237933 on Build GRCh38
rs886062711 Chr.8: 133237582-133237582 on Build GRCh38
rs191345335 Chr.8: 133237557-133237557 on Build GRCh38
rs970652341 Chr.8: 133237571-133237571 on Build GRCh38
rs761054650 Chr.8: 133237684-133237684 on Build GRCh38
rs1032770932 Chr.8: 133237722-133237722 on Build GRCh38
rs980642396 Chr.8: 133237714-133237715 on Build GRCh38
rs764696488 Chr.8: 133237744-133237744 on Build GRCh38
rs543373151 Chr.8: 133237701-133237701 on Build GRCh38
rs964049216 Chr.8: 133237530-133237530 on Build GRCh38
rs944288492 Chr.8: 133237692-133237692 on Build GRCh38
rs776620022 Chr.8: 133237946-133237946 on Build GRCh38
rs572727755 Chr.8: 133237537-133237537 on Build GRCh38
rs773742921 Chr.8: 133237581-133237581 on Build GRCh38
rs1041250053 Chr.8: 133237699-133237699 on Build GRCh38
rs981948726 Chr.8: 133237774-133237774 on Build GRCh38
rs1020783450 Chr.8: 133237738-133237738 on Build GRCh38
rs539924589 Chr.8: 133237539-133237539 on Build GRCh38
rs890611804 Chr.8: 133237662-133237662 on Build GRCh38
rs557737109 Chr.8: 133237578-133237578 on Build GRCh38
rs10675 Chr.8: 133237743-133237743 on Build GRCh38
rs969668808 Chr.8: 133237551-133237551 on Build GRCh38
rs561506934 Chr.8: 133237683-133237683 on Build GRCh38
rs893877081 Chr.8: 133237736-133237736 on Build GRCh38
rs184598535 Chr.8: 133237647-133237647 on Build GRCh38
rs1033555679 Chr.8: 133237790-133237790 on Build GRCh38
rs932193715 Chr.8: 133237636-133237636 on Build GRCh38
rs944950081 Chr.8: 133237888-133237888 on Build GRCh38
rs887335765 Chr.8: 133237517-133237517 on Build GRCh38
rs892508822 Chr.8: 133237695-133237695 on Build GRCh38
rs1043843453 Chr.8: 133237947-133237947 on Build GRCh38
rs1041216652 Chr.8: 133237927-133237927 on Build GRCh38
rs547240463 Chr.8: 133237891-133237891 on Build GRCh38
rs12668 Chr.8: 133237538-133237538 on Build GRCh38
rs932331118 Chr.8: 133237781-133237781 on Build GRCh38
rs538964895 Chr.8: 133237796-133237796 on Build GRCh38
rs532646260 Chr.8: 133237931-133237931 on Build GRCh38
rs983642550 Chr.8: 133237918-133237918 on Build GRCh38
rs996781179 Chr.8: 133237712-133237712 on Build GRCh38

View Full Product Details