Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 142660380-142660912 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
JRK
Entrez Gene ID
8629
Gene Name
Jrk helix-turn-helix protein
Gene Aliases
JH8, jerky
UniGene
Hs.535903
Gene Chromosome Location
Chr.8: 142651501-142670132 on Build GRCh38
Transcripts
NM_001077527.2 NM_001279352.1 NM_003724.3 XM_006716677.3 XM_006716678.3 XM_011517354.1 XM_011517355.1

SNP Detail

SNP ID Chromosome Location
rs1003927970 Chr.8: 142660744-142660744 on Build GRCh38
rs782629877 Chr.8: 142660716-142660716 on Build GRCh38
rs978722558 Chr.8: 142660822-142660822 on Build GRCh38
rs966597395 Chr.8: 142660887-142660887 on Build GRCh38
rs978890542 Chr.8: 142660524-142660524 on Build GRCh38
rs988944548 Chr.8: 142660586-142660586 on Build GRCh38
rs996621827 Chr.8: 142660454-142660454 on Build GRCh38
rs959989297 Chr.8: 142660516-142660516 on Build GRCh38
rs782161424 Chr.8: 142660478-142660478 on Build GRCh38
rs912021687 Chr.8: 142660660-142660660 on Build GRCh38
rs782184413 Chr.8: 142660831-142660831 on Build GRCh38
rs587704622 Chr.8: 142660697-142660697 on Build GRCh38
rs1026718034 Chr.8: 142660457-142660457 on Build GRCh38
rs1023284390 Chr.8: 142660807-142660807 on Build GRCh38
rs926035077 Chr.8: 142660556-142660556 on Build GRCh38
rs4736365 Chr.8: 142660785-142660785 on Build GRCh38
rs4736364 Chr.8: 142660701-142660701 on Build GRCh38
rs781803489 Chr.8: 142660575-142660575 on Build GRCh38
rs1015402993 Chr.8: 142660512-142660512 on Build GRCh38
rs1048207004 Chr.8: 142660728-142660728 on Build GRCh38
rs200289800 Chr.8: 142660699-142660700 on Build GRCh38
rs587724782 Chr.8: 142660461-142660461 on Build GRCh38
rs763628611 Chr.8: 142660760-142660760 on Build GRCh38
rs587693073 Chr.8: 142660796-142660796 on Build GRCh38
rs587603302 Chr.8: 142660508-142660508 on Build GRCh38
rs147786364 Chr.8: 142660840-142660840 on Build GRCh38
rs4736363 Chr.8: 142660700-142660700 on Build GRCh38
rs71507442 Chr.8: 142660700-142660701 on Build GRCh38
rs782612834 Chr.8: 142660698-142660698 on Build GRCh38
rs944995490 Chr.8: 142660667-142660667 on Build GRCh38
rs912076070 Chr.8: 142660882-142660882 on Build GRCh38
rs1004041157 Chr.8: 142660511-142660511 on Build GRCh38
rs112107379 Chr.8: 142660523-142660523 on Build GRCh38
rs1022980625 Chr.8: 142660520-142660520 on Build GRCh38
rs988832084 Chr.8: 142660852-142660852 on Build GRCh38
rs970122107 Chr.8: 142660813-142660813 on Build GRCh38
rs888205385 Chr.8: 142660733-142660733 on Build GRCh38
rs993133665 Chr.8: 142660517-142660517 on Build GRCh38
rs782466190 Chr.8: 142660591-142660591 on Build GRCh38
rs765279659 Chr.8: 142660865-142660865 on Build GRCh38
rs587738195 Chr.8: 142660749-142660749 on Build GRCh38

View Full Product Details