Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 143791282-143791785 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SCRIB
Entrez Gene ID
23513
Gene Name
scribbled planar cell polarity protein
Gene Aliases
CRIB1, SCRB1, SCRIB1, Vartul
UniGene
Hs.436329
Gene Chromosome Location
Chr.8: 143790920-143815379 on Build GRCh38
Transcripts
NM_015356.4 NM_182706.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3646121 c.4799C>T p.S1600F Chr.8: 143791412-143791412 on Build GRCh38
6773414 c.4733C>T p.A1578V Chr.8: 143791703-143791703 on Build GRCh38
5434012 c.4770+1G>A p.? Chr.8: 143791665-143791665 on Build GRCh38
1097015 c.4710G>T p.K1570N Chr.8: 143791726-143791726 on Build GRCh38
6499638 c.4831G>A p.E1611K Chr.8: 143791300-143791300 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs901584211 Chr.8: 143791678-143791678 on Build GRCh38
rs530339737 Chr.8: 143791525-143791525 on Build GRCh38
rs782154485 Chr.8: 143791348-143791348 on Build GRCh38
rs554743454 Chr.8: 143791566-143791566 on Build GRCh38
rs782676114 Chr.8: 143791676-143791676 on Build GRCh38
rs782581658 Chr.8: 143791761-143791761 on Build GRCh38
rs781787552 Chr.8: 143791648-143791648 on Build GRCh38
rs782770759 Chr.8: 143791343-143791343 on Build GRCh38
rs782165990 Chr.8: 143791320-143791321 on Build GRCh38
rs781860410 Chr.8: 143791634-143791634 on Build GRCh38
rs781923036 Chr.8: 143791701-143791701 on Build GRCh38
rs940053854 Chr.8: 143791449-143791449 on Build GRCh38
rs782531324 Chr.8: 143791760-143791760 on Build GRCh38
rs781848499 Chr.8: 143791347-143791347 on Build GRCh38
rs782418099 Chr.8: 143791480-143791480 on Build GRCh38
rs200783841 Chr.8: 143791304-143791304 on Build GRCh38
rs532895515 Chr.8: 143791502-143791502 on Build GRCh38
rs782125930 Chr.8: 143791744-143791744 on Build GRCh38
rs781873171 Chr.8: 143791467-143791467 on Build GRCh38
rs574303852 Chr.8: 143791320-143791320 on Build GRCh38
rs782442684 Chr.8: 143791376-143791376 on Build GRCh38
rs782812845 Chr.8: 143791718-143791718 on Build GRCh38
rs980290475 Chr.8: 143791610-143791610 on Build GRCh38
rs558561221 Chr.8: 143791556-143791556 on Build GRCh38
rs782554857 Chr.8: 143791637-143791637 on Build GRCh38
rs887620899 Chr.8: 143791739-143791739 on Build GRCh38
rs565277281 Chr.8: 143791497-143791497 on Build GRCh38
rs548729661 Chr.8: 143791544-143791544 on Build GRCh38
rs782324976 Chr.8: 143791700-143791700 on Build GRCh38
rs891468268 Chr.8: 143791536-143791536 on Build GRCh38
rs782166837 Chr.8: 143791534-143791534 on Build GRCh38
rs541690927 Chr.8: 143791703-143791703 on Build GRCh38
rs782345419 Chr.8: 143791453-143791453 on Build GRCh38
rs557094565 Chr.8: 143791729-143791729 on Build GRCh38
rs146664605 Chr.8: 143791305-143791305 on Build GRCh38
rs782382186 Chr.8: 143791314-143791314 on Build GRCh38
rs560261414 Chr.8: 143791706-143791706 on Build GRCh38
rs376658629 Chr.8: 143791352-143791352 on Build GRCh38
rs1029274869 Chr.8: 143791567-143791567 on Build GRCh38
rs201700942 Chr.8: 143791379-143791379 on Build GRCh38
rs9314417 Chr.8: 143791609-143791609 on Build GRCh38
rs909855857 Chr.8: 143791581-143791581 on Build GRCh38
rs1054645186 Chr.8: 143791621-143791621 on Build GRCh38
rs570105502 Chr.8: 143791582-143791582 on Build GRCh38
rs565546823 Chr.8: 143791656-143791656 on Build GRCh38
rs972755047 Chr.8: 143791602-143791602 on Build GRCh38
rs781893499 Chr.8: 143791508-143791508 on Build GRCh38
rs782263775 Chr.8: 143791661-143791661 on Build GRCh38
rs372232275 Chr.8: 143791328-143791328 on Build GRCh38
rs782520307 Chr.8: 143791397-143791397 on Build GRCh38
rs782010485 Chr.8: 143791492-143791492 on Build GRCh38
rs374751896 Chr.8: 143791469-143791469 on Build GRCh38
rs782495308 Chr.8: 143791649-143791649 on Build GRCh38
rs558316607 Chr.8: 143791574-143791574 on Build GRCh38
rs71318627 Chr.8: 143791556-143791557 on Build GRCh38
rs781833586 Chr.8: 143791391-143791391 on Build GRCh38
rs782693712 Chr.8: 143791624-143791624 on Build GRCh38
rs782070566 Chr.8: 143791615-143791615 on Build GRCh38
rs781810795 Chr.8: 143791368-143791368 on Build GRCh38
rs782034459 Chr.8: 143791316-143791316 on Build GRCh38
rs149841848 Chr.8: 143791436-143791436 on Build GRCh38
rs782291991 Chr.8: 143791412-143791412 on Build GRCh38
rs782735344 Chr.8: 143791733-143791733 on Build GRCh38
rs781971473 Chr.8: 143791326-143791326 on Build GRCh38
rs782781865 Chr.8: 143791628-143791628 on Build GRCh38
rs566329991 Chr.8: 143791517-143791517 on Build GRCh38
rs369874730 Chr.8: 143791755-143791755 on Build GRCh38
rs373471641 Chr.8: 143791619-143791619 on Build GRCh38
rs782103174 Chr.8: 143791658-143791658 on Build GRCh38
rs975040340 Chr.8: 143791363-143791363 on Build GRCh38
rs1040045238 Chr.8: 143791660-143791660 on Build GRCh38
rs369889971 Chr.8: 143791372-143791372 on Build GRCh38
rs782449541 Chr.8: 143791755-143791756 on Build GRCh38
rs376717824 Chr.8: 143791754-143791754 on Build GRCh38
rs1028793323 Chr.8: 143791727-143791727 on Build GRCh38
rs534280317 Chr.8: 143791594-143791594 on Build GRCh38
rs1006119494 Chr.8: 143791750-143791750 on Build GRCh38
rs782275223 Chr.8: 143791696-143791696 on Build GRCh38
rs1018668826 Chr.8: 143791550-143791550 on Build GRCh38
rs782498175 Chr.8: 143791663-143791663 on Build GRCh38
rs373434132 Chr.8: 143791503-143791503 on Build GRCh38
rs1009910296 Chr.8: 143791545-143791545 on Build GRCh38
rs782810844 Chr.8: 143791638-143791638 on Build GRCh38
rs184899027 Chr.8: 143791319-143791319 on Build GRCh38
rs574564419 Chr.8: 143791603-143791603 on Build GRCh38
rs782092419 Chr.8: 143791626-143791626 on Build GRCh38
rs200271090 Chr.8: 143791408-143791408 on Build GRCh38
rs782006464 Chr.8: 143791709-143791709 on Build GRCh38
rs782767624 Chr.8: 143791455-143791456 on Build GRCh38
rs927453926 Chr.8: 143791611-143791611 on Build GRCh38
rs782222427 Chr.8: 143791448-143791448 on Build GRCh38
rs782118671 Chr.8: 143791713-143791713 on Build GRCh38
rs996332567 Chr.8: 143791717-143791717 on Build GRCh38
rs371196827 Chr.8: 143791468-143791468 on Build GRCh38
rs9314416 Chr.8: 143791496-143791496 on Build GRCh38
rs782020257 Chr.8: 143791726-143791726 on Build GRCh38
rs561238876 Chr.8: 143791435-143791435 on Build GRCh38
rs540079348 Chr.8: 143791690-143791690 on Build GRCh38

View Full Product Details