Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 32466061-32466588 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX58
Entrez Gene ID
23586
Gene Name
DEXD/H-box helicase 58
Gene Aliases
RIG-I, RIGI, RLR-1, SGMRT2
UniGene
Hs.190622
Gene Chromosome Location
Chr.9: 32455302-32526324 on Build GRCh38
Transcripts
NM_014314.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6258149 c.2124+10C>A p.? Chr.9: 32466280-32466280 on Build GRCh38
168026 c.2272G>T p.E758* Chr.9: 32466355-32466355 on Build GRCh38
7364010 c.2321C>A p.A774E Chr.9: 32466306-32466306 on Build GRCh38
6258148 c.2337+10C>A p.? Chr.9: 32466280-32466280 on Build GRCh38
5979674 c.2059G>T p.E687* Chr.9: 32466355-32466355 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781464142 Chr.9: 32466345-32466345 on Build GRCh38
rs762952485 Chr.9: 32466284-32466284 on Build GRCh38
rs369407062 Chr.9: 32466282-32466282 on Build GRCh38
rs748767192 Chr.9: 32466154-32466154 on Build GRCh38
rs376877588 Chr.9: 32466141-32466141 on Build GRCh38
rs1024817804 Chr.9: 32466503-32466503 on Build GRCh38
rs146146421 Chr.9: 32466208-32466208 on Build GRCh38
rs2274864 Chr.9: 32466186-32466186 on Build GRCh38
rs754416719 Chr.9: 32466444-32466444 on Build GRCh38
rs760553867 Chr.9: 32466434-32466434 on Build GRCh38
rs753504869 Chr.9: 32466443-32466443 on Build GRCh38
rs772925636 Chr.9: 32466388-32466388 on Build GRCh38
rs374631860 Chr.9: 32466145-32466145 on Build GRCh38
rs144181118 Chr.9: 32466310-32466310 on Build GRCh38
rs867148941 Chr.9: 32466461-32466461 on Build GRCh38
rs560209567 Chr.9: 32466136-32466136 on Build GRCh38
rs765664278 Chr.9: 32466185-32466189 on Build GRCh38
rs776842809 Chr.9: 32466269-32466269 on Build GRCh38
rs904822679 Chr.9: 32466307-32466307 on Build GRCh38
rs112363590 Chr.9: 32466323-32466323 on Build GRCh38
rs883948 Chr.9: 32466280-32466280 on Build GRCh38
rs45460592 Chr.9: 32466464-32466464 on Build GRCh38
rs764082325 Chr.9: 32466290-32466290 on Build GRCh38
rs376349759 Chr.9: 32466322-32466322 on Build GRCh38
rs1046722296 Chr.9: 32466300-32466300 on Build GRCh38
rs764525003 Chr.9: 32466191-32466191 on Build GRCh38
rs56377445 Chr.9: 32466192-32466192 on Build GRCh38
rs746076420 Chr.9: 32466347-32466347 on Build GRCh38
rs539721474 Chr.9: 32466380-32466380 on Build GRCh38
rs553716308 Chr.9: 32466355-32466355 on Build GRCh38
rs754509057 Chr.9: 32466336-32466336 on Build GRCh38
rs772611141 Chr.9: 32466241-32466241 on Build GRCh38
rs751946210 Chr.9: 32466460-32466464 on Build GRCh38
rs959538227 Chr.9: 32466105-32466105 on Build GRCh38
rs45589431 Chr.9: 32466279-32466279 on Build GRCh38
rs763131643 Chr.9: 32466376-32466377 on Build GRCh38
rs753309872 Chr.9: 32466334-32466334 on Build GRCh38
rs757526001 Chr.9: 32466475-32466497 on Build GRCh38
rs990691218 Chr.9: 32466118-32466118 on Build GRCh38
rs142767943 Chr.9: 32466363-32466363 on Build GRCh38
rs372515643 Chr.9: 32466328-32466328 on Build GRCh38
rs765885518 Chr.9: 32466329-32466329 on Build GRCh38
rs747516523 Chr.9: 32466342-32466342 on Build GRCh38
rs372312705 Chr.9: 32466311-32466311 on Build GRCh38
rs764791411 Chr.9: 32466463-32466463 on Build GRCh38
rs1049967126 Chr.9: 32466214-32466214 on Build GRCh38
rs764494800 Chr.9: 32466427-32466427 on Build GRCh38
rs747327882 Chr.9: 32466253-32466253 on Build GRCh38
rs981492319 Chr.9: 32466167-32466167 on Build GRCh38
rs765902183 Chr.9: 32466442-32466442 on Build GRCh38
rs778286801 Chr.9: 32466249-32466249 on Build GRCh38
rs749330817 Chr.9: 32466364-32466364 on Build GRCh38
rs372618054 Chr.9: 32466132-32466132 on Build GRCh38
rs200275444 Chr.9: 32466268-32466268 on Build GRCh38
rs774428075 Chr.9: 32466369-32466369 on Build GRCh38
rs942743406 Chr.9: 32466171-32466171 on Build GRCh38
rs368263807 Chr.9: 32466327-32466327 on Build GRCh38
rs148502654 Chr.9: 32466341-32466341 on Build GRCh38
rs908547290 Chr.9: 32466169-32466169 on Build GRCh38
rs775480990 Chr.9: 32466283-32466283 on Build GRCh38
rs748636479 Chr.9: 32466240-32466240 on Build GRCh38
rs1032064406 Chr.9: 32466312-32466312 on Build GRCh38
rs373292486 Chr.9: 32466402-32466402 on Build GRCh38
rs570533013 Chr.9: 32466373-32466373 on Build GRCh38
rs1039042127 Chr.9: 32466189-32466189 on Build GRCh38
rs755750875 Chr.9: 32466091-32466091 on Build GRCh38
rs768864196 Chr.9: 32466365-32466365 on Build GRCh38

View Full Product Details