Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 36632839-36633340 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MELK
Entrez Gene ID
9833
Gene Name
maternal embryonic leucine zipper kinase
Gene Aliases
HPK38
UniGene
Hs.184339
Gene Chromosome Location
Chr.9: 36572862-36677683 on Build GRCh38
Transcripts
NM_001256685.1 NM_001256687.1 NM_001256688.1 NM_001256689.1 NM_001256690.1 NM_001256691.1 NM_001256692.1 NM_001256693.1 NM_014791.3 NR_046337.1 XM_011518076.2 XM_011518077.1 XM_011518078.2 XM_011518079.1 XM_011518080.1 XM_011518081.2 XM_011518082.2 XM_011518083.2 XM_011518084.2 XM_011518085.1 XM_011518086.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1659036 c.817T>C p.W273R Chr.9: 36633183-36633183 on Build GRCh38
6483675 c.758C>G p.S253C Chr.9: 36633124-36633124 on Build GRCh38
6296500 c.811G>T p.V271F Chr.9: 36633177-36633177 on Build GRCh38
6183739 c.744A>T p.P248P Chr.9: 36633110-36633110 on Build GRCh38
6434399 c.825C>A p.S275R Chr.9: 36633191-36633191 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs891280689 Chr.9: 36633302-36633302 on Build GRCh38
rs1047692436 Chr.9: 36633121-36633121 on Build GRCh38
rs777806003 Chr.9: 36633071-36633072 on Build GRCh38
rs1056733651 Chr.9: 36633027-36633027 on Build GRCh38
rs549358592 Chr.9: 36633114-36633114 on Build GRCh38
rs930874479 Chr.9: 36633043-36633043 on Build GRCh38
rs796296807 Chr.9: 36633128-36633129 on Build GRCh38
rs1030266194 Chr.9: 36633264-36633264 on Build GRCh38
rs766156695 Chr.9: 36633192-36633192 on Build GRCh38
rs756205353 Chr.9: 36633221-36633221 on Build GRCh38
rs146486300 Chr.9: 36633126-36633126 on Build GRCh38
rs917741656 Chr.9: 36633036-36633036 on Build GRCh38
rs7039344 Chr.9: 36633007-36633007 on Build GRCh38
rs566083413 Chr.9: 36633125-36633125 on Build GRCh38
rs768103782 Chr.9: 36633153-36633153 on Build GRCh38
rs996719015 Chr.9: 36633310-36633310 on Build GRCh38
rs529655835 Chr.9: 36633098-36633098 on Build GRCh38
rs371065264 Chr.9: 36633053-36633053 on Build GRCh38
rs997274135 Chr.9: 36633222-36633222 on Build GRCh38
rs557988412 Chr.9: 36633159-36633159 on Build GRCh38
rs934766353 Chr.9: 36633190-36633190 on Build GRCh38
rs765060064 Chr.9: 36633054-36633054 on Build GRCh38
rs543008462 Chr.9: 36632990-36632990 on Build GRCh38
rs762738147 Chr.9: 36633182-36633182 on Build GRCh38
rs1009661976 Chr.9: 36633315-36633315 on Build GRCh38
rs544779873 Chr.9: 36632962-36632962 on Build GRCh38
rs751109036 Chr.9: 36633018-36633018 on Build GRCh38
rs558961126 Chr.9: 36632900-36632900 on Build GRCh38
rs769455465 Chr.9: 36633169-36633169 on Build GRCh38
rs759495589 Chr.9: 36633195-36633195 on Build GRCh38
rs767622049 Chr.9: 36633105-36633105 on Build GRCh38
rs777338925 Chr.9: 36633225-36633226 on Build GRCh38
rs778092292 Chr.9: 36633119-36633119 on Build GRCh38
rs908255796 Chr.9: 36632921-36632921 on Build GRCh38
rs771179956 Chr.9: 36633226-36633226 on Build GRCh38
rs865966244 Chr.9: 36632964-36632964 on Build GRCh38
rs138651367 Chr.9: 36632988-36632988 on Build GRCh38
rs750247449 Chr.9: 36633056-36633056 on Build GRCh38
rs776178978 Chr.9: 36633157-36633157 on Build GRCh38
rs185469521 Chr.9: 36633269-36633269 on Build GRCh38
rs1045775545 Chr.9: 36633144-36633144 on Build GRCh38
rs756061198 Chr.9: 36632886-36632886 on Build GRCh38
rs200387406 Chr.9: 36633095-36633095 on Build GRCh38
rs762684988 Chr.9: 36633084-36633084 on Build GRCh38
rs979378172 Chr.9: 36632973-36632973 on Build GRCh38
rs772791136 Chr.9: 36633174-36633174 on Build GRCh38
rs777917422 Chr.9: 36632928-36632928 on Build GRCh38
rs376375697 Chr.9: 36633051-36633051 on Build GRCh38
rs751443564 Chr.9: 36633102-36633102 on Build GRCh38
rs746375465 Chr.9: 36633152-36633152 on Build GRCh38
rs193187446 Chr.9: 36633207-36633207 on Build GRCh38
rs755078966 Chr.9: 36633104-36633104 on Build GRCh38
rs988333641 Chr.9: 36632867-36632867 on Build GRCh38
rs747428704 Chr.9: 36633191-36633191 on Build GRCh38
rs949997188 Chr.9: 36633138-36633138 on Build GRCh38
rs201211316 Chr.9: 36633124-36633124 on Build GRCh38
rs758392144 Chr.9: 36633117-36633117 on Build GRCh38
rs774211922 Chr.9: 36633194-36633194 on Build GRCh38
rs779634205 Chr.9: 36633148-36633148 on Build GRCh38
rs756637702 Chr.9: 36633046-36633046 on Build GRCh38
rs188337639 Chr.9: 36632914-36632914 on Build GRCh38

View Full Product Details