Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 15511359-15511863 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACE2
Entrez Gene ID
59272
Gene Name
angiotensin I converting enzyme 2
Gene Aliases
ACEH
UniGene
Hs.178098
Gene Chromosome Location
Chr.X: 15494525-15602069 on Build GRCh38
Transcripts
XM_011545551.2 NM_001320866.1 NM_001721.6 NM_203281.2 XM_017029752.1

Gene Symbol
BMX
Entrez Gene ID
660
Gene Name
BMX non-receptor tyrosine kinase
Gene Aliases
ETK, PSCTK2, PSCTK3
UniGene
Hs.495731
Gene Chromosome Location
Chr.X: 15500777-15556529 on Build GRCh38
Transcripts
XM_011545551.2 NM_001320866.1 NM_001721.6 NM_203281.2 XM_017029752.1

SNP Detail

SNP ID Chromosome Location
rs56231264 Chr.X: 15511563-15511563 on Build GRCh38
rs914617893 Chr.X: 15511482-15511482 on Build GRCh38
rs374982760 Chr.X: 15511408-15511408 on Build GRCh38
rs869132740 Chr.X: 15511614-15511615 on Build GRCh38
rs879056556 Chr.X: 15511441-15511441 on Build GRCh38
rs773110270 Chr.X: 15511565-15511565 on Build GRCh38
rs759857171 Chr.X: 15511525-15511526 on Build GRCh38
rs376304899 Chr.X: 15511562-15511562 on Build GRCh38
rs746549419 Chr.X: 15511513-15511513 on Build GRCh38
rs752896501 Chr.X: 15511555-15511555 on Build GRCh38
rs754855100 Chr.X: 15511535-15511535 on Build GRCh38
rs188248881 Chr.X: 15511808-15511808 on Build GRCh38
rs769701179 Chr.X: 15511607-15511607 on Build GRCh38
rs892044633 Chr.X: 15511717-15511717 on Build GRCh38
rs750248462 Chr.X: 15511407-15511407 on Build GRCh38
rs867697129 Chr.X: 15511651-15511651 on Build GRCh38
rs752105094 Chr.X: 15511719-15511719 on Build GRCh38
rs1019973928 Chr.X: 15511612-15511612 on Build GRCh38
rs746299719 Chr.X: 15511811-15511811 on Build GRCh38
rs758070329 Chr.X: 15511725-15511725 on Build GRCh38
rs981332515 Chr.X: 15511779-15511779 on Build GRCh38
rs1005963145 Chr.X: 15511656-15511656 on Build GRCh38
rs1015385610 Chr.X: 15511406-15511406 on Build GRCh38
rs763154649 Chr.X: 15511570-15511571 on Build GRCh38
rs779395852 Chr.X: 15511487-15511487 on Build GRCh38
rs903176633 Chr.X: 15511526-15511526 on Build GRCh38
rs947474924 Chr.X: 15511489-15511489 on Build GRCh38
rs757699086 Chr.X: 15511440-15511440 on Build GRCh38
rs887553811 Chr.X: 15511632-15511632 on Build GRCh38
rs991304519 Chr.X: 15511835-15511835 on Build GRCh38
rs753747603 Chr.X: 15511533-15511533 on Build GRCh38
rs944278540 Chr.X: 15511631-15511631 on Build GRCh38
rs200465814 Chr.X: 15511429-15511429 on Build GRCh38
rs112128562 Chr.X: 15511611-15511611 on Build GRCh38
rs373097386 Chr.X: 15511537-15511537 on Build GRCh38
rs996986806 Chr.X: 15511599-15511599 on Build GRCh38
rs765389495 Chr.X: 15511534-15511534 on Build GRCh38
rs778413330 Chr.X: 15511701-15511701 on Build GRCh38
rs961698581 Chr.X: 15511726-15511726 on Build GRCh38
rs113096962 Chr.X: 15511603-15511603 on Build GRCh38
rs150627254 Chr.X: 15511488-15511488 on Build GRCh38
rs373085295 Chr.X: 15511405-15511405 on Build GRCh38
rs767655164 Chr.X: 15511390-15511390 on Build GRCh38
rs1042139244 Chr.X: 15511514-15511514 on Build GRCh38
rs369144737 Chr.X: 15511476-15511476 on Build GRCh38
rs141827642 Chr.X: 15511457-15511457 on Build GRCh38
rs754195845 Chr.X: 15511430-15511430 on Build GRCh38

View Full Product Details