Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 16844396-16844936 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RBBP7
Entrez Gene ID
5931
Gene Name
RB binding protein 7, chromatin remodeling factor
Gene Aliases
RbAp46
UniGene
Hs.495755
Gene Chromosome Location
Chr.X: 16844650-16870411 on Build GRCh38
Transcripts
NM_001198719.1 NM_002893.3 NM_001168683.1 NM_018360.2 XM_017029631.1

Gene Symbol
TXLNG
Entrez Gene ID
55787
Gene Name
taxilin gamma
Gene Aliases
CXorf15, ELRG, FIAT, LSR5, TXLNGX
UniGene
Hs.555961
Gene Chromosome Location
Chr.X: 16786432-16844519 on Build GRCh38
Transcripts
NM_001198719.1 NM_002893.3 NM_001168683.1 NM_018360.2 XM_017029631.1

SNP Detail

SNP ID Chromosome Location
rs886109673 Chr.X: 16844489-16844489 on Build GRCh38
rs397840029 Chr.X: 16844625-16844625 on Build GRCh38
rs11542192 Chr.X: 16844783-16844783 on Build GRCh38
rs3214937 Chr.X: 16844617-16844617 on Build GRCh38
rs930827918 Chr.X: 16844818-16844818 on Build GRCh38
rs200947083 Chr.X: 16844617-16844617 on Build GRCh38
rs756188425 Chr.X: 16844894-16844894 on Build GRCh38
rs199744009 Chr.X: 16844752-16844752 on Build GRCh38
rs185289966 Chr.X: 16844596-16844596 on Build GRCh38
rs977808611 Chr.X: 16844731-16844731 on Build GRCh38
rs769447719 Chr.X: 16844881-16844881 on Build GRCh38
rs900777575 Chr.X: 16844747-16844747 on Build GRCh38
rs756438498 Chr.X: 16844616-16844617 on Build GRCh38
rs907989245 Chr.X: 16844877-16844877 on Build GRCh38
rs189795519 Chr.X: 16844887-16844887 on Build GRCh38
rs750937673 Chr.X: 16844745-16844746 on Build GRCh38
rs1047391676 Chr.X: 16844465-16844465 on Build GRCh38
rs954813960 Chr.X: 16844832-16844833 on Build GRCh38
rs770770607 Chr.X: 16844903-16844903 on Build GRCh38
rs975397381 Chr.X: 16844751-16844751 on Build GRCh38
rs772617352 Chr.X: 16844674-16844674 on Build GRCh38
rs1022065112 Chr.X: 16844638-16844638 on Build GRCh38
rs752937466 Chr.X: 16844604-16844604 on Build GRCh38
rs759649052 Chr.X: 16844439-16844439 on Build GRCh38
rs757760467 Chr.X: 16844623-16844623 on Build GRCh38
rs1033559386 Chr.X: 16844508-16844508 on Build GRCh38
rs1010709712 Chr.X: 16844635-16844635 on Build GRCh38
rs779457169 Chr.X: 16844681-16844681 on Build GRCh38
rs1003864005 Chr.X: 16844480-16844480 on Build GRCh38
rs966352898 Chr.X: 16844749-16844749 on Build GRCh38
rs1016542505 Chr.X: 16844711-16844711 on Build GRCh38
rs747785211 Chr.X: 16844880-16844880 on Build GRCh38
rs775802793 Chr.X: 16844851-16844851 on Build GRCh38
rs780598436 Chr.X: 16844796-16844796 on Build GRCh38
rs1033366379 Chr.X: 16844625-16844625 on Build GRCh38
rs1006443401 Chr.X: 16844525-16844526 on Build GRCh38
rs938466527 Chr.X: 16844428-16844429 on Build GRCh38
rs754256821 Chr.X: 16844617-16844618 on Build GRCh38
rs911534923 Chr.X: 16844746-16844746 on Build GRCh38

View Full Product Details