Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 17687457-17687952 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NHS
Entrez Gene ID
4810
Gene Name
NHS actin remodeling regulator
Gene Aliases
CTRCT40, CXN, SCML1
UniGene
Hs.201623
Gene Chromosome Location
Chr.X: 17375420-17735994 on Build GRCh38
Transcripts
NM_001136024.3 NM_001291867.1 NM_001291868.1 NM_198270.3

SNP Detail

SNP ID Chromosome Location
rs772563986 Chr.X: 17687811-17687811 on Build GRCh38
rs756659708 Chr.X: 17687764-17687764 on Build GRCh38
rs773424232 Chr.X: 17687647-17687647 on Build GRCh38
rs200952266 Chr.X: 17687794-17687794 on Build GRCh38
rs770771757 Chr.X: 17687894-17687895 on Build GRCh38
rs999015963 Chr.X: 17687569-17687569 on Build GRCh38
rs894438729 Chr.X: 17687511-17687511 on Build GRCh38
rs766403255 Chr.X: 17687849-17687849 on Build GRCh38
rs760065199 Chr.X: 17687737-17687737 on Build GRCh38
rs758384516 Chr.X: 17687778-17687778 on Build GRCh38
rs771955679 Chr.X: 17687666-17687666 on Build GRCh38
rs773546351 Chr.X: 17687720-17687720 on Build GRCh38
rs764528482 Chr.X: 17687817-17687817 on Build GRCh38
rs919155040 Chr.X: 17687534-17687534 on Build GRCh38
rs749420963 Chr.X: 17687867-17687867 on Build GRCh38
rs779172578 Chr.X: 17687900-17687900 on Build GRCh38
rs747851640 Chr.X: 17687506-17687506 on Build GRCh38
rs768063132 Chr.X: 17687738-17687738 on Build GRCh38
rs771305748 Chr.X: 17687730-17687730 on Build GRCh38
rs140369750 Chr.X: 17687787-17687787 on Build GRCh38
rs999129147 Chr.X: 17687889-17687889 on Build GRCh38
rs144769657 Chr.X: 17687744-17687744 on Build GRCh38
rs780014924 Chr.X: 17687676-17687676 on Build GRCh38
rs745989162 Chr.X: 17687808-17687808 on Build GRCh38
rs150379955 Chr.X: 17687822-17687822 on Build GRCh38
rs999078013 Chr.X: 17687650-17687650 on Build GRCh38
rs187750900 Chr.X: 17687592-17687592 on Build GRCh38
rs774771955 Chr.X: 17687733-17687733 on Build GRCh38
rs750334033 Chr.X: 17687776-17687776 on Build GRCh38
rs397759640 Chr.X: 17687732-17687733 on Build GRCh38
rs760893537 Chr.X: 17687813-17687813 on Build GRCh38
rs967969953 Chr.X: 17687646-17687646 on Build GRCh38
rs1021845408 Chr.X: 17687540-17687540 on Build GRCh38
rs200300826 Chr.X: 17687807-17687807 on Build GRCh38
rs371785134 Chr.X: 17687766-17687766 on Build GRCh38
rs5901624 Chr.X: 17687730-17687731 on Build GRCh38
rs1049024471 Chr.X: 17687541-17687541 on Build GRCh38
rs78153843 Chr.X: 17687864-17687864 on Build GRCh38
rs1028084772 Chr.X: 17687628-17687628 on Build GRCh38
rs138104885 Chr.X: 17687842-17687842 on Build GRCh38
rs751527423 Chr.X: 17687860-17687860 on Build GRCh38
rs875989805 Chr.X: 17687870-17687870 on Build GRCh38
rs776035809 Chr.X: 17687812-17687812 on Build GRCh38
rs757440505 Chr.X: 17687868-17687868 on Build GRCh38
rs769730155 Chr.X: 17687724-17687725 on Build GRCh38
rs901306368 Chr.X: 17687917-17687917 on Build GRCh38
rs146835111 Chr.X: 17687925-17687925 on Build GRCh38
rs146456925 Chr.X: 17687878-17687878 on Build GRCh38
rs775351254 Chr.X: 17687729-17687730 on Build GRCh38
rs775797533 Chr.X: 17687913-17687913 on Build GRCh38
rs746772731 Chr.X: 17687786-17687786 on Build GRCh38
rs182673124 Chr.X: 17687512-17687512 on Build GRCh38
rs1031825068 Chr.X: 17687905-17687905 on Build GRCh38
rs201472570 Chr.X: 17687729-17687729 on Build GRCh38
rs866091099 Chr.X: 17687789-17687789 on Build GRCh38
rs1039809530 Chr.X: 17687581-17687581 on Build GRCh38
rs869108101 Chr.X: 17687733-17687734 on Build GRCh38
rs746181221 Chr.X: 17687623-17687623 on Build GRCh38
rs965137308 Chr.X: 17687594-17687594 on Build GRCh38
rs17312492 Chr.X: 17687565-17687565 on Build GRCh38
rs763641131 Chr.X: 17687754-17687754 on Build GRCh38
rs888406866 Chr.X: 17687559-17687559 on Build GRCh38
rs146725938 Chr.X: 17687763-17687763 on Build GRCh38
rs761169168 Chr.X: 17687748-17687748 on Build GRCh38
rs754663070 Chr.X: 17687863-17687863 on Build GRCh38
rs763102874 Chr.X: 17687681-17687681 on Build GRCh38
rs780176861 Chr.X: 17687783-17687783 on Build GRCh38
rs769872475 Chr.X: 17687698-17687698 on Build GRCh38
rs794727314 Chr.X: 17687836-17687836 on Build GRCh38
rs904773347 Chr.X: 17687866-17687866 on Build GRCh38
rs745877435 Chr.X: 17687693-17687693 on Build GRCh38
rs749528520 Chr.X: 17687790-17687790 on Build GRCh38
rs201401939 Chr.X: 17687820-17687820 on Build GRCh38
rs762927798 Chr.X: 17687721-17687721 on Build GRCh38
rs765028709 Chr.X: 17687837-17687837 on Build GRCh38
rs758044602 Chr.X: 17687846-17687846 on Build GRCh38
rs769910916 Chr.X: 17687600-17687600 on Build GRCh38
rs367740284 Chr.X: 17687924-17687924 on Build GRCh38
rs747585263 Chr.X: 17687915-17687915 on Build GRCh38
rs972522687 Chr.X: 17687743-17687743 on Build GRCh38
rs753235773 Chr.X: 17687907-17687907 on Build GRCh38
rs1012940111 Chr.X: 17687533-17687533 on Build GRCh38
rs777799752 Chr.X: 17687788-17687788 on Build GRCh38

View Full Product Details