Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 27747859-27748358 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCAF8L2
Entrez Gene ID
347442
Gene Name
DDB1 and CUL4 associated factor 8 like 2
Gene Aliases
WDR42C
UniGene
Hs.720885
Gene Chromosome Location
Chr.X: 27746809-27748821 on Build GRCh38
Transcripts
NM_001136533.1

SNP Detail

SNP ID Chromosome Location
rs762061938 Chr.X: 27748333-27748333 on Build GRCh38
rs769551706 Chr.X: 27748256-27748256 on Build GRCh38
rs966102479 Chr.X: 27747897-27747897 on Build GRCh38
rs777879257 Chr.X: 27748015-27748015 on Build GRCh38
rs976570296 Chr.X: 27748244-27748244 on Build GRCh38
rs5926895 Chr.X: 27747928-27747928 on Build GRCh38
rs751379029 Chr.X: 27748011-27748011 on Build GRCh38
rs368376110 Chr.X: 27748122-27748122 on Build GRCh38
rs750469981 Chr.X: 27748220-27748220 on Build GRCh38
rs761465771 Chr.X: 27748115-27748115 on Build GRCh38
rs372484466 Chr.X: 27747887-27747887 on Build GRCh38
rs765849497 Chr.X: 27748320-27748320 on Build GRCh38
rs1029038901 Chr.X: 27748249-27748249 on Build GRCh38
rs772892127 Chr.X: 27748266-27748266 on Build GRCh38
rs1811385 Chr.X: 27747972-27747972 on Build GRCh38
rs748841788 Chr.X: 27748278-27748278 on Build GRCh38
rs61730883 Chr.X: 27748125-27748125 on Build GRCh38
rs756400313 Chr.X: 27748012-27748012 on Build GRCh38
rs774285351 Chr.X: 27747991-27747991 on Build GRCh38
rs776310062 Chr.X: 27747942-27747942 on Build GRCh38
rs77842814 Chr.X: 27748322-27748322 on Build GRCh38
rs766389302 Chr.X: 27748158-27748158 on Build GRCh38
rs112456497 Chr.X: 27747915-27747915 on Build GRCh38
rs966189526 Chr.X: 27748228-27748228 on Build GRCh38
rs775333139 Chr.X: 27748328-27748328 on Build GRCh38
rs766335953 Chr.X: 27747996-27747996 on Build GRCh38
rs767272705 Chr.X: 27748191-27748191 on Build GRCh38
rs746306525 Chr.X: 27747909-27747909 on Build GRCh38
rs754033019 Chr.X: 27748020-27748020 on Build GRCh38
rs781702266 Chr.X: 27748121-27748121 on Build GRCh38
rs772488367 Chr.X: 27747911-27747911 on Build GRCh38
rs78540534 Chr.X: 27747929-27747929 on Build GRCh38
rs61730884 Chr.X: 27748287-27748287 on Build GRCh38
rs746428763 Chr.X: 27748062-27748062 on Build GRCh38
rs372427910 Chr.X: 27747890-27747890 on Build GRCh38
rs868486185 Chr.X: 27748160-27748160 on Build GRCh38
rs747493517 Chr.X: 27748227-27748227 on Build GRCh38
rs369460010 Chr.X: 27748028-27748028 on Build GRCh38
rs774851804 Chr.X: 27747906-27747906 on Build GRCh38
rs768160600 Chr.X: 27747930-27747930 on Build GRCh38
rs780562926 Chr.X: 27748222-27748222 on Build GRCh38
rs972232899 Chr.X: 27748305-27748305 on Build GRCh38
rs1011873595 Chr.X: 27748155-27748155 on Build GRCh38
rs111559275 Chr.X: 27747923-27747923 on Build GRCh38
rs764785756 Chr.X: 27747969-27747969 on Build GRCh38
rs761253838 Chr.X: 27747950-27747950 on Build GRCh38
rs770506122 Chr.X: 27748280-27748280 on Build GRCh38
rs921920331 Chr.X: 27747918-27747918 on Build GRCh38
rs747363358 Chr.X: 27748103-27748103 on Build GRCh38
rs185264593 Chr.X: 27748253-27748253 on Build GRCh38
rs763034072 Chr.X: 27748138-27748138 on Build GRCh38
rs1056165880 Chr.X: 27748136-27748136 on Build GRCh38
rs750395244 Chr.X: 27747988-27747988 on Build GRCh38
rs909436093 Chr.X: 27748088-27748088 on Build GRCh38
rs761676142 Chr.X: 27748039-27748039 on Build GRCh38
rs775815150 Chr.X: 27747927-27747927 on Build GRCh38
rs754070602 Chr.X: 27748192-27748192 on Build GRCh38
rs780433932 Chr.X: 27748070-27748070 on Build GRCh38
rs779012547 Chr.X: 27748213-27748213 on Build GRCh38
rs769018767 Chr.X: 27748107-27748107 on Build GRCh38
rs759260989 Chr.X: 27748177-27748177 on Build GRCh38
rs757287423 Chr.X: 27748024-27748024 on Build GRCh38
rs751476265 Chr.X: 27748176-27748176 on Build GRCh38
rs772541793 Chr.X: 27748068-27748068 on Build GRCh38
rs757488857 Chr.X: 27748206-27748206 on Build GRCh38
rs777958909 Chr.X: 27747885-27747885 on Build GRCh38
rs759509987 Chr.X: 27748313-27748313 on Build GRCh38
rs776165375 Chr.X: 27748109-27748109 on Build GRCh38

View Full Product Details