Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 37801023-37801520 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYBB
Entrez Gene ID
1536
Gene Name
cytochrome b-245 beta chain
Gene Aliases
AMCBX2, CGD, GP91-1, GP91-PHOX, GP91PHOX, IMD34, NOX2, p91-PHOX
UniGene
Hs.292356
Gene Chromosome Location
Chr.X: 37780017-37813461 on Build GRCh38
Transcripts
NM_000397.3

SNP Detail

SNP ID Chromosome Location
rs782770664 Chr.X: 37801130-37801130 on Build GRCh38
rs1055865879 Chr.X: 37801363-37801363 on Build GRCh38
rs782787406 Chr.X: 37801064-37801064 on Build GRCh38
rs782029265 Chr.X: 37801413-37801413 on Build GRCh38
rs782105194 Chr.X: 37801112-37801112 on Build GRCh38
rs35670962 Chr.X: 37801175-37801175 on Build GRCh38
rs886039337 Chr.X: 37801319-37801319 on Build GRCh38
rs782610130 Chr.X: 37801268-37801268 on Build GRCh38
rs782685319 Chr.X: 37801212-37801212 on Build GRCh38
rs377381024 Chr.X: 37801368-37801368 on Build GRCh38
rs782206726 Chr.X: 37801282-37801282 on Build GRCh38
rs189505035 Chr.X: 37801101-37801101 on Build GRCh38
rs781880651 Chr.X: 37801205-37801205 on Build GRCh38
rs782383208 Chr.X: 37801300-37801300 on Build GRCh38
rs963359162 Chr.X: 37801073-37801073 on Build GRCh38
rs782402308 Chr.X: 37801311-37801311 on Build GRCh38
rs912006447 Chr.X: 37801177-37801177 on Build GRCh38
rs1015253648 Chr.X: 37801055-37801055 on Build GRCh38
rs781930966 Chr.X: 37801352-37801352 on Build GRCh38
rs781948097 Chr.X: 37801160-37801160 on Build GRCh38
rs946038531 Chr.X: 37801191-37801191 on Build GRCh38
rs906330192 Chr.X: 37801464-37801464 on Build GRCh38
rs781983828 Chr.X: 37801304-37801304 on Build GRCh38
rs977485921 Chr.X: 37801223-37801223 on Build GRCh38
rs201609728 Chr.X: 37801356-37801356 on Build GRCh38
rs782282539 Chr.X: 37801214-37801214 on Build GRCh38
rs782293433 Chr.X: 37801306-37801306 on Build GRCh38
rs1028831270 Chr.X: 37801109-37801109 on Build GRCh38
rs950248707 Chr.X: 37801394-37801394 on Build GRCh38
rs782506166 Chr.X: 37801208-37801208 on Build GRCh38
rs182092761 Chr.X: 37801239-37801239 on Build GRCh38
rs782767612 Chr.X: 37801398-37801398 on Build GRCh38
rs187079945 Chr.X: 37801393-37801393 on Build GRCh38
rs1046112471 Chr.X: 37801433-37801433 on Build GRCh38
rs546163876 Chr.X: 37801382-37801382 on Build GRCh38

View Full Product Details