Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 46500236-46500733 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF674
Entrez Gene ID
641339
Gene Name
zinc finger protein 674
Gene Aliases
MRX92, ZNF673B
UniGene
Hs.636105
Gene Chromosome Location
Chr.X: 46497725-46545459 on Build GRCh38
Transcripts
NM_001039891.2 NM_001146291.1 NM_001190417.1 XM_011543941.2 XM_011543943.2 XM_011543944.2 XM_011543945.2 XM_017029728.1 XM_017029729.1 XM_017029730.1 XM_017029731.1 XM_017029732.1 XM_017029733.1

SNP Detail

SNP ID Chromosome Location
rs773266198 Chr.X: 46500653-46500653 on Build GRCh38
rs778745042 Chr.X: 46500691-46500691 on Build GRCh38
rs920432447 Chr.X: 46500271-46500271 on Build GRCh38
rs776820356 Chr.X: 46500319-46500319 on Build GRCh38
rs767120139 Chr.X: 46500445-46500445 on Build GRCh38
rs377460705 Chr.X: 46500576-46500576 on Build GRCh38
rs376244972 Chr.X: 46500624-46500624 on Build GRCh38
rs374577863 Chr.X: 46500572-46500572 on Build GRCh38
rs756969719 Chr.X: 46500441-46500441 on Build GRCh38
rs748331751 Chr.X: 46500524-46500524 on Build GRCh38
rs768828974 Chr.X: 46500300-46500300 on Build GRCh38
rs180754888 Chr.X: 46500505-46500505 on Build GRCh38
rs775076231 Chr.X: 46500705-46500705 on Build GRCh38
rs370285911 Chr.X: 46500603-46500603 on Build GRCh38
rs747988366 Chr.X: 46500324-46500324 on Build GRCh38
rs771665182 Chr.X: 46500564-46500564 on Build GRCh38
rs755063842 Chr.X: 46500502-46500502 on Build GRCh38
rs758505232 Chr.X: 46500402-46500402 on Build GRCh38
rs1027923825 Chr.X: 46500376-46500376 on Build GRCh38
rs777476294 Chr.X: 46500548-46500548 on Build GRCh38
rs762296664 Chr.X: 46500450-46500450 on Build GRCh38
rs61730637 Chr.X: 46500561-46500561 on Build GRCh38
rs750035369 Chr.X: 46500320-46500320 on Build GRCh38
rs780611116 Chr.X: 46500488-46500488 on Build GRCh38
rs752919527 Chr.X: 46500652-46500652 on Build GRCh38
rs769547318 Chr.X: 46500346-46500346 on Build GRCh38
rs925874735 Chr.X: 46500604-46500604 on Build GRCh38
rs888092311 Chr.X: 46500446-46500446 on Build GRCh38
rs763363600 Chr.X: 46500598-46500598 on Build GRCh38
rs142411452 Chr.X: 46500464-46500464 on Build GRCh38
rs908558783 Chr.X: 46500681-46500681 on Build GRCh38
rs766844138 Chr.X: 46500599-46500599 on Build GRCh38
rs771724523 Chr.X: 46500699-46500699 on Build GRCh38
rs758530476 Chr.X: 46500288-46500288 on Build GRCh38
rs751648956 Chr.X: 46500500-46500500 on Build GRCh38
rs778934477 Chr.X: 46500365-46500378 on Build GRCh38
rs773799168 Chr.X: 46500438-46500438 on Build GRCh38
rs1055550584 Chr.X: 46500327-46500327 on Build GRCh38
rs587780526 Chr.X: 46500372-46500372 on Build GRCh38
rs752856034 Chr.X: 46500386-46500387 on Build GRCh38
rs750888011 Chr.X: 46500597-46500597 on Build GRCh38
rs200552019 Chr.X: 46500294-46500294 on Build GRCh38
rs189178099 Chr.X: 46500281-46500281 on Build GRCh38
rs777957299 Chr.X: 46500655-46500655 on Build GRCh38
rs868197297 Chr.X: 46500339-46500339 on Build GRCh38
rs781188926 Chr.X: 46500636-46500636 on Build GRCh38
rs749081744 Chr.X: 46500682-46500682 on Build GRCh38
rs544778598 Chr.X: 46500478-46500478 on Build GRCh38
rs199534410 Chr.X: 46500411-46500411 on Build GRCh38
rs750983120 Chr.X: 46500473-46500473 on Build GRCh38
rs982843155 Chr.X: 46500656-46500656 on Build GRCh38
rs183994849 Chr.X: 46500265-46500265 on Build GRCh38
rs187237351 Chr.X: 46500690-46500690 on Build GRCh38
rs778073001 Chr.X: 46500598-46500599 on Build GRCh38
rs370874445 Chr.X: 46500356-46500356 on Build GRCh38
rs752364265 Chr.X: 46500448-46500448 on Build GRCh38
rs765705003 Chr.X: 46500593-46500593 on Build GRCh38
rs902832661 Chr.X: 46500546-46500546 on Build GRCh38
rs762817626 Chr.X: 46500386-46500386 on Build GRCh38
rs895603983 Chr.X: 46500329-46500329 on Build GRCh38
rs769589423 Chr.X: 46500543-46500543 on Build GRCh38
rs773153851 Chr.X: 46500355-46500355 on Build GRCh38
rs779944408 Chr.X: 46500293-46500293 on Build GRCh38
rs755764590 Chr.X: 46500383-46500383 on Build GRCh38
rs942671211 Chr.X: 46500310-46500310 on Build GRCh38
rs746340164 Chr.X: 46500282-46500282 on Build GRCh38
rs745766277 Chr.X: 46500696-46500696 on Build GRCh38
rs749104969 Chr.X: 46500549-46500549 on Build GRCh38
rs377291882 Chr.X: 46500560-46500560 on Build GRCh38
rs201399961 Chr.X: 46500540-46500540 on Build GRCh38

View Full Product Details