Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 49166799-49167305 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAGIX
Entrez Gene ID
79917
Gene Name
MAGI family member, X-linked
Gene Aliases
JM10, PDZX
UniGene
Hs.193170
Gene Chromosome Location
Chr.X: 49162569-49168157 on Build GRCh38
Transcripts
NM_001099681.2 NM_001099682.2 NM_024859.3

SNP Detail

SNP ID Chromosome Location
rs782423984 Chr.X: 49167234-49167234 on Build GRCh38
rs897857774 Chr.X: 49167002-49167002 on Build GRCh38
rs1025079272 Chr.X: 49167042-49167042 on Build GRCh38
rs909758366 Chr.X: 49167224-49167224 on Build GRCh38
rs970860555 Chr.X: 49167043-49167043 on Build GRCh38
rs1049602779 Chr.X: 49166866-49166866 on Build GRCh38
rs922427322 Chr.X: 49166845-49166845 on Build GRCh38
rs1021031895 Chr.X: 49167125-49167125 on Build GRCh38
rs932451882 Chr.X: 49167201-49167201 on Build GRCh38
rs782234255 Chr.X: 49167173-49167173 on Build GRCh38
rs1038104261 Chr.X: 49166973-49166973 on Build GRCh38
rs370026551 Chr.X: 49167246-49167246 on Build GRCh38
rs976890563 Chr.X: 49167162-49167162 on Build GRCh38
rs888371256 Chr.X: 49166914-49166914 on Build GRCh38
rs190285403 Chr.X: 49167056-49167056 on Build GRCh38
rs782148523 Chr.X: 49167124-49167124 on Build GRCh38
rs781929000 Chr.X: 49167210-49167210 on Build GRCh38
rs782195261 Chr.X: 49167104-49167104 on Build GRCh38
rs782513463 Chr.X: 49166920-49166920 on Build GRCh38
rs782007022 Chr.X: 49166916-49166916 on Build GRCh38
rs150086636 Chr.X: 49167029-49167029 on Build GRCh38
rs782567408 Chr.X: 49167101-49167101 on Build GRCh38
rs985262333 Chr.X: 49167209-49167209 on Build GRCh38
rs782821147 Chr.X: 49167138-49167138 on Build GRCh38
rs929339747 Chr.X: 49167283-49167283 on Build GRCh38
rs1036977757 Chr.X: 49167262-49167262 on Build GRCh38
rs55999418 Chr.X: 49167117-49167117 on Build GRCh38
rs1033899318 Chr.X: 49167116-49167116 on Build GRCh38
rs781963036 Chr.X: 49167122-49167122 on Build GRCh38
rs782348330 Chr.X: 49166824-49166824 on Build GRCh38
rs376721276 Chr.X: 49167085-49167085 on Build GRCh38
rs782068870 Chr.X: 49167276-49167276 on Build GRCh38

View Full Product Details