Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 102601386-102601896 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARMCX5
Entrez Gene ID
64860
Gene Name
armadillo repeat containing, X-linked 5
Gene Aliases
GASP5
UniGene
Hs.522729
Gene Chromosome Location
Chr.X: 102599168-102604159 on Build GRCh38
Transcripts
NM_001168478.1 NM_001168479.1 NM_001168480.1 NM_001168482.1 NM_001168485.1 NM_022838.3 NM_001199818.1

Gene Symbol
ARMCX5-GPRASP2
Entrez Gene ID
100528062
Gene Name
ARMCX5-GPRASP2 readthrough
Gene Aliases
-
UniGene
Hs.522729
Gene Chromosome Location
Chr.X: 102599348-102717733 on Build GRCh38
Transcripts
NM_001168478.1 NM_001168479.1 NM_001168480.1 NM_001168482.1 NM_001168485.1 NM_022838.3 NM_001199818.1

SNP Detail

SNP ID Chromosome Location
rs999822452 Chr.X: 102601627-102601627 on Build GRCh38
rs886885043 Chr.X: 102601534-102601534 on Build GRCh38
rs929475545 Chr.X: 102601873-102601873 on Build GRCh38
rs910587975 Chr.X: 102601754-102601754 on Build GRCh38
rs751447954 Chr.X: 102601433-102601433 on Build GRCh38
rs918633690 Chr.X: 102601825-102601825 on Build GRCh38
rs2379211 Chr.X: 102601790-102601790 on Build GRCh38
rs780927012 Chr.X: 102601768-102601769 on Build GRCh38
rs1035352140 Chr.X: 102601679-102601679 on Build GRCh38
rs988393425 Chr.X: 102601694-102601694 on Build GRCh38
rs929480897 Chr.X: 102601418-102601418 on Build GRCh38
rs1005203863 Chr.X: 102601608-102601608 on Build GRCh38
rs1012340998 Chr.X: 102601739-102601739 on Build GRCh38
rs752443504 Chr.X: 102601762-102601762 on Build GRCh38
rs962730698 Chr.X: 102601752-102601752 on Build GRCh38
rs367693400 Chr.X: 102601729-102601729 on Build GRCh38
rs951440634 Chr.X: 102601855-102601855 on Build GRCh38
rs183954945 Chr.X: 102601623-102601623 on Build GRCh38
rs968129893 Chr.X: 102601738-102601738 on Build GRCh38
rs568255652 Chr.X: 102601859-102601859 on Build GRCh38
rs769300984 Chr.X: 102601847-102601847 on Build GRCh38
rs1016648841 Chr.X: 102601617-102601617 on Build GRCh38
rs1043362969 Chr.X: 102601610-102601610 on Build GRCh38
rs41310699 Chr.X: 102601641-102601641 on Build GRCh38
rs960324 Chr.X: 102601823-102601823 on Build GRCh38
rs757681358 Chr.X: 102601443-102601446 on Build GRCh38
rs923529474 Chr.X: 102601849-102601849 on Build GRCh38
rs780981996 Chr.X: 102601683-102601683 on Build GRCh38
rs979401291 Chr.X: 102601747-102601747 on Build GRCh38
rs1002086684 Chr.X: 102601669-102601669 on Build GRCh38

View Full Product Details