Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 102714037-102714540 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARMCX5-GPRASP2
Entrez Gene ID
100528062
Gene Name
ARMCX5-GPRASP2 readthrough
Gene Aliases
-
UniGene
Hs.522729
Gene Chromosome Location
Chr.X: 102599348-102717733 on Build GRCh38
Transcripts
NM_001199818.1 NM_001004051.3 NM_001184874.2 NM_001184875.2 NM_001184876.2 NM_138437.5

Gene Symbol
GPRASP2
Entrez Gene ID
114928
Gene Name
G protein-coupled receptor associated sorting protein 2
Gene Aliases
GASP2
UniGene
Hs.522729
Gene Chromosome Location
Chr.X: 102712176-102717733 on Build GRCh38
Transcripts
NM_001199818.1 NM_001004051.3 NM_001184874.2 NM_001184875.2 NM_001184876.2 NM_138437.5

SNP Detail

SNP ID Chromosome Location
rs1054440458 Chr.X: 102714295-102714295 on Build GRCh38
rs761258010 Chr.X: 102714396-102714396 on Build GRCh38
rs143017019 Chr.X: 102714218-102714218 on Build GRCh38
rs747385137 Chr.X: 102714217-102714217 on Build GRCh38
rs1016619613 Chr.X: 102714106-102714106 on Build GRCh38
rs777322256 Chr.X: 102714220-102714220 on Build GRCh38
rs2073118 Chr.X: 102714234-102714234 on Build GRCh38
rs936094346 Chr.X: 102714281-102714281 on Build GRCh38
rs1045719753 Chr.X: 102714355-102714355 on Build GRCh38
rs772575308 Chr.X: 102714359-102714359 on Build GRCh38
rs774838789 Chr.X: 102714491-102714491 on Build GRCh38
rs780472647 Chr.X: 102714195-102714195 on Build GRCh38
rs191969773 Chr.X: 102714468-102714468 on Build GRCh38
rs985630473 Chr.X: 102714265-102714265 on Build GRCh38
rs1054969646 Chr.X: 102714383-102714383 on Build GRCh38
rs58847439 Chr.X: 102714376-102714376 on Build GRCh38
rs148285111 Chr.X: 102714280-102714280 on Build GRCh38
rs576701889 Chr.X: 102714072-102714072 on Build GRCh38
rs58893995 Chr.X: 102714438-102714438 on Build GRCh38
rs911349179 Chr.X: 102714279-102714279 on Build GRCh38
rs774063771 Chr.X: 102714259-102714259 on Build GRCh38
rs6621548 Chr.X: 102714473-102714473 on Build GRCh38
rs983834700 Chr.X: 102714092-102714092 on Build GRCh38
rs946063124 Chr.X: 102714323-102714323 on Build GRCh38
rs973030557 Chr.X: 102714118-102714118 on Build GRCh38
rs748472018 Chr.X: 102714330-102714330 on Build GRCh38
rs775513998 Chr.X: 102714428-102714428 on Build GRCh38
rs188641020 Chr.X: 102714151-102714151 on Build GRCh38
rs762149837 Chr.X: 102714484-102714484 on Build GRCh38
rs891816199 Chr.X: 102714310-102714310 on Build GRCh38
rs779987375 Chr.X: 102714073-102714073 on Build GRCh38
rs373096911 Chr.X: 102714120-102714120 on Build GRCh38
rs961138905 Chr.X: 102714481-102714481 on Build GRCh38
rs1029326542 Chr.X: 102714515-102714515 on Build GRCh38
rs749011998 Chr.X: 102714221-102714221 on Build GRCh38
rs111896745 Chr.X: 102714474-102714474 on Build GRCh38
rs376702099 Chr.X: 102714116-102714116 on Build GRCh38
rs779222910 Chr.X: 102714297-102714297 on Build GRCh38
rs1003461020 Chr.X: 102714374-102714374 on Build GRCh38
rs768725155 Chr.X: 102714258-102714258 on Build GRCh38

View Full Product Details