Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 104112325-104112823 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC25A53
Entrez Gene ID
401612
Gene Name
solute carrier family 25 member 53
Gene Aliases
MCART6
UniGene
Hs.660022
Gene Chromosome Location
Chr.X: 104099214-104157027 on Build GRCh38
Transcripts
NM_001012755.4 XM_005262129.4 XM_011530952.2 XM_011530953.2 XM_017029514.1 XM_017029515.1 XM_017029516.1 NM_001143978.2 NR_026694.2 XM_011531012.2

Gene Symbol
ZCCHC18
Entrez Gene ID
644353
Gene Name
zinc finger CCHC-type containing 18
Gene Aliases
PNMA7B, SIZN2
UniGene
Hs.648338
Gene Chromosome Location
Chr.X: 104112416-104121330 on Build GRCh38
Transcripts
NM_001012755.4 XM_005262129.4 XM_011530952.2 XM_011530953.2 XM_017029514.1 XM_017029515.1 XM_017029516.1 NM_001143978.2 NR_026694.2 XM_011531012.2

SNP Detail

SNP ID Chromosome Location
rs781827550 Chr.X: 104112555-104112555 on Build GRCh38
rs782622417 Chr.X: 104112524-104112524 on Build GRCh38
rs781883366 Chr.X: 104112461-104112461 on Build GRCh38
rs188920726 Chr.X: 104112599-104112599 on Build GRCh38
rs183928076 Chr.X: 104112574-104112574 on Build GRCh38
rs782440597 Chr.X: 104112518-104112518 on Build GRCh38
rs782800709 Chr.X: 104112417-104112417 on Build GRCh38
rs375512265 Chr.X: 104112627-104112627 on Build GRCh38
rs782511183 Chr.X: 104112347-104112347 on Build GRCh38

View Full Product Details