Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 109658981-109659513 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACSL4
Entrez Gene ID
2182
Gene Name
acyl-CoA synthetase long-chain family member 4
Gene Aliases
ACS4, FACL4, LACS4, MRX63, MRX68
UniGene
Hs.268785
Gene Chromosome Location
Chr.X: 109641330-109733403 on Build GRCh38
Transcripts
NM_001318509.1 NM_001318510.1 NM_004458.2 NM_022977.2 XM_005262109.2 XM_006724635.2 XM_011530888.2 XM_011530889.2

SNP Detail

SNP ID Chromosome Location
rs921638453 Chr.X: 109659310-109659310 on Build GRCh38
rs567273157 Chr.X: 109659131-109659131 on Build GRCh38
rs768577722 Chr.X: 109659484-109659484 on Build GRCh38
rs756641850 Chr.X: 109659405-109659405 on Build GRCh38
rs988110707 Chr.X: 109659385-109659385 on Build GRCh38
rs952250039 Chr.X: 109659177-109659177 on Build GRCh38
rs116830999 Chr.X: 109659343-109659343 on Build GRCh38
rs749815932 Chr.X: 109659427-109659427 on Build GRCh38
rs747364547 Chr.X: 109659003-109659004 on Build GRCh38
rs953387465 Chr.X: 109659377-109659377 on Build GRCh38
rs762308327 Chr.X: 109659326-109659326 on Build GRCh38
rs145872865 Chr.X: 109659357-109659357 on Build GRCh38
rs773383629 Chr.X: 109659280-109659280 on Build GRCh38
rs776688086 Chr.X: 109659066-109659066 on Build GRCh38
rs188972332 Chr.X: 109659239-109659239 on Build GRCh38
rs976714333 Chr.X: 109659309-109659309 on Build GRCh38
rs944988806 Chr.X: 109659281-109659281 on Build GRCh38
rs1044675047 Chr.X: 109659026-109659026 on Build GRCh38
rs892888627 Chr.X: 109659371-109659371 on Build GRCh38
rs779450925 Chr.X: 109659345-109659348 on Build GRCh38
rs959612617 Chr.X: 109659087-109659087 on Build GRCh38
rs904908471 Chr.X: 109659044-109659044 on Build GRCh38
rs963417207 Chr.X: 109659302-109659304 on Build GRCh38
rs1027683631 Chr.X: 109659152-109659152 on Build GRCh38
rs372304436 Chr.X: 109659373-109659373 on Build GRCh38
rs752680113 Chr.X: 109659322-109659322 on Build GRCh38
rs375278019 Chr.X: 109659112-109659112 on Build GRCh38
rs750845761 Chr.X: 109659394-109659394 on Build GRCh38
rs765152419 Chr.X: 109659311-109659311 on Build GRCh38
rs768994066 Chr.X: 109659035-109659035 on Build GRCh38
rs763686403 Chr.X: 109659338-109659338 on Build GRCh38
rs780359191 Chr.X: 109659421-109659421 on Build GRCh38
rs983583224 Chr.X: 109659214-109659214 on Build GRCh38
rs1004018522 Chr.X: 109659181-109659181 on Build GRCh38

View Full Product Details