Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 119454184-119454709 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC25A43
Entrez Gene ID
203427
Gene Name
solute carrier family 25 member 43
Gene Aliases
-
UniGene
Hs.496658
Gene Chromosome Location
Chr.X: 119399295-119454478 on Build GRCh38
Transcripts
NM_145305.2

SNP Detail

SNP ID Chromosome Location
rs918556039 Chr.X: 119454653-119454653 on Build GRCh38
rs1048518020 Chr.X: 119454270-119454270 on Build GRCh38
rs750220294 Chr.X: 119454415-119454418 on Build GRCh38
rs769092481 Chr.X: 119454402-119454402 on Build GRCh38
rs757120184 Chr.X: 119454400-119454400 on Build GRCh38
rs980703902 Chr.X: 119454609-119454609 on Build GRCh38
rs753359771 Chr.X: 119454379-119454379 on Build GRCh38
rs917064670 Chr.X: 119454279-119454279 on Build GRCh38
rs1023843858 Chr.X: 119454463-119454463 on Build GRCh38
rs867898631 Chr.X: 119454416-119454416 on Build GRCh38
rs41300942 Chr.X: 119454301-119454301 on Build GRCh38
rs756544470 Chr.X: 119454254-119454254 on Build GRCh38
rs748302142 Chr.X: 119454639-119454639 on Build GRCh38
rs999418651 Chr.X: 119454340-119454340 on Build GRCh38
rs11558701 Chr.X: 119454277-119454277 on Build GRCh38
rs935721653 Chr.X: 119454610-119454610 on Build GRCh38
rs912990816 Chr.X: 119454652-119454653 on Build GRCh38
rs868223949 Chr.X: 119454418-119454418 on Build GRCh38
rs972993384 Chr.X: 119454647-119454647 on Build GRCh38
rs1007171380 Chr.X: 119454305-119454305 on Build GRCh38
rs781350138 Chr.X: 119454523-119454523 on Build GRCh38
rs10907045 Chr.X: 119454408-119454408 on Build GRCh38
rs567108533 Chr.X: 119454428-119454428 on Build GRCh38
rs928882836 Chr.X: 119454613-119454613 on Build GRCh38
rs780623804 Chr.X: 119454258-119454258 on Build GRCh38
rs960255401 Chr.X: 119454642-119454642 on Build GRCh38
rs760355234 Chr.X: 119454284-119454284 on Build GRCh38
rs969860642 Chr.X: 119454304-119454304 on Build GRCh38
rs747178251 Chr.X: 119454451-119454451 on Build GRCh38
rs755263845 Chr.X: 119454472-119454473 on Build GRCh38
rs901110063 Chr.X: 119454336-119454336 on Build GRCh38
rs970241842 Chr.X: 119454524-119454524 on Build GRCh38
rs779929448 Chr.X: 119454448-119454448 on Build GRCh38
rs1040955452 Chr.X: 119454307-119454307 on Build GRCh38

View Full Product Details