Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 142202225-142202753 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAGEC2
Entrez Gene ID
51438
Gene Name
MAGE family member C2
Gene Aliases
CT10, HCA587, MAGEE1
UniGene
Hs.123536
Gene Chromosome Location
Chr.X: 142202342-142205290 on Build GRCh38
Transcripts
NM_016249.3

SNP Detail

SNP ID Chromosome Location
rs915750764 Chr.X: 142202413-142202413 on Build GRCh38
rs889211745 Chr.X: 142202688-142202688 on Build GRCh38
rs1004119934 Chr.X: 142202622-142202622 on Build GRCh38
rs148758205 Chr.X: 142202597-142202597 on Build GRCh38
rs762934960 Chr.X: 142202690-142202690 on Build GRCh38
rs916872254 Chr.X: 142202467-142202467 on Build GRCh38
rs966986968 Chr.X: 142202304-142202304 on Build GRCh38
rs760342104 Chr.X: 142202517-142202517 on Build GRCh38
rs914173229 Chr.X: 142202296-142202296 on Build GRCh38
rs759186743 Chr.X: 142202378-142202378 on Build GRCh38
rs991178820 Chr.X: 142202414-142202414 on Build GRCh38
rs188818047 Chr.X: 142202689-142202689 on Build GRCh38
rs996937518 Chr.X: 142202719-142202719 on Build GRCh38
rs989740221 Chr.X: 142202257-142202257 on Build GRCh38
rs951204268 Chr.X: 142202285-142202285 on Build GRCh38
rs777370461 Chr.X: 142202644-142202644 on Build GRCh38
rs958154164 Chr.X: 142202256-142202256 on Build GRCh38
rs753432473 Chr.X: 142202673-142202673 on Build GRCh38
rs1028515883 Chr.X: 142202249-142202249 on Build GRCh38
rs901148925 Chr.X: 142202674-142202674 on Build GRCh38
rs542956529 Chr.X: 142202516-142202517 on Build GRCh38
rs1041455562 Chr.X: 142202653-142202653 on Build GRCh38
rs375508379 Chr.X: 142202531-142202531 on Build GRCh38
rs977652762 Chr.X: 142202527-142202527 on Build GRCh38
rs769579340 Chr.X: 142202582-142202582 on Build GRCh38
rs928309155 Chr.X: 142202335-142202335 on Build GRCh38
rs1055429935 Chr.X: 142202388-142202389 on Build GRCh38
rs766972187 Chr.X: 142202325-142202325 on Build GRCh38
rs943417407 Chr.X: 142202697-142202697 on Build GRCh38
rs938449582 Chr.X: 142202382-142202382 on Build GRCh38
rs527670234 Chr.X: 142202686-142202686 on Build GRCh38
rs142430022 Chr.X: 142202640-142202640 on Build GRCh38
rs931180070 Chr.X: 142202415-142202415 on Build GRCh38
rs374515121 Chr.X: 142202531-142202532 on Build GRCh38
rs767885246 Chr.X: 142202544-142202544 on Build GRCh38
rs950299546 Chr.X: 142202482-142202482 on Build GRCh38
rs1016681102 Chr.X: 142202342-142202342 on Build GRCh38
rs752009219 Chr.X: 142202443-142202443 on Build GRCh38
rs983869430 Chr.X: 142202327-142202327 on Build GRCh38
rs958660816 Chr.X: 142202371-142202371 on Build GRCh38
rs759972981 Chr.X: 142202533-142202533 on Build GRCh38
rs1048219262 Chr.X: 142202506-142202506 on Build GRCh38

View Full Product Details