Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 198639123-198639663 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTPRC
Entrez Gene ID
5788
Gene Name
protein tyrosine phosphatase, receptor type C
Gene Aliases
B220, CD45, CD45R, GP180, L-CA, LCA, LY5, T200
UniGene
Hs.654514
Gene Chromosome Location
Chr.1: 198638968-198757476 on Build GRCh38
Transcripts
NM_001267798.1 NM_002838.4 NM_080921.3 NR_052021.1 XM_006711472.3 XM_006711473.3 XM_006711474.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5691087 c.26C>T p.A9V Chr.1: 198639300-198639300 on Build GRCh38
26016 c.67G>A p.G23R Chr.1: 198639341-198639341 on Build GRCh38
1160886 c.40_5718>T p.L15fs*26 Chr.1: 198639314-198639331 on Build GRCh38
3400174 c.55G>T p.V19L Chr.1: 198639329-198639329 on Build GRCh38
6490014 c.49A>T p.T17S Chr.1: 198639323-198639323 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779729917 Chr.1: 198639354-198639354 on Build GRCh38
rs1017588531 Chr.1: 198639181-198639181 on Build GRCh38
rs779020551 Chr.1: 198639232-198639232 on Build GRCh38
rs1039306236 Chr.1: 198639557-198639557 on Build GRCh38
rs752452436 Chr.1: 198639452-198639452 on Build GRCh38
rs528738710 Chr.1: 198639399-198639399 on Build GRCh38
rs769521927 Chr.1: 198639260-198639260 on Build GRCh38
rs567062751 Chr.1: 198639544-198639544 on Build GRCh38
rs368929179 Chr.1: 198639275-198639275 on Build GRCh38
rs541883459 Chr.1: 198639231-198639231 on Build GRCh38
rs900469946 Chr.1: 198639576-198639576 on Build GRCh38
rs781080051 Chr.1: 198639320-198639320 on Build GRCh38
rs768227005 Chr.1: 198639273-198639273 on Build GRCh38
rs748901652 Chr.1: 198639360-198639360 on Build GRCh38
rs751543114 Chr.1: 198639580-198639580 on Build GRCh38
rs746884648 Chr.1: 198639239-198639239 on Build GRCh38
rs184778190 Chr.1: 198639527-198639527 on Build GRCh38
rs910014416 Chr.1: 198639604-198639604 on Build GRCh38
rs773900940 Chr.1: 198639279-198639279 on Build GRCh38
rs778513212 Chr.1: 198639368-198639368 on Build GRCh38
rs766720230 Chr.1: 198639292-198639292 on Build GRCh38
rs756237254 Chr.1: 198639229-198639229 on Build GRCh38
rs758522951 Chr.1: 198639317-198639317 on Build GRCh38
rs745653911 Chr.1: 198639253-198639253 on Build GRCh38
rs550322178 Chr.1: 198639506-198639506 on Build GRCh38
rs998303873 Chr.1: 198639566-198639566 on Build GRCh38
rs930116604 Chr.1: 198639156-198639156 on Build GRCh38
rs61757803 Chr.1: 198639296-198639296 on Build GRCh38
rs771747130 Chr.1: 198639372-198639372 on Build GRCh38
rs747635998 Chr.1: 198639371-198639371 on Build GRCh38
rs910138701 Chr.1: 198639200-198639200 on Build GRCh38
rs199852332 Chr.1: 198639325-198639325 on Build GRCh38
rs143814848 Chr.1: 198639245-198639245 on Build GRCh38
rs755927316 Chr.1: 198639350-198639350 on Build GRCh38
rs374062143 Chr.1: 198639543-198639543 on Build GRCh38
rs564768902 Chr.1: 198639352-198639352 on Build GRCh38
rs926264983 Chr.1: 198639384-198639385 on Build GRCh38
rs141414674 Chr.1: 198639307-198639307 on Build GRCh38
rs547201711 Chr.1: 198639456-198639456 on Build GRCh38
rs892324446 Chr.1: 198639463-198639463 on Build GRCh38
rs1056616032 Chr.1: 198639461-198639461 on Build GRCh38
rs759684142 Chr.1: 198639299-198639299 on Build GRCh38
rs772724488 Chr.1: 198639276-198639276 on Build GRCh38
rs1008890871 Chr.1: 198639174-198639174 on Build GRCh38
rs773764906 Chr.1: 198639375-198639375 on Build GRCh38
rs753531027 Chr.1: 198639261-198639261 on Build GRCh38
rs376285687 Chr.1: 198639265-198639265 on Build GRCh38
rs771268163 Chr.1: 198639389-198639389 on Build GRCh38
rs752797028 Chr.1: 198639312-198639312 on Build GRCh38
rs912060915 Chr.1: 198639548-198639549 on Build GRCh38
rs768452006 Chr.1: 198639367-198639367 on Build GRCh38
rs773998713 Chr.1: 198639274-198639274 on Build GRCh38
rs563770736 Chr.1: 198639465-198639465 on Build GRCh38
rs944917319 Chr.1: 198639549-198639549 on Build GRCh38
rs951467427 Chr.1: 198639601-198639601 on Build GRCh38
rs780916589 Chr.1: 198639246-198639246 on Build GRCh38
rs574858681 Chr.1: 198639209-198639209 on Build GRCh38
rs765018292 Chr.1: 198639432-198639432 on Build GRCh38
rs568795394 Chr.1: 198639521-198639521 on Build GRCh38
rs557675850 Chr.1: 198639574-198639574 on Build GRCh38
rs759182888 Chr.1: 198639271-198639271 on Build GRCh38
rs761141412 Chr.1: 198639383-198639383 on Build GRCh38
rs983373472 Chr.1: 198639602-198639602 on Build GRCh38
rs761069010 Chr.1: 198639284-198639284 on Build GRCh38
rs1007446801 Chr.1: 198639534-198639534 on Build GRCh38
rs568091555 Chr.1: 198639458-198639458 on Build GRCh38
rs983083787 Chr.1: 198639195-198639195 on Build GRCh38

View Full Product Details