Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 200174213-200174715 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NR5A2
Entrez Gene ID
2494
Gene Name
nuclear receptor subfamily 5 group A member 2
Gene Aliases
B1F, B1F2, CPF, FTF, FTZ-F1, FTZ-F1beta, LRH-1, LRH1, hB1F-2
UniGene
Hs.33446
Gene Chromosome Location
Chr.1: 200027602-200177424 on Build GRCh38
Transcripts
NM_001276464.1 NM_003822.4 NM_205860.2 XM_005245062.3 XM_011509380.1 XM_011509381.2 XM_011509382.1 XM_011509384.2 XM_017000904.1

SNP Detail

SNP ID Chromosome Location
rs562141771 Chr.1: 200174606-200174606 on Build GRCh38
rs749349138 Chr.1: 200174589-200174589 on Build GRCh38
rs757657182 Chr.1: 200174262-200174262 on Build GRCh38
rs946893494 Chr.1: 200174363-200174363 on Build GRCh38
rs536706732 Chr.1: 200174626-200174626 on Build GRCh38
rs576777421 Chr.1: 200174313-200174313 on Build GRCh38
rs953092035 Chr.1: 200174335-200174335 on Build GRCh38
rs531645020 Chr.1: 200174532-200174532 on Build GRCh38
rs866343588 Chr.1: 200174267-200174267 on Build GRCh38
rs187662223 Chr.1: 200174282-200174282 on Build GRCh38
rs1060062 Chr.1: 200174587-200174587 on Build GRCh38
rs986314754 Chr.1: 200174352-200174352 on Build GRCh38
rs554148554 Chr.1: 200174395-200174395 on Build GRCh38
rs866274476 Chr.1: 200174333-200174333 on Build GRCh38
rs1043945798 Chr.1: 200174412-200174412 on Build GRCh38
rs1000118987 Chr.1: 200174537-200174537 on Build GRCh38
rs377747524 Chr.1: 200174607-200174607 on Build GRCh38
rs1023453606 Chr.1: 200174271-200174273 on Build GRCh38
rs868753440 Chr.1: 200174318-200174318 on Build GRCh38
rs1056371499 Chr.1: 200174469-200174469 on Build GRCh38
rs992239683 Chr.1: 200174256-200174256 on Build GRCh38
rs779843510 Chr.1: 200174500-200174500 on Build GRCh38
rs1060061 Chr.1: 200174314-200174314 on Build GRCh38
rs780120680 Chr.1: 200174539-200174539 on Build GRCh38
rs182552712 Chr.1: 200174438-200174438 on Build GRCh38
rs543874380 Chr.1: 200174317-200174317 on Build GRCh38
rs924203896 Chr.1: 200174432-200174432 on Build GRCh38
rs966875200 Chr.1: 200174278-200174278 on Build GRCh38
rs763328155 Chr.1: 200174262-200174262 on Build GRCh38
rs764395661 Chr.1: 200174428-200174428 on Build GRCh38
rs1029991054 Chr.1: 200174595-200174595 on Build GRCh38
rs537191680 Chr.1: 200174285-200174285 on Build GRCh38
rs895192152 Chr.1: 200174522-200174522 on Build GRCh38
rs911365750 Chr.1: 200174360-200174361 on Build GRCh38
rs953715717 Chr.1: 200174676-200174676 on Build GRCh38
rs377340850 Chr.1: 200174247-200174247 on Build GRCh38
rs995402165 Chr.1: 200174490-200174490 on Build GRCh38
rs1055446685 Chr.1: 200174508-200174508 on Build GRCh38
rs374542267 Chr.1: 200174574-200174574 on Build GRCh38
rs1032470876 Chr.1: 200174538-200174538 on Build GRCh38
rs1021797009 Chr.1: 200174630-200174630 on Build GRCh38
rs1025013802 Chr.1: 200174271-200174271 on Build GRCh38
rs922173992 Chr.1: 200174605-200174605 on Build GRCh38
rs986437250 Chr.1: 200174688-200174688 on Build GRCh38
rs760079892 Chr.1: 200174238-200174238 on Build GRCh38
rs370677122 Chr.1: 200174252-200174252 on Build GRCh38
rs1045572970 Chr.1: 200174504-200174504 on Build GRCh38
rs192579356 Chr.1: 200174296-200174296 on Build GRCh38
rs932296905 Chr.1: 200174368-200174368 on Build GRCh38
rs116603498 Chr.1: 200174479-200174479 on Build GRCh38
rs746081498 Chr.1: 200174400-200174400 on Build GRCh38
rs561192271 Chr.1: 200174531-200174531 on Build GRCh38
rs1013523758 Chr.1: 200174523-200174523 on Build GRCh38
rs1050869159 Chr.1: 200174383-200174383 on Build GRCh38
rs769182489 Chr.1: 200174507-200174507 on Build GRCh38

View Full Product Details