Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 201405125-201405633 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TNNI1
Entrez Gene ID
7135
Gene Name
troponin I1, slow skeletal type
Gene Aliases
SSTNI, TNN1
UniGene
Hs.320890
Gene Chromosome Location
Chr.1: 201403767-201421746 on Build GRCh38
Transcripts
NM_003281.3

SNP Detail

SNP ID Chromosome Location
rs989695395 Chr.1: 201405532-201405532 on Build GRCh38
rs193186676 Chr.1: 201405245-201405245 on Build GRCh38
rs1005993668 Chr.1: 201405598-201405598 on Build GRCh38
rs769247005 Chr.1: 201405281-201405281 on Build GRCh38
rs370521094 Chr.1: 201405300-201405300 on Build GRCh38
rs1053197293 Chr.1: 201405187-201405187 on Build GRCh38
rs775090507 Chr.1: 201405299-201405299 on Build GRCh38
rs2799680 Chr.1: 201405549-201405549 on Build GRCh38
rs866967075 Chr.1: 201405420-201405420 on Build GRCh38
rs548563631 Chr.1: 201405293-201405293 on Build GRCh38
rs1256944 Chr.1: 201405525-201405525 on Build GRCh38
rs923212371 Chr.1: 201405487-201405487 on Build GRCh38
rs554560458 Chr.1: 201405573-201405573 on Build GRCh38
rs537556108 Chr.1: 201405310-201405310 on Build GRCh38
rs779554102 Chr.1: 201405544-201405544 on Build GRCh38
rs187074962 Chr.1: 201405244-201405244 on Build GRCh38
rs1050543864 Chr.1: 201405262-201405262 on Build GRCh38
rs943244374 Chr.1: 201405375-201405375 on Build GRCh38
rs994340668 Chr.1: 201405561-201405561 on Build GRCh38
rs891845697 Chr.1: 201405520-201405520 on Build GRCh38
rs1027759669 Chr.1: 201405331-201405331 on Build GRCh38
rs995376756 Chr.1: 201405312-201405312 on Build GRCh38
rs141957386 Chr.1: 201405531-201405531 on Build GRCh38
rs931685119 Chr.1: 201405246-201405246 on Build GRCh38
rs891906386 Chr.1: 201405189-201405189 on Build GRCh38
rs185124375 Chr.1: 201405368-201405368 on Build GRCh38
rs866333631 Chr.1: 201405319-201405319 on Build GRCh38
rs950482796 Chr.1: 201405344-201405344 on Build GRCh38
rs762566542 Chr.1: 201405382-201405382 on Build GRCh38
rs974702413 Chr.1: 201405421-201405421 on Build GRCh38
rs896094851 Chr.1: 201405555-201405555 on Build GRCh38
rs944322095 Chr.1: 201405576-201405576 on Build GRCh38
rs868476998 Chr.1: 201405476-201405476 on Build GRCh38
rs751109318 Chr.1: 201405279-201405279 on Build GRCh38
rs865859781 Chr.1: 201405537-201405537 on Build GRCh38
rs138836335 Chr.1: 201405199-201405199 on Build GRCh38
rs527808002 Chr.1: 201405502-201405502 on Build GRCh38
rs1053403236 Chr.1: 201405519-201405519 on Build GRCh38
rs1001757375 Chr.1: 201405248-201405248 on Build GRCh38
rs766044523 Chr.1: 201405233-201405233 on Build GRCh38
rs540075931 Chr.1: 201405388-201405388 on Build GRCh38
rs551068600 Chr.1: 201405363-201405363 on Build GRCh38
rs923825028 Chr.1: 201405526-201405526 on Build GRCh38
rs999829416 Chr.1: 201405284-201405284 on Build GRCh38
rs971367228 Chr.1: 201405469-201405469 on Build GRCh38
rs911595361 Chr.1: 201405574-201405574 on Build GRCh38
rs950408189 Chr.1: 201405575-201405575 on Build GRCh38

View Full Product Details