Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 203489159-203489656 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRELP
Entrez Gene ID
5549
Gene Name
proline and arginine rich end leucine rich repeat protein
Gene Aliases
MST161, MSTP161, SLRR2A
UniGene
Hs.632481
Gene Chromosome Location
Chr.1: 203475755-203491352 on Build GRCh38
Transcripts
NM_002725.3 NM_201348.1

SNP Detail

SNP ID Chromosome Location
rs907739414 Chr.1: 203489454-203489454 on Build GRCh38
rs990830388 Chr.1: 203489616-203489616 on Build GRCh38
rs533487024 Chr.1: 203489622-203489622 on Build GRCh38
rs1021119810 Chr.1: 203489519-203489519 on Build GRCh38
rs531481195 Chr.1: 203489633-203489633 on Build GRCh38
rs562397542 Chr.1: 203489197-203489197 on Build GRCh38
rs1032286269 Chr.1: 203489388-203489388 on Build GRCh38
rs899712029 Chr.1: 203489344-203489344 on Build GRCh38
rs529032369 Chr.1: 203489516-203489516 on Build GRCh38
rs763981930 Chr.1: 203489591-203489591 on Build GRCh38
rs765124007 Chr.1: 203489193-203489193 on Build GRCh38
rs75953692 Chr.1: 203489187-203489187 on Build GRCh38
rs577475874 Chr.1: 203489398-203489398 on Build GRCh38
rs140810007 Chr.1: 203489526-203489526 on Build GRCh38
rs895488507 Chr.1: 203489630-203489638 on Build GRCh38
rs973632361 Chr.1: 203489473-203489473 on Build GRCh38
rs189534375 Chr.1: 203489419-203489419 on Build GRCh38
rs931627948 Chr.1: 203489492-203489492 on Build GRCh38
rs531765170 Chr.1: 203489440-203489440 on Build GRCh38
rs770618943 Chr.1: 203489256-203489256 on Build GRCh38
rs551175208 Chr.1: 203489332-203489332 on Build GRCh38
rs917977169 Chr.1: 203489303-203489303 on Build GRCh38
rs780837014 Chr.1: 203489347-203489347 on Build GRCh38
rs9475 Chr.1: 203489279-203489279 on Build GRCh38
rs959896074 Chr.1: 203489609-203489611 on Build GRCh38
rs762604414 Chr.1: 203489224-203489224 on Build GRCh38
rs1047183189 Chr.1: 203489246-203489246 on Build GRCh38
rs919488230 Chr.1: 203489498-203489498 on Build GRCh38
rs941940836 Chr.1: 203489460-203489460 on Build GRCh38
rs1010424261 Chr.1: 203489513-203489513 on Build GRCh38
rs1057078040 Chr.1: 203489497-203489511 on Build GRCh38
rs559888522 Chr.1: 203489466-203489466 on Build GRCh38
rs929635631 Chr.1: 203489242-203489242 on Build GRCh38
rs113642415 Chr.1: 203489521-203489521 on Build GRCh38
rs866299371 Chr.1: 203489325-203489325 on Build GRCh38
rs1025437793 Chr.1: 203489621-203489621 on Build GRCh38
rs1039458534 Chr.1: 203489580-203489580 on Build GRCh38
rs28655574 Chr.1: 203489468-203489468 on Build GRCh38
rs970760671 Chr.1: 203489422-203489422 on Build GRCh38
rs979707762 Chr.1: 203489194-203489194 on Build GRCh38
rs762128320 Chr.1: 203489386-203489387 on Build GRCh38
rs1009539885 Chr.1: 203489415-203489415 on Build GRCh38
rs569408886 Chr.1: 203489472-203489472 on Build GRCh38
rs763980022 Chr.1: 203489267-203489267 on Build GRCh38
rs1002327409 Chr.1: 203489435-203489435 on Build GRCh38
rs144758400 Chr.1: 203489632-203489632 on Build GRCh38
rs892394202 Chr.1: 203489390-203489390 on Build GRCh38
rs959335143 Chr.1: 203489220-203489220 on Build GRCh38
rs943783257 Chr.1: 203489550-203489550 on Build GRCh38
rs1033838774 Chr.1: 203489443-203489443 on Build GRCh38
rs551851492 Chr.1: 203489631-203489631 on Build GRCh38
rs949536413 Chr.1: 203489401-203489401 on Build GRCh38
rs35582620 Chr.1: 203489418-203489419 on Build GRCh38
rs1037554735 Chr.1: 203489467-203489467 on Build GRCh38

View Full Product Details