Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 206110034-206109538 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AVPR1B
Entrez Gene ID
553
Gene Name
arginine vasopressin receptor 1B
Gene Aliases
AVPR3
UniGene
Hs.1372
Gene Chromosome Location
Chr.1: 206109849-206117048 on Build GRCh38
Transcripts
NM_000707.3

View Full Product Details