Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 208022111-208022611 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLXNA2
Entrez Gene ID
5362
Gene Name
plexin A2
Gene Aliases
OCT, PLXN2
UniGene
Hs.497626
Gene Chromosome Location
Chr.1: 208022242-208244384 on Build GRCh38
Transcripts
NM_025179.3 XM_005273164.2

SNP Detail

SNP ID Chromosome Location
rs989437816 Chr.1: 208022334-208022334 on Build GRCh38
rs931394117 Chr.1: 208022522-208022522 on Build GRCh38
rs751948539 Chr.1: 208022433-208022433 on Build GRCh38
rs937611521 Chr.1: 208022478-208022478 on Build GRCh38
rs879167177 Chr.1: 208022445-208022445 on Build GRCh38
rs764390420 Chr.1: 208022408-208022408 on Build GRCh38
rs757692316 Chr.1: 208022502-208022502 on Build GRCh38
rs1047632529 Chr.1: 208022567-208022567 on Build GRCh38
rs1050365867 Chr.1: 208022187-208022187 on Build GRCh38
rs28626591 Chr.1: 208022152-208022152 on Build GRCh38
rs192223321 Chr.1: 208022289-208022289 on Build GRCh38
rs941528260 Chr.1: 208022560-208022560 on Build GRCh38
rs919948107 Chr.1: 208022521-208022521 on Build GRCh38
rs1015789460 Chr.1: 208022238-208022238 on Build GRCh38
rs957116247 Chr.1: 208022366-208022366 on Build GRCh38
rs981698261 Chr.1: 208022157-208022157 on Build GRCh38
rs1024607253 Chr.1: 208022271-208022271 on Build GRCh38
rs928851574 Chr.1: 208022477-208022477 on Build GRCh38
rs555068498 Chr.1: 208022383-208022383 on Build GRCh38
rs1029094830 Chr.1: 208022313-208022313 on Build GRCh38
rs867447417 Chr.1: 208022576-208022576 on Build GRCh38
rs753529089 Chr.1: 208022239-208022239 on Build GRCh38
rs928631989 Chr.1: 208022163-208022163 on Build GRCh38
rs931401020 Chr.1: 208022203-208022203 on Build GRCh38
rs886267947 Chr.1: 208022570-208022570 on Build GRCh38
rs932751295 Chr.1: 208022466-208022466 on Build GRCh38
rs867253697 Chr.1: 208022575-208022575 on Build GRCh38
rs1005785623 Chr.1: 208022220-208022220 on Build GRCh38
rs995819366 Chr.1: 208022303-208022303 on Build GRCh38
rs557122772 Chr.1: 208022209-208022209 on Build GRCh38
rs116467274 Chr.1: 208022571-208022571 on Build GRCh38
rs1014487724 Chr.1: 208022586-208022586 on Build GRCh38
rs977171358 Chr.1: 208022386-208022386 on Build GRCh38
rs1004802941 Chr.1: 208022240-208022240 on Build GRCh38
rs2767568 Chr.1: 208022561-208022561 on Build GRCh38
rs1033544071 Chr.1: 208022296-208022296 on Build GRCh38
rs898554666 Chr.1: 208022189-208022189 on Build GRCh38
rs990335529 Chr.1: 208022379-208022379 on Build GRCh38
rs1193096 Chr.1: 208022171-208022171 on Build GRCh38
rs530337867 Chr.1: 208022260-208022261 on Build GRCh38
rs937307467 Chr.1: 208022168-208022168 on Build GRCh38
rs901770282 Chr.1: 208022293-208022293 on Build GRCh38
rs536375661 Chr.1: 208022330-208022330 on Build GRCh38
rs573442054 Chr.1: 208022532-208022532 on Build GRCh38
rs981581836 Chr.1: 208022422-208022422 on Build GRCh38
rs888647214 Chr.1: 208022539-208022539 on Build GRCh38
rs1020448249 Chr.1: 208022389-208022389 on Build GRCh38
rs991475064 Chr.1: 208022518-208022518 on Build GRCh38
rs397732495 Chr.1: 208022267-208022268 on Build GRCh38
rs913824196 Chr.1: 208022339-208022339 on Build GRCh38
rs1040236630 Chr.1: 208022588-208022588 on Build GRCh38

View Full Product Details