Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 212729423-212729949 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NSL1
Entrez Gene ID
25936
Gene Name
NSL1, MIS12 kinetochore complex component
Gene Aliases
C1orf48, DC8, MIS14
UniGene
Hs.497692
Gene Chromosome Location
Chr.1: 212726153-212798900 on Build GRCh38
Transcripts
NM_001042549.1 NM_001297736.1 NM_001297737.1 NM_001297739.1 NM_015471.3

SNP Detail

SNP ID Chromosome Location
rs895744286 Chr.1: 212729618-212729618 on Build GRCh38
rs775463168 Chr.1: 212729496-212729496 on Build GRCh38
rs528238035 Chr.1: 212729729-212729730 on Build GRCh38
rs149266235 Chr.1: 212729745-212729745 on Build GRCh38
rs867118963 Chr.1: 212729622-212729622 on Build GRCh38
rs1051054368 Chr.1: 212729688-212729688 on Build GRCh38
rs112919348 Chr.1: 212729871-212729871 on Build GRCh38
rs1039884486 Chr.1: 212729525-212729525 on Build GRCh38
rs535899202 Chr.1: 212729614-212729614 on Build GRCh38
rs898658380 Chr.1: 212729537-212729537 on Build GRCh38
rs965273697 Chr.1: 212729498-212729498 on Build GRCh38
rs142748361 Chr.1: 212729615-212729615 on Build GRCh38
rs1039744263 Chr.1: 212729720-212729720 on Build GRCh38
rs887358161 Chr.1: 212729775-212729775 on Build GRCh38
rs913221627 Chr.1: 212729854-212729854 on Build GRCh38
rs984315131 Chr.1: 212729476-212729476 on Build GRCh38
rs12747251 Chr.1: 212729747-212729747 on Build GRCh38
rs538834122 Chr.1: 212729558-212729558 on Build GRCh38
rs1047203748 Chr.1: 212729756-212729756 on Build GRCh38
rs775638623 Chr.1: 212729836-212729836 on Build GRCh38
rs778722625 Chr.1: 212729453-212729453 on Build GRCh38
rs747211033 Chr.1: 212729521-212729521 on Build GRCh38
rs12128903 Chr.1: 212729709-212729709 on Build GRCh38
rs947477844 Chr.1: 212729583-212729583 on Build GRCh38
rs927373512 Chr.1: 212729890-212729890 on Build GRCh38
rs555144427 Chr.1: 212729853-212729853 on Build GRCh38
rs11120006 Chr.1: 212729469-212729469 on Build GRCh38
rs188006388 Chr.1: 212729514-212729514 on Build GRCh38
rs565017319 Chr.1: 212729656-212729656 on Build GRCh38
rs537136804 Chr.1: 212729576-212729576 on Build GRCh38
rs146068744 Chr.1: 212729616-212729616 on Build GRCh38
rs553018981 Chr.1: 212729458-212729458 on Build GRCh38
rs964882000 Chr.1: 212729711-212729711 on Build GRCh38
rs11120007 Chr.1: 212729559-212729559 on Build GRCh38
rs1035387787 Chr.1: 212729814-212729814 on Build GRCh38
rs933213920 Chr.1: 212729665-212729665 on Build GRCh38
rs1043565477 Chr.1: 212729609-212729609 on Build GRCh38
rs989047576 Chr.1: 212729833-212729833 on Build GRCh38
rs765715599 Chr.1: 212729671-212729671 on Build GRCh38
rs974935498 Chr.1: 212729502-212729502 on Build GRCh38
rs933281671 Chr.1: 212729550-212729550 on Build GRCh38
rs1016414377 Chr.1: 212729484-212729484 on Build GRCh38
rs1046762034 Chr.1: 212729637-212729637 on Build GRCh38
rs954661592 Chr.1: 212729917-212729917 on Build GRCh38
rs1002557911 Chr.1: 212729567-212729567 on Build GRCh38
rs139958351 Chr.1: 212729649-212729649 on Build GRCh38
rs543308742 Chr.1: 212729750-212729750 on Build GRCh38
rs386639134 Chr.1: 212729615-212729616 on Build GRCh38
rs894142998 Chr.1: 212729819-212729819 on Build GRCh38
rs912556351 Chr.1: 212729697-212729697 on Build GRCh38
rs954958927 Chr.1: 212729778-212729778 on Build GRCh38
rs766701382 Chr.1: 212729861-212729861 on Build GRCh38
rs769056644 Chr.1: 212729555-212729555 on Build GRCh38

View Full Product Details