Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 220153558-220154106 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAB3GAP2
Entrez Gene ID
25782
Gene Name
RAB3 GTPase activating non-catalytic protein subunit 2
Gene Aliases
RAB3-GAP150, RAB3GAP150, SPG69, WARBM2, p150
UniGene
Hs.654849
Gene Chromosome Location
Chr.1: 220148268-220272501 on Build GRCh38
Transcripts
NM_012414.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1601663 c.3604A>T p.S1202C Chr.1: 220154009-220154009 on Build GRCh38
6400016 c.3565G>T p.A1189S Chr.1: 220154048-220154048 on Build GRCh38
1959163 c.3563delA p.N1188fs*7 Chr.1: 220154050-220154050 on Build GRCh38
5352645 c.3632C>A p.S1211Y Chr.1: 220153981-220153981 on Build GRCh38
6124187 c.3608G>A p.G1203E Chr.1: 220154005-220154005 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs538580550 Chr.1: 220153894-220153894 on Build GRCh38
rs891492677 Chr.1: 220153795-220153795 on Build GRCh38
rs537731357 Chr.1: 220153856-220153856 on Build GRCh38
rs887988484 Chr.1: 220153763-220153763 on Build GRCh38
rs997371913 Chr.1: 220153929-220153930 on Build GRCh38
rs186369879 Chr.1: 220153591-220153591 on Build GRCh38
rs139407102 Chr.1: 220153997-220153997 on Build GRCh38
rs557396940 Chr.1: 220153920-220153921 on Build GRCh38
rs747881174 Chr.1: 220154075-220154075 on Build GRCh38
rs780158398 Chr.1: 220154048-220154048 on Build GRCh38
rs755893160 Chr.1: 220154046-220154046 on Build GRCh38
rs1009854592 Chr.1: 220153823-220153823 on Build GRCh38
rs568734157 Chr.1: 220153701-220153701 on Build GRCh38
rs1048266620 Chr.1: 220153723-220153723 on Build GRCh38
rs766889252 Chr.1: 220153935-220153938 on Build GRCh38
rs551129561 Chr.1: 220153756-220153756 on Build GRCh38
rs765602795 Chr.1: 220153920-220153920 on Build GRCh38
rs901566929 Chr.1: 220153827-220153827 on Build GRCh38
rs781081339 Chr.1: 220154034-220154034 on Build GRCh38
rs907298690 Chr.1: 220153846-220153846 on Build GRCh38
rs918004721 Chr.1: 220153727-220153727 on Build GRCh38
rs992266587 Chr.1: 220153708-220153708 on Build GRCh38
rs950496458 Chr.1: 220153991-220153991 on Build GRCh38
rs536489899 Chr.1: 220153822-220153822 on Build GRCh38
rs751576247 Chr.1: 220154068-220154069 on Build GRCh38
rs962024187 Chr.1: 220153693-220153693 on Build GRCh38
rs1017455183 Chr.1: 220153928-220153928 on Build GRCh38
rs763596081 Chr.1: 220153791-220153791 on Build GRCh38
rs543785834 Chr.1: 220154055-220154055 on Build GRCh38
rs1017425396 Chr.1: 220153667-220153674 on Build GRCh38
rs577447105 Chr.1: 220153794-220153794 on Build GRCh38
rs587777169 Chr.1: 220153976-220153976 on Build GRCh38
rs984824792 Chr.1: 220153685-220153685 on Build GRCh38
rs757400581 Chr.1: 220154027-220154027 on Build GRCh38
rs749173523 Chr.1: 220154050-220154050 on Build GRCh38
rs1040132734 Chr.1: 220153826-220153826 on Build GRCh38
rs753152798 Chr.1: 220153922-220153922 on Build GRCh38
rs996402766 Chr.1: 220153608-220153608 on Build GRCh38
rs916111898 Chr.1: 220153776-220153776 on Build GRCh38
rs1053129858 Chr.1: 220153871-220153871 on Build GRCh38
rs191171363 Chr.1: 220153645-220153645 on Build GRCh38
rs1014830345 Chr.1: 220153996-220153996 on Build GRCh38
rs952007605 Chr.1: 220153870-220153870 on Build GRCh38
rs983144414 Chr.1: 220153741-220153741 on Build GRCh38
rs150546097 Chr.1: 220153847-220153847 on Build GRCh38
rs763433911 Chr.1: 220153921-220153921 on Build GRCh38
rs373538022 Chr.1: 220154045-220154045 on Build GRCh38
rs778549979 Chr.1: 220154069-220154069 on Build GRCh38
rs948238350 Chr.1: 220153739-220153739 on Build GRCh38
rs558398224 Chr.1: 220153919-220153919 on Build GRCh38
rs1038627544 Chr.1: 220153775-220153775 on Build GRCh38
rs751933206 Chr.1: 220153595-220153597 on Build GRCh38
rs571688600 Chr.1: 220153692-220153692 on Build GRCh38
rs941630222 Chr.1: 220153758-220153758 on Build GRCh38
rs952016895 Chr.1: 220153653-220153653 on Build GRCh38
rs555022251 Chr.1: 220153679-220153679 on Build GRCh38
rs765761497 Chr.1: 220153917-220153917 on Build GRCh38
rs925561073 Chr.1: 220153765-220153765 on Build GRCh38
rs1046297965 Chr.1: 220153824-220153824 on Build GRCh38
rs1030542656 Chr.1: 220153941-220153944 on Build GRCh38
rs537406824 Chr.1: 220153746-220153746 on Build GRCh38
rs574931984 Chr.1: 220153929-220153929 on Build GRCh38

View Full Product Details