Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 225402613-225403139 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LBR
Entrez Gene ID
3930
Gene Name
lamin B receptor
Gene Aliases
DHCR14B, LMN2R, PHA, TDRD18
UniGene
Hs.435166
Gene Chromosome Location
Chr.1: 225401502-225428855 on Build GRCh38
Transcripts
NM_002296.3 NM_194442.2 XM_005273125.2 XM_011544185.2

SNP Detail

SNP ID Chromosome Location
rs985568984 Chr.1: 225403094-225403094 on Build GRCh38
rs995607324 Chr.1: 225402784-225402784 on Build GRCh38
rs16844841 Chr.1: 225402973-225402973 on Build GRCh38
rs1033852204 Chr.1: 225403024-225403024 on Build GRCh38
rs80028106 Chr.1: 225402647-225402647 on Build GRCh38
rs565044223 Chr.1: 225402686-225402686 on Build GRCh38
rs572258776 Chr.1: 225402721-225402721 on Build GRCh38
rs922480181 Chr.1: 225402776-225402776 on Build GRCh38
rs1021432027 Chr.1: 225402983-225402983 on Build GRCh38
rs972407929 Chr.1: 225403096-225403096 on Build GRCh38
rs774945026 Chr.1: 225402964-225402964 on Build GRCh38
rs932603176 Chr.1: 225403061-225403061 on Build GRCh38
rs983829757 Chr.1: 225402644-225402644 on Build GRCh38
rs1009617892 Chr.1: 225402982-225402982 on Build GRCh38
rs11551874 Chr.1: 225403048-225403048 on Build GRCh38
rs761467375 Chr.1: 225403009-225403009 on Build GRCh38
rs562482402 Chr.1: 225402830-225402830 on Build GRCh38
rs933747456 Chr.1: 225403106-225403106 on Build GRCh38
rs922373633 Chr.1: 225403101-225403101 on Build GRCh38
rs1004156634 Chr.1: 225403012-225403012 on Build GRCh38
rs979056811 Chr.1: 225403051-225403051 on Build GRCh38
rs767353012 Chr.1: 225403034-225403034 on Build GRCh38
rs1035810101 Chr.1: 225402959-225402959 on Build GRCh38
rs1026639850 Chr.1: 225402807-225402807 on Build GRCh38
rs886046050 Chr.1: 225402679-225402679 on Build GRCh38
rs1053715754 Chr.1: 225402963-225402963 on Build GRCh38
rs762715244 Chr.1: 225402971-225402971 on Build GRCh38
rs925324699 Chr.1: 225403060-225403060 on Build GRCh38
rs113990759 Chr.1: 225403108-225403108 on Build GRCh38
rs771329334 Chr.1: 225402873-225402873 on Build GRCh38
rs944152738 Chr.1: 225402691-225402691 on Build GRCh38
rs755810784 Chr.1: 225403068-225403068 on Build GRCh38
rs544921637 Chr.1: 225402668-225402668 on Build GRCh38
rs939760688 Chr.1: 225402684-225402684 on Build GRCh38
rs1000066037 Chr.1: 225402902-225402902 on Build GRCh38
rs1037318610 Chr.1: 225402706-225402706 on Build GRCh38
rs751092768 Chr.1: 225402992-225402992 on Build GRCh38
rs1001311354 Chr.1: 225402908-225402908 on Build GRCh38

View Full Product Details