Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 244844384-244844899 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COX20
Entrez Gene ID
116228
Gene Name
COX20, cytochrome c oxidase assembly factor
Gene Aliases
FAM36A
UniGene
Hs.411490
Gene Chromosome Location
Chr.1: 244835306-244845063 on Build GRCh38
Transcripts
NM_001312871.1 NM_001312872.1 NM_001312873.1 NM_001312874.1 NM_198076.5 NR_132419.1 NR_132420.1 NR_132421.1 XM_017001115.1 XM_017001116.1 XM_017001117.1

Gene Symbol
HNRNPU
Entrez Gene ID
3192
Gene Name
heterogeneous nuclear ribonucleoprotein U
Gene Aliases
HNRNPU-AS1, HNRPU, SAF-A, SAFA, U21.1, hnRNP U
UniGene
Hs.106212
Gene Chromosome Location
Chr.1: 244842123-244864720 on Build GRCh38
Transcripts
NM_001312871.1 NM_001312872.1 NM_001312873.1 NM_001312874.1 NM_198076.5 NR_132419.1 NR_132420.1 NR_132421.1 XM_017001115.1 XM_017001116.1 XM_017001117.1

SNP Detail

SNP ID Chromosome Location
rs535412994 Chr.1: 244844641-244844641 on Build GRCh38
rs1001689661 Chr.1: 244844722-244844722 on Build GRCh38
rs1014299917 Chr.1: 244844714-244844714 on Build GRCh38
rs925421581 Chr.1: 244844592-244844592 on Build GRCh38
rs557498518 Chr.1: 244844431-244844431 on Build GRCh38
rs181870217 Chr.1: 244844531-244844531 on Build GRCh38
rs974936410 Chr.1: 244844746-244844747 on Build GRCh38
rs906133467 Chr.1: 244844720-244844720 on Build GRCh38
rs146150737 Chr.1: 244844860-244844860 on Build GRCh38
rs1002140393 Chr.1: 244844591-244844591 on Build GRCh38
rs149571617 Chr.1: 244844497-244844497 on Build GRCh38
rs895206309 Chr.1: 244844702-244844702 on Build GRCh38
rs758951475 Chr.1: 244844435-244844435 on Build GRCh38
rs970245085 Chr.1: 244844577-244844577 on Build GRCh38
rs920661084 Chr.1: 244844677-244844677 on Build GRCh38
rs978945460 Chr.1: 244844700-244844700 on Build GRCh38
rs370969811 Chr.1: 244844485-244844485 on Build GRCh38
rs940688645 Chr.1: 244844801-244844801 on Build GRCh38
rs922171597 Chr.1: 244844610-244844610 on Build GRCh38
rs576394539 Chr.1: 244844814-244844814 on Build GRCh38
rs541234844 Chr.1: 244844407-244844407 on Build GRCh38
rs907899718 Chr.1: 244844794-244844797 on Build GRCh38
rs374710215 Chr.1: 244844697-244844697 on Build GRCh38
rs11579106 Chr.1: 244844813-244844813 on Build GRCh38
rs557542970 Chr.1: 244844750-244844750 on Build GRCh38
rs368887269 Chr.1: 244844777-244844777 on Build GRCh38
rs188266796 Chr.1: 244844619-244844619 on Build GRCh38
rs1013885205 Chr.1: 244844666-244844666 on Build GRCh38
rs933915526 Chr.1: 244844416-244844416 on Build GRCh38
rs1051784124 Chr.1: 244844765-244844767 on Build GRCh38
rs181430818 Chr.1: 244844629-244844629 on Build GRCh38
rs911157449 Chr.1: 244844645-244844645 on Build GRCh38
rs547992617 Chr.1: 244844576-244844576 on Build GRCh38
rs77239968 Chr.1: 244844765-244844765 on Build GRCh38
rs907989318 Chr.1: 244844850-244844850 on Build GRCh38
rs185418438 Chr.1: 244844618-244844618 on Build GRCh38
rs527751822 Chr.1: 244844563-244844563 on Build GRCh38
rs1020140819 Chr.1: 244844857-244844857 on Build GRCh38
rs895752065 Chr.1: 244844830-244844830 on Build GRCh38
rs912899897 Chr.1: 244844625-244844625 on Build GRCh38
rs930722874 Chr.1: 244844683-244844683 on Build GRCh38
rs1018166615 Chr.1: 244844797-244844797 on Build GRCh38
rs562196723 Chr.1: 244844523-244844523 on Build GRCh38
rs780129762 Chr.1: 244844421-244844421 on Build GRCh38
rs1039722339 Chr.1: 244844608-244844608 on Build GRCh38
rs986452672 Chr.1: 244844794-244844794 on Build GRCh38
rs959908183 Chr.1: 244844662-244844662 on Build GRCh38
rs34289944 Chr.1: 244844779-244844780 on Build GRCh38
rs781666921 Chr.1: 244844713-244844713 on Build GRCh38
rs1016481279 Chr.1: 244844466-244844467 on Build GRCh38
rs999858751 Chr.1: 244844871-244844871 on Build GRCh38
rs541989344 Chr.1: 244844503-244844503 on Build GRCh38
rs1001365467 Chr.1: 244844651-244844653 on Build GRCh38
rs1047721509 Chr.1: 244844696-244844696 on Build GRCh38
rs933348944 Chr.1: 244844615-244844615 on Build GRCh38
rs189516393 Chr.1: 244844486-244844486 on Build GRCh38
rs955078538 Chr.1: 244844628-244844628 on Build GRCh38
rs555026821 Chr.1: 244844669-244844669 on Build GRCh38
rs756248304 Chr.1: 244844764-244844765 on Build GRCh38
rs919758231 Chr.1: 244844828-244844828 on Build GRCh38
rs572506013 Chr.1: 244844868-244844868 on Build GRCh38
rs184993621 Chr.1: 244844452-244844452 on Build GRCh38
rs955026672 Chr.1: 244844789-244844789 on Build GRCh38
rs778578348 Chr.1: 244844829-244844829 on Build GRCh38
rs556880389 Chr.1: 244844611-244844611 on Build GRCh38
rs942530528 Chr.1: 244844655-244844655 on Build GRCh38
rs536996774 Chr.1: 244844779-244844779 on Build GRCh38
rs868149257 Chr.1: 244844741-244844741 on Build GRCh38
rs1039104035 Chr.1: 244844477-244844477 on Build GRCh38
rs936835957 Chr.1: 244844597-244844597 on Build GRCh38
rs543275294 Chr.1: 244844472-244844472 on Build GRCh38
rs937172706 Chr.1: 244844821-244844821 on Build GRCh38
rs959730822 Chr.1: 244844507-244844507 on Build GRCh38
rs534371083 Chr.1: 244844725-244844725 on Build GRCh38
rs532222284 Chr.1: 244844721-244844721 on Build GRCh38
rs942560646 Chr.1: 244844453-244844453 on Build GRCh38
rs983528250 Chr.1: 244844836-244844836 on Build GRCh38
rs1038630084 Chr.1: 244844659-244844659 on Build GRCh38
rs1037772203 Chr.1: 244844647-244844647 on Build GRCh38
rs140206023 Chr.1: 244844866-244844866 on Build GRCh38
rs551811303 Chr.1: 244844736-244844736 on Build GRCh38
rs112391413 Chr.1: 244844614-244844614 on Build GRCh38
rs895021673 Chr.1: 244844683-244844700 on Build GRCh38
rs922183053 Chr.1: 244844747-244844747 on Build GRCh38
rs1045969640 Chr.1: 244844719-244844719 on Build GRCh38

View Full Product Details