Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 246743879-246744278 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SCCPDH
Entrez Gene ID
51097
Gene Name
saccharopine dehydrogenase (putative)
Gene Aliases
CGI-49, NET11
UniGene
Hs.498397
Gene Chromosome Location
Chr.1: 246724076-246768137 on Build GRCh38
Transcripts
NM_016002.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1748282 c.554C>T p.S185L Chr.1: 246744115-246744115 on Build GRCh38
6772610 c.527C>T p.A176V Chr.1: 246744088-246744088 on Build GRCh38
3486513 c.540C>T p.F180F Chr.1: 246744101-246744101 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs535509823 Chr.1: 246743999-246743999 on Build GRCh38
rs911727632 Chr.1: 246743944-246743944 on Build GRCh38
rs1056028850 Chr.1: 246744115-246744115 on Build GRCh38
rs373603494 Chr.1: 246743938-246743938 on Build GRCh38
rs767317874 Chr.1: 246744051-246744051 on Build GRCh38
rs748630635 Chr.1: 246744147-246744147 on Build GRCh38
rs765236254 Chr.1: 246744031-246744031 on Build GRCh38
rs1019733374 Chr.1: 246744229-246744229 on Build GRCh38
rs7513496 Chr.1: 246743904-246743904 on Build GRCh38
rs577431501 Chr.1: 246744251-246744251 on Build GRCh38
rs1014026476 Chr.1: 246743919-246743919 on Build GRCh38
rs557502680 Chr.1: 246744220-246744220 on Build GRCh38
rs763102089 Chr.1: 246744050-246744050 on Build GRCh38
rs940441221 Chr.1: 246744073-246744073 on Build GRCh38
rs993992642 Chr.1: 246743964-246743964 on Build GRCh38
rs767602130 Chr.1: 246744092-246744092 on Build GRCh38
rs17852359 Chr.1: 246744121-246744121 on Build GRCh38
rs190753539 Chr.1: 246744066-246744066 on Build GRCh38
rs769020383 Chr.1: 246744136-246744136 on Build GRCh38
rs935541443 Chr.1: 246744197-246744197 on Build GRCh38
rs967465860 Chr.1: 246743935-246743935 on Build GRCh38
rs1008310170 Chr.1: 246744208-246744208 on Build GRCh38
rs962543412 Chr.1: 246743956-246743956 on Build GRCh38
rs571056965 Chr.1: 246743911-246743911 on Build GRCh38
rs144263778 Chr.1: 246743990-246743990 on Build GRCh38
rs370581831 Chr.1: 246744135-246744135 on Build GRCh38
rs1020301766 Chr.1: 246744188-246744188 on Build GRCh38
rs761893083 Chr.1: 246744027-246744027 on Build GRCh38
rs868119045 Chr.1: 246743948-246743948 on Build GRCh38
rs143004003 Chr.1: 246744089-246744089 on Build GRCh38
rs572392917 Chr.1: 246744058-246744058 on Build GRCh38
rs1052507338 Chr.1: 246743903-246743903 on Build GRCh38
rs777055643 Chr.1: 246744139-246744139 on Build GRCh38
rs965902813 Chr.1: 246743987-246743987 on Build GRCh38
rs1016819863 Chr.1: 246743947-246743947 on Build GRCh38
rs879023065 Chr.1: 246744217-246744217 on Build GRCh38
rs1011543006 Chr.1: 246744153-246744153 on Build GRCh38
rs201020594 Chr.1: 246744047-246744047 on Build GRCh38
rs758036900 Chr.1: 246744122-246744122 on Build GRCh38
rs764936830 Chr.1: 246744018-246744018 on Build GRCh38
rs76587444 Chr.1: 246743997-246743997 on Build GRCh38
rs147652217 Chr.1: 246744075-246744075 on Build GRCh38
rs780368365 Chr.1: 246744131-246744131 on Build GRCh38
rs749173555 Chr.1: 246744032-246744032 on Build GRCh38
rs903226206 Chr.1: 246744191-246744191 on Build GRCh38
rs753029388 Chr.1: 246744104-246744117 on Build GRCh38
rs986929647 Chr.1: 246744014-246744014 on Build GRCh38
rs138814160 Chr.1: 246743998-246743998 on Build GRCh38
rs141926725 Chr.1: 246744108-246744108 on Build GRCh38

View Full Product Details