Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 248179904-248180420 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR2M2
Entrez Gene ID
391194
Gene Name
olfactory receptor family 2 subfamily M member 2
Gene Aliases
OR2M2Q, OST423
UniGene
Hs.534728
Gene Chromosome Location
Chr.1: 248179986-248181029 on Build GRCh38
Transcripts
NM_001004688.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5627734 c.112G>T p.A38S Chr.1: 248180097-248180097 on Build GRCh38
236516 c.139C>T p.L47F Chr.1: 248180124-248180124 on Build GRCh38
71970 c.17A>G p.Q6R Chr.1: 248180002-248180002 on Build GRCh38
3487010 c.128C>T p.S43F Chr.1: 248180113-248180113 on Build GRCh38
6878189 c.304T>A p.F102I Chr.1: 248180289-248180289 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768992900 Chr.1: 248180273-248180273 on Build GRCh38
rs144481353 Chr.1: 248180018-248180018 on Build GRCh38
rs772473592 Chr.1: 248180262-248180262 on Build GRCh38
rs773244655 Chr.1: 248180139-248180139 on Build GRCh38
rs9435919 Chr.1: 248180241-248180241 on Build GRCh38
rs149789397 Chr.1: 248180063-248180063 on Build GRCh38
rs375347393 Chr.1: 248180216-248180216 on Build GRCh38
rs891640490 Chr.1: 248180288-248180288 on Build GRCh38
rs776463824 Chr.1: 248179954-248179954 on Build GRCh38
rs386641960 Chr.1: 248180239-248180241 on Build GRCh38
rs771012232 Chr.1: 248180385-248180385 on Build GRCh38
rs200062907 Chr.1: 248180187-248180187 on Build GRCh38
rs760969800 Chr.1: 248180340-248180340 on Build GRCh38
rs148878003 Chr.1: 248180075-248180075 on Build GRCh38
rs146796261 Chr.1: 248180059-248180059 on Build GRCh38
rs761347728 Chr.1: 248179955-248179955 on Build GRCh38
rs568039481 Chr.1: 248180132-248180132 on Build GRCh38
rs200932016 Chr.1: 248180089-248180089 on Build GRCh38
rs747296081 Chr.1: 248180346-248180346 on Build GRCh38
rs374044525 Chr.1: 248180388-248180388 on Build GRCh38
rs386641959 Chr.1: 248180211-248180213 on Build GRCh38
rs753353772 Chr.1: 248180329-248180329 on Build GRCh38
rs773551153 Chr.1: 248180291-248180291 on Build GRCh38
rs1043945042 Chr.1: 248180271-248180271 on Build GRCh38
rs970245791 Chr.1: 248180312-248180312 on Build GRCh38
rs1037090915 Chr.1: 248180202-248180202 on Build GRCh38
rs138681109 Chr.1: 248180153-248180153 on Build GRCh38
rs1014034130 Chr.1: 248180061-248180061 on Build GRCh38
rs768276374 Chr.1: 248179951-248179951 on Build GRCh38
rs145713159 Chr.1: 248180060-248180060 on Build GRCh38
rs375841086 Chr.1: 248179937-248179937 on Build GRCh38
rs766881231 Chr.1: 248179983-248179983 on Build GRCh38
rs747621126 Chr.1: 248180103-248180103 on Build GRCh38
rs765566768 Chr.1: 248180284-248180284 on Build GRCh38
rs777318730 Chr.1: 248180219-248180219 on Build GRCh38
rs769987965 Chr.1: 248180354-248180354 on Build GRCh38
rs772574077 Chr.1: 248180161-248180161 on Build GRCh38
rs965719032 Chr.1: 248180085-248180085 on Build GRCh38
rs775390459 Chr.1: 248180392-248180392 on Build GRCh38
rs771283691 Chr.1: 248180056-248180056 on Build GRCh38
rs569581349 Chr.1: 248180051-248180051 on Build GRCh38
rs1004220556 Chr.1: 248180022-248180022 on Build GRCh38
rs566573970 Chr.1: 248180300-248180300 on Build GRCh38
rs534047479 Chr.1: 248180302-248180303 on Build GRCh38
rs563979686 Chr.1: 248180349-248180349 on Build GRCh38
rs750073988 Chr.1: 248180337-248180337 on Build GRCh38
rs868746148 Chr.1: 248180272-248180272 on Build GRCh38
rs746245069 Chr.1: 248180246-248180246 on Build GRCh38
rs139067314 Chr.1: 248180210-248180210 on Build GRCh38
rs759682878 Chr.1: 248179987-248179987 on Build GRCh38
rs778595351 Chr.1: 248180333-248180333 on Build GRCh38
rs749121760 Chr.1: 248180272-248180273 on Build GRCh38
rs973630780 Chr.1: 248180119-248180119 on Build GRCh38
rs751350435 Chr.1: 248180083-248180083 on Build GRCh38
rs778976585 Chr.1: 248180303-248180304 on Build GRCh38
rs766685038 Chr.1: 248180348-248180348 on Build GRCh38
rs201832076 Chr.1: 248180029-248180029 on Build GRCh38
rs372848803 Chr.1: 248180054-248180054 on Build GRCh38
rs192342576 Chr.1: 248180222-248180222 on Build GRCh38
rs763902972 Chr.1: 248180327-248180327 on Build GRCh38
rs1048473015 Chr.1: 248179936-248179936 on Build GRCh38
rs963663282 Chr.1: 248180114-248180114 on Build GRCh38
rs200421339 Chr.1: 248180331-248180331 on Build GRCh38
rs766252365 Chr.1: 248180080-248180080 on Build GRCh38
rs748910573 Chr.1: 248180131-248180131 on Build GRCh38
rs372047333 Chr.1: 248180015-248180015 on Build GRCh38
rs756316643 Chr.1: 248180014-248180014 on Build GRCh38
rs759468683 Chr.1: 248180296-248180296 on Build GRCh38
rs147227258 Chr.1: 248180030-248180030 on Build GRCh38
rs746532195 Chr.1: 248179949-248179949 on Build GRCh38
rs142679516 Chr.1: 248180185-248180185 on Build GRCh38
rs751744630 Chr.1: 248180047-248180048 on Build GRCh38
rs760725854 Chr.1: 248180177-248180177 on Build GRCh38
rs779999518 Chr.1: 248179969-248179970 on Build GRCh38
rs201343435 Chr.1: 248180372-248180372 on Build GRCh38
rs780488430 Chr.1: 248180345-248180345 on Build GRCh38
rs777503138 Chr.1: 248180088-248180088 on Build GRCh38
rs140281039 Chr.1: 248180016-248180016 on Build GRCh38
rs754995992 Chr.1: 248180088-248180088 on Build GRCh38
rs1024447172 Chr.1: 248180074-248180074 on Build GRCh38
rs746194128 Chr.1: 248180158-248180158 on Build GRCh38
rs776881155 Chr.1: 248180274-248180274 on Build GRCh38
rs148032327 Chr.1: 248180143-248180143 on Build GRCh38
rs143926960 Chr.1: 248180217-248180217 on Build GRCh38
rs761787327 Chr.1: 248180042-248180042 on Build GRCh38
rs747650826 Chr.1: 248180035-248180035 on Build GRCh38
rs144359026 Chr.1: 248180215-248180215 on Build GRCh38
rs142698993 Chr.1: 248180350-248180350 on Build GRCh38
rs780206895 Chr.1: 248180017-248180017 on Build GRCh38
rs747527899 Chr.1: 248180268-248180268 on Build GRCh38
rs771936111 Chr.1: 248179943-248179943 on Build GRCh38
rs755634605 Chr.1: 248180104-248180104 on Build GRCh38
rs758731719 Chr.1: 248180344-248180344 on Build GRCh38
rs753285621 Chr.1: 248180275-248180275 on Build GRCh38
rs539993597 Chr.1: 248180052-248180052 on Build GRCh38
rs774882674 Chr.1: 248180057-248180057 on Build GRCh38
rs940130237 Chr.1: 248180186-248180186 on Build GRCh38
rs781114023 Chr.1: 248180092-248180092 on Build GRCh38
rs61156873 Chr.1: 248180245-248180245 on Build GRCh38
rs780070308 Chr.1: 248179945-248179945 on Build GRCh38
rs769437782 Chr.1: 248179971-248179971 on Build GRCh38
rs199810194 Chr.1: 248180358-248180358 on Build GRCh38
rs190830713 Chr.1: 248179985-248179985 on Build GRCh38
rs141475463 Chr.1: 248180041-248180041 on Build GRCh38
rs775930351 Chr.1: 248180266-248180266 on Build GRCh38
rs200933239 Chr.1: 248180004-248180004 on Build GRCh38
rs764201656 Chr.1: 248180180-248180180 on Build GRCh38
rs760686932 Chr.1: 248180311-248180311 on Build GRCh38
rs764238697 Chr.1: 248180073-248180073 on Build GRCh38
rs754228786 Chr.1: 248180386-248180386 on Build GRCh38
rs776765473 Chr.1: 248180181-248180181 on Build GRCh38
rs776007697 Chr.1: 248180166-248180166 on Build GRCh38
rs767557930 Chr.1: 248179992-248179992 on Build GRCh38
rs763308358 Chr.1: 248179978-248179978 on Build GRCh38
rs763217545 Chr.1: 248180380-248180380 on Build GRCh38
rs1051872902 Chr.1: 248180165-248180165 on Build GRCh38
rs558437234 Chr.1: 248180054-248180054 on Build GRCh38
rs770843804 Chr.1: 248180287-248180301 on Build GRCh38
rs1024065657 Chr.1: 248180306-248180306 on Build GRCh38
rs200093216 Chr.1: 248179972-248179972 on Build GRCh38
rs146197738 Chr.1: 248180298-248180298 on Build GRCh38
rs752628601 Chr.1: 248180302-248180302 on Build GRCh38
rs758306379 Chr.1: 248180078-248180078 on Build GRCh38
rs1052953931 Chr.1: 248180287-248180287 on Build GRCh38
rs34678055 Chr.1: 248180050-248180051 on Build GRCh38
rs754757872 Chr.1: 248180213-248180213 on Build GRCh38
rs369271344 Chr.1: 248180278-248180278 on Build GRCh38
rs1052265847 Chr.1: 248180048-248180048 on Build GRCh38

View Full Product Details