Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 248855633-248856122 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF692
Entrez Gene ID
55657
Gene Name
zinc finger protein 692
Gene Aliases
AREBP, Zfp692
UniGene
Hs.377705
Gene Chromosome Location
Chr.1: 248850004-248860759 on Build GRCh38
Transcripts
NM_001136036.2 NM_001193328.1 NM_017865.3 XM_011544220.1 XM_011544221.1 XM_011544222.1 XM_011544223.2 XM_011544227.1 XM_017001698.1 XM_017001699.1 XM_017001700.1 XM_017001701.1 XM_017001702.1 XM_017001703.1 XM_017001704.1 XM_017001705.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1185818 c.743C>T p.A248V Chr.1: 248855863-248855863 on Build GRCh38
6832487 c.882-5C>A p.? Chr.1: 248855640-248855640 on Build GRCh38
907409 c.695C>A p.P232H Chr.1: 248855911-248855911 on Build GRCh38
2234135 c.882-4G>A p.? Chr.1: 248855639-248855639 on Build GRCh38
6236629 c.761C>A p.A254D Chr.1: 248855845-248855845 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763152595 Chr.1: 248855891-248855891 on Build GRCh38
rs751559099 Chr.1: 248855736-248855736 on Build GRCh38
rs1022404354 Chr.1: 248856051-248856051 on Build GRCh38
rs376322471 Chr.1: 248855769-248855769 on Build GRCh38
rs772520912 Chr.1: 248855865-248855865 on Build GRCh38
rs764273651 Chr.1: 248855714-248855714 on Build GRCh38
rs756789831 Chr.1: 248855830-248855830 on Build GRCh38
rs901173786 Chr.1: 248855991-248855991 on Build GRCh38
rs746761126 Chr.1: 248855667-248855667 on Build GRCh38
rs56106447 Chr.1: 248855670-248855670 on Build GRCh38
rs149636647 Chr.1: 248855934-248855934 on Build GRCh38
rs201502808 Chr.1: 248855724-248855724 on Build GRCh38
rs144605555 Chr.1: 248855839-248855839 on Build GRCh38
rs902797200 Chr.1: 248856057-248856057 on Build GRCh38
rs189959177 Chr.1: 248856017-248856017 on Build GRCh38
rs541550111 Chr.1: 248855956-248855956 on Build GRCh38
rs551986367 Chr.1: 248855799-248855799 on Build GRCh38
rs754320102 Chr.1: 248855703-248855704 on Build GRCh38
rs752477387 Chr.1: 248855751-248855751 on Build GRCh38
rs773408037 Chr.1: 248855890-248855890 on Build GRCh38
rs200437158 Chr.1: 248855690-248855690 on Build GRCh38
rs371122838 Chr.1: 248855691-248855691 on Build GRCh38
rs375365732 Chr.1: 248855752-248855752 on Build GRCh38
rs1006873361 Chr.1: 248855870-248855870 on Build GRCh38
rs368069363 Chr.1: 248855938-248855938 on Build GRCh38
rs772751428 Chr.1: 248855676-248855676 on Build GRCh38
rs990320221 Chr.1: 248856044-248856044 on Build GRCh38
rs145545056 Chr.1: 248855911-248855911 on Build GRCh38
rs140745254 Chr.1: 248855823-248855823 on Build GRCh38
rs757317426 Chr.1: 248855857-248855858 on Build GRCh38
rs780554191 Chr.1: 248855958-248855958 on Build GRCh38
rs149407405 Chr.1: 248855733-248855733 on Build GRCh38
rs746369683 Chr.1: 248855783-248855783 on Build GRCh38
rs748767951 Chr.1: 248855680-248855680 on Build GRCh38
rs753671789 Chr.1: 248855926-248855926 on Build GRCh38
rs992011816 Chr.1: 248855931-248855931 on Build GRCh38
rs777319905 Chr.1: 248855754-248855754 on Build GRCh38
rs753474261 Chr.1: 248855838-248855838 on Build GRCh38
rs747334413 Chr.1: 248855880-248855880 on Build GRCh38
rs775899584 Chr.1: 248855788-248855788 on Build GRCh38
rs760971005 Chr.1: 248855797-248855797 on Build GRCh38
rs144577893 Chr.1: 248855930-248855930 on Build GRCh38
rs1049815853 Chr.1: 248856012-248856012 on Build GRCh38
rs755028449 Chr.1: 248855744-248855744 on Build GRCh38
rs1051370745 Chr.1: 248856074-248856074 on Build GRCh38
rs760918890 Chr.1: 248855705-248855705 on Build GRCh38
rs925058803 Chr.1: 248856008-248856008 on Build GRCh38
rs769401689 Chr.1: 248855675-248855675 on Build GRCh38
rs779956211 Chr.1: 248856065-248856065 on Build GRCh38
rs768917141 Chr.1: 248855883-248855883 on Build GRCh38
rs969485534 Chr.1: 248855944-248855944 on Build GRCh38
rs752493862 Chr.1: 248855826-248855826 on Build GRCh38
rs747828016 Chr.1: 248855673-248855673 on Build GRCh38
rs202234605 Chr.1: 248855701-248855701 on Build GRCh38
rs546231655 Chr.1: 248855855-248855855 on Build GRCh38
rs374297190 Chr.1: 248855692-248855692 on Build GRCh38
rs913379672 Chr.1: 248856063-248856063 on Build GRCh38
rs767854754 Chr.1: 248855700-248855700 on Build GRCh38
rs199939289 Chr.1: 248855928-248855928 on Build GRCh38
rs779542496 Chr.1: 248855809-248855809 on Build GRCh38
rs947844944 Chr.1: 248855759-248855759 on Build GRCh38
rs745430605 Chr.1: 248855861-248855861 on Build GRCh38
rs770437402 Chr.1: 248855771-248855771 on Build GRCh38
rs765398736 Chr.1: 248855806-248855806 on Build GRCh38
rs371975767 Chr.1: 248855730-248855730 on Build GRCh38
rs1051114896 Chr.1: 248855862-248855862 on Build GRCh38
rs749059735 Chr.1: 248856084-248856084 on Build GRCh38
rs193006136 Chr.1: 248856035-248856035 on Build GRCh38
rs1035238673 Chr.1: 248856077-248856077 on Build GRCh38
rs13313009 Chr.1: 248856006-248856006 on Build GRCh38
rs867948263 Chr.1: 248855936-248855936 on Build GRCh38
rs1018346979 Chr.1: 248855668-248855668 on Build GRCh38
rs200124290 Chr.1: 248855672-248855672 on Build GRCh38
rs759517802 Chr.1: 248855821-248855821 on Build GRCh38
rs770185355 Chr.1: 248855889-248855889 on Build GRCh38
rs762124843 Chr.1: 248855888-248855888 on Build GRCh38
rs377420863 Chr.1: 248855686-248855686 on Build GRCh38
rs777423632 Chr.1: 248855832-248855832 on Build GRCh38
rs13313088 Chr.1: 248855917-248855917 on Build GRCh38
rs778452229 Chr.1: 248855781-248855781 on Build GRCh38
rs144411823 Chr.1: 248856080-248856080 on Build GRCh38
rs781133318 Chr.1: 248855750-248855750 on Build GRCh38
rs768617761 Chr.1: 248856093-248856093 on Build GRCh38
rs372241595 Chr.1: 248855971-248855971 on Build GRCh38
rs748576242 Chr.1: 248855989-248855989 on Build GRCh38
rs777114657 Chr.1: 248855885-248855885 on Build GRCh38
rs772605168 Chr.1: 248855784-248855784 on Build GRCh38
rs1028326783 Chr.1: 248855720-248855720 on Build GRCh38
rs568999759 Chr.1: 248855964-248855964 on Build GRCh38
rs770095521 Chr.1: 248855993-248855993 on Build GRCh38
rs778362635 Chr.1: 248855843-248855843 on Build GRCh38
rs992464427 Chr.1: 248856095-248856095 on Build GRCh38
rs781668524 Chr.1: 248855986-248855986 on Build GRCh38
rs150141583 Chr.1: 248855905-248855905 on Build GRCh38
rs138288849 Chr.1: 248855805-248855805 on Build GRCh38
rs780164764 Chr.1: 248855652-248855652 on Build GRCh38
rs750601922 Chr.1: 248855814-248855814 on Build GRCh38
rs375816292 Chr.1: 248855803-248855803 on Build GRCh38
rs780457451 Chr.1: 248855866-248855866 on Build GRCh38
rs776722798 Chr.1: 248855798-248855798 on Build GRCh38

View Full Product Details