Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 5639154-5639650 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASB13
Entrez Gene ID
79754
Gene Name
ankyrin repeat and SOCS box containing 13
Gene Aliases
-
UniGene
Hs.445899
Gene Chromosome Location
Chr.10: 5638857-5666595 on Build GRCh38
Transcripts
NM_024701.3 NR_024581.1 NR_037164.1

SNP Detail

SNP ID Chromosome Location
rs1019259071 Chr.10: 5639271-5639271 on Build GRCh38
rs567899803 Chr.10: 5639551-5639551 on Build GRCh38
rs1051042390 Chr.10: 5639303-5639303 on Build GRCh38
rs528027051 Chr.10: 5639548-5639548 on Build GRCh38
rs7901492 Chr.10: 5639385-5639385 on Build GRCh38
rs185058931 Chr.10: 5639609-5639609 on Build GRCh38
rs551977648 Chr.10: 5639421-5639421 on Build GRCh38
rs932542122 Chr.10: 5639299-5639299 on Build GRCh38
rs374771648 Chr.10: 5639489-5639489 on Build GRCh38
rs141053750 Chr.10: 5639210-5639210 on Build GRCh38
rs772593955 Chr.10: 5639209-5639209 on Build GRCh38
rs557974912 Chr.10: 5639615-5639615 on Build GRCh38
rs924694899 Chr.10: 5639422-5639422 on Build GRCh38
rs374500796 Chr.10: 5639261-5639261 on Build GRCh38
rs931345933 Chr.10: 5639505-5639505 on Build GRCh38
rs921257441 Chr.10: 5639481-5639481 on Build GRCh38
rs1025019811 Chr.10: 5639396-5639396 on Build GRCh38
rs995111449 Chr.10: 5639367-5639367 on Build GRCh38
rs898353408 Chr.10: 5639557-5639557 on Build GRCh38
rs61225117 Chr.10: 5639330-5639330 on Build GRCh38
rs57467389 Chr.10: 5639608-5639608 on Build GRCh38
rs995206115 Chr.10: 5639563-5639563 on Build GRCh38
rs549752963 Chr.10: 5639596-5639596 on Build GRCh38
rs547187775 Chr.10: 5639180-5639180 on Build GRCh38
rs549418908 Chr.10: 5639549-5639549 on Build GRCh38
rs556262915 Chr.10: 5639208-5639208 on Build GRCh38
rs558818885 Chr.10: 5639211-5639211 on Build GRCh38
rs1050959511 Chr.10: 5639512-5639512 on Build GRCh38
rs1024528860 Chr.10: 5639592-5639592 on Build GRCh38
rs1049875588 Chr.10: 5639213-5639213 on Build GRCh38
rs1023120724 Chr.10: 5639591-5639591 on Build GRCh38
rs868257996 Chr.10: 5639577-5639577 on Build GRCh38
rs936051016 Chr.10: 5639426-5639426 on Build GRCh38
rs371897607 Chr.10: 5639273-5639273 on Build GRCh38
rs561977346 Chr.10: 5639343-5639343 on Build GRCh38
rs539284636 Chr.10: 5639216-5639216 on Build GRCh38
rs765185154 Chr.10: 5639570-5639570 on Build GRCh38
rs1015309160 Chr.10: 5639336-5639336 on Build GRCh38
rs7908974 Chr.10: 5639427-5639427 on Build GRCh38
rs932678009 Chr.10: 5639511-5639511 on Build GRCh38
rs192266135 Chr.10: 5639418-5639418 on Build GRCh38
rs187938497 Chr.10: 5639304-5639304 on Build GRCh38
rs1011271633 Chr.10: 5639586-5639586 on Build GRCh38
rs150253226 Chr.10: 5639286-5639286 on Build GRCh38
rs77761888 Chr.10: 5639571-5639571 on Build GRCh38
rs1050257228 Chr.10: 5639507-5639507 on Build GRCh38
rs912477089 Chr.10: 5639531-5639531 on Build GRCh38
rs991695536 Chr.10: 5639395-5639395 on Build GRCh38
rs1007421355 Chr.10: 5639246-5639246 on Build GRCh38
rs138580570 Chr.10: 5639408-5639408 on Build GRCh38
rs957069955 Chr.10: 5639298-5639298 on Build GRCh38
rs117090098 Chr.10: 5639344-5639344 on Build GRCh38
rs940008637 Chr.10: 5639196-5639196 on Build GRCh38
rs201026861 Chr.10: 5639282-5639283 on Build GRCh38
rs977491012 Chr.10: 5639202-5639202 on Build GRCh38
rs1033290580 Chr.10: 5639374-5639374 on Build GRCh38
rs544198735 Chr.10: 5639366-5639366 on Build GRCh38
rs12573500 Chr.10: 5639176-5639176 on Build GRCh38
rs761935662 Chr.10: 5639469-5639469 on Build GRCh38

View Full Product Details