Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 27735135-27735649 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MKX
Entrez Gene ID
283078
Gene Name
mohawk homeobox
Gene Aliases
C10orf48, IFRX, IRXL1
UniGene
Hs.128193
Gene Chromosome Location
Chr.10: 27672874-27745849 on Build GRCh38
Transcripts
NM_001242702.1 NM_173576.2 XM_017016105.1 XM_017016106.1 XM_017016107.1 XM_017016108.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1257757 c.398G>A p.R133Q Chr.10: 27735325-27735325 on Build GRCh38
684589 c.349G>A p.V117M Chr.10: 27735374-27735374 on Build GRCh38
3436976 c.467G>A p.R156Q Chr.10: 27735256-27735256 on Build GRCh38
3436977 c.395T>C p.V132A Chr.10: 27735328-27735328 on Build GRCh38
3436975 c.502G>A p.D168N Chr.10: 27735221-27735221 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370766157 Chr.10: 27735592-27735592 on Build GRCh38
rs190790004 Chr.10: 27735256-27735256 on Build GRCh38
rs748997066 Chr.10: 27735257-27735257 on Build GRCh38
rs768432622 Chr.10: 27735565-27735565 on Build GRCh38
rs570486813 Chr.10: 27735164-27735164 on Build GRCh38
rs891249446 Chr.10: 27735194-27735194 on Build GRCh38
rs897134065 Chr.10: 27735229-27735229 on Build GRCh38
rs762056002 Chr.10: 27735343-27735343 on Build GRCh38
rs778649989 Chr.10: 27735422-27735422 on Build GRCh38
rs766648845 Chr.10: 27735369-27735369 on Build GRCh38
rs1009972472 Chr.10: 27735434-27735434 on Build GRCh38
rs770657390 Chr.10: 27735272-27735272 on Build GRCh38
rs912557253 Chr.10: 27735600-27735600 on Build GRCh38
rs200930878 Chr.10: 27735332-27735332 on Build GRCh38
rs182085022 Chr.10: 27735418-27735418 on Build GRCh38
rs141823732 Chr.10: 27735510-27735510 on Build GRCh38
rs145362609 Chr.10: 27735224-27735224 on Build GRCh38
rs761988512 Chr.10: 27735213-27735213 on Build GRCh38
rs979665187 Chr.10: 27735499-27735499 on Build GRCh38
rs776528222 Chr.10: 27735342-27735342 on Build GRCh38
rs868638409 Chr.10: 27735219-27735219 on Build GRCh38
rs750804503 Chr.10: 27735344-27735344 on Build GRCh38
rs868332651 Chr.10: 27735363-27735363 on Build GRCh38
rs369678208 Chr.10: 27735249-27735249 on Build GRCh38
rs988297039 Chr.10: 27735445-27735445 on Build GRCh38
rs750749306 Chr.10: 27735225-27735225 on Build GRCh38
rs113819945 Chr.10: 27735583-27735583 on Build GRCh38
rs932635443 Chr.10: 27735529-27735529 on Build GRCh38
rs921536535 Chr.10: 27735513-27735513 on Build GRCh38
rs144375154 Chr.10: 27735223-27735223 on Build GRCh38
rs1000278003 Chr.10: 27735551-27735551 on Build GRCh38
rs764062798 Chr.10: 27735197-27735197 on Build GRCh38
rs1034726188 Chr.10: 27735435-27735435 on Build GRCh38
rs34806745 Chr.10: 27735248-27735248 on Build GRCh38
rs778882954 Chr.10: 27735273-27735273 on Build GRCh38
rs200079170 Chr.10: 27735385-27735385 on Build GRCh38
rs185887031 Chr.10: 27735538-27735538 on Build GRCh38
rs374145415 Chr.10: 27735325-27735325 on Build GRCh38
rs1032557425 Chr.10: 27735579-27735579 on Build GRCh38
rs940097278 Chr.10: 27735606-27735606 on Build GRCh38
rs960454949 Chr.10: 27735441-27735441 on Build GRCh38
rs1021097624 Chr.10: 27735455-27735455 on Build GRCh38
rs755418682 Chr.10: 27735384-27735384 on Build GRCh38
rs754153747 Chr.10: 27735204-27735204 on Build GRCh38
rs546140690 Chr.10: 27735448-27735448 on Build GRCh38
rs765614697 Chr.10: 27735214-27735214 on Build GRCh38
rs986832677 Chr.10: 27735561-27735561 on Build GRCh38
rs772092632 Chr.10: 27735174-27735174 on Build GRCh38
rs752150488 Chr.10: 27735382-27735382 on Build GRCh38
rs758914357 Chr.10: 27735230-27735230 on Build GRCh38
rs149049062 Chr.10: 27735242-27735242 on Build GRCh38
rs151140216 Chr.10: 27735329-27735329 on Build GRCh38
rs575454652 Chr.10: 27735428-27735428 on Build GRCh38
rs763367127 Chr.10: 27735354-27735354 on Build GRCh38
rs753356998 Chr.10: 27735401-27735401 on Build GRCh38
rs968632053 Chr.10: 27735494-27735494 on Build GRCh38
rs760582157 Chr.10: 27735183-27735183 on Build GRCh38
rs745591021 Chr.10: 27735283-27735283 on Build GRCh38
rs766976774 Chr.10: 27735367-27735367 on Build GRCh38
rs377601436 Chr.10: 27735424-27735424 on Build GRCh38
rs760522501 Chr.10: 27735330-27735330 on Build GRCh38

View Full Product Details