Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 73912094-73912572 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C10orf55
Entrez Gene ID
414236
Gene Name
chromosome 10 open reading frame 55
Gene Aliases
bA417O11.3
UniGene
Hs.585453
Gene Chromosome Location
Chr.10: 73909969-73922777 on Build GRCh38
Transcripts
NM_001001791.2 NM_001145031.2 NM_001319191.1 NM_002658.4 XM_011539866.2

Gene Symbol
PLAU
Entrez Gene ID
5328
Gene Name
plasminogen activator, urokinase
Gene Aliases
ATF, BDPLT5, QPD, UPA, URK, u-PA
UniGene
Hs.77274
Gene Chromosome Location
Chr.10: 73909182-73917501 on Build GRCh38
Transcripts
NM_001001791.2 NM_001145031.2 NM_001319191.1 NM_002658.4 XM_011539866.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5837316 c.87G>A p.S29S Chr.10: 73912216-73912216 on Build GRCh38
1349096 c.171C>T p.F57F Chr.10: 73912300-73912300 on Build GRCh38
5837318 c.29C>T p.S10L Chr.10: 73912216-73912216 on Build GRCh38
5837319 c.98G>A p.C33Y Chr.10: 73912227-73912227 on Build GRCh38
5483485 c.179A>G p.Q60R Chr.10: 73912308-73912308 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775296618 Chr.10: 73912251-73912251 on Build GRCh38
rs1002137585 Chr.10: 73912538-73912538 on Build GRCh38
rs200927138 Chr.10: 73912240-73912240 on Build GRCh38
rs774324202 Chr.10: 73912153-73912153 on Build GRCh38
rs372720180 Chr.10: 73912194-73912194 on Build GRCh38
rs760952116 Chr.10: 73912466-73912466 on Build GRCh38
rs767790362 Chr.10: 73912335-73912335 on Build GRCh38
rs755164208 Chr.10: 73912329-73912329 on Build GRCh38
rs1014074238 Chr.10: 73912545-73912545 on Build GRCh38
rs773881195 Chr.10: 73912379-73912379 on Build GRCh38
rs892303177 Chr.10: 73912158-73912158 on Build GRCh38
rs374232402 Chr.10: 73912260-73912260 on Build GRCh38
rs770661625 Chr.10: 73912134-73912134 on Build GRCh38
rs1011294897 Chr.10: 73912141-73912141 on Build GRCh38
rs755285429 Chr.10: 73912186-73912186 on Build GRCh38
rs895666064 Chr.10: 73912531-73912531 on Build GRCh38
rs147372618 Chr.10: 73912292-73912292 on Build GRCh38
rs936296982 Chr.10: 73912434-73912434 on Build GRCh38
rs888800813 Chr.10: 73912394-73912394 on Build GRCh38
rs764051398 Chr.10: 73912272-73912272 on Build GRCh38
rs2227581 Chr.10: 73912117-73912117 on Build GRCh38
rs751776753 Chr.10: 73912318-73912318 on Build GRCh38
rs774129403 Chr.10: 73912216-73912216 on Build GRCh38
rs1034339928 Chr.10: 73912179-73912179 on Build GRCh38
rs568507000 Chr.10: 73912516-73912516 on Build GRCh38
rs756588363 Chr.10: 73912193-73912193 on Build GRCh38
rs866831923 Chr.10: 73912364-73912364 on Build GRCh38
rs142059320 Chr.10: 73912291-73912291 on Build GRCh38
rs760592972 Chr.10: 73912160-73912160 on Build GRCh38
rs778398901 Chr.10: 73912363-73912363 on Build GRCh38
rs1046247003 Chr.10: 73912473-73912473 on Build GRCh38
rs1055657823 Chr.10: 73912511-73912511 on Build GRCh38
rs762710927 Chr.10: 73912133-73912133 on Build GRCh38
rs780219863 Chr.10: 73912183-73912183 on Build GRCh38
rs758648701 Chr.10: 73912177-73912177 on Build GRCh38
rs55744193 Chr.10: 73912301-73912301 on Build GRCh38
rs5786128 Chr.10: 73912161-73912161 on Build GRCh38
rs2227557 Chr.10: 73912472-73912472 on Build GRCh38
rs748612292 Chr.10: 73912189-73912189 on Build GRCh38
rs753060051 Chr.10: 73912341-73912341 on Build GRCh38
rs754283224 Chr.10: 73912352-73912352 on Build GRCh38
rs772716620 Chr.10: 73912128-73912128 on Build GRCh38
rs148701277 Chr.10: 73912159-73912159 on Build GRCh38
rs759380746 Chr.10: 73912157-73912157 on Build GRCh38
rs55695810 Chr.10: 73912467-73912467 on Build GRCh38
rs781782139 Chr.10: 73912188-73912188 on Build GRCh38
rs146168437 Chr.10: 73912375-73912375 on Build GRCh38
rs755452241 Chr.10: 73912447-73912447 on Build GRCh38
rs747752586 Chr.10: 73912116-73912116 on Build GRCh38
rs533020384 Chr.10: 73912413-73912413 on Build GRCh38
rs1025371355 Chr.10: 73912248-73912248 on Build GRCh38
rs543197904 Chr.10: 73912429-73912429 on Build GRCh38
rs750452037 Chr.10: 73912170-73912170 on Build GRCh38
rs748862531 Chr.10: 73912367-73912367 on Build GRCh38
rs115703038 Chr.10: 73912488-73912488 on Build GRCh38
rs1044084049 Chr.10: 73912470-73912470 on Build GRCh38
rs765244714 Chr.10: 73912163-73912163 on Build GRCh38
rs748772626 Chr.10: 73912198-73912198 on Build GRCh38
rs370557565 Chr.10: 73912215-73912215 on Build GRCh38
rs375409521 Chr.10: 73912300-73912300 on Build GRCh38

View Full Product Details