Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 79382190-79382703 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZCCHC24
Entrez Gene ID
219654
Gene Name
zinc finger CCHC-type containing 24
Gene Aliases
C10orf56, Z3CXXC8
UniGene
Hs.523080
Gene Chromosome Location
Chr.10: 79382325-79445627 on Build GRCh38
Transcripts
NM_153367.3

SNP Detail

SNP ID Chromosome Location
rs80126415 Chr.10: 79382329-79382329 on Build GRCh38
rs572889462 Chr.10: 79382567-79382567 on Build GRCh38
rs962528720 Chr.10: 79382454-79382454 on Build GRCh38
rs542335363 Chr.10: 79382535-79382535 on Build GRCh38
rs1022083468 Chr.10: 79382376-79382376 on Build GRCh38
rs11002951 Chr.10: 79382489-79382489 on Build GRCh38
rs929987778 Chr.10: 79382476-79382476 on Build GRCh38
rs974113695 Chr.10: 79382465-79382465 on Build GRCh38
rs565578645 Chr.10: 79382622-79382622 on Build GRCh38
rs6858 Chr.10: 79382609-79382609 on Build GRCh38
rs749599872 Chr.10: 79382385-79382385 on Build GRCh38
rs960981228 Chr.10: 79382571-79382571 on Build GRCh38
rs961324298 Chr.10: 79382593-79382593 on Build GRCh38
rs865784163 Chr.10: 79382355-79382355 on Build GRCh38
rs1050301980 Chr.10: 79382640-79382640 on Build GRCh38
rs902294457 Chr.10: 79382551-79382551 on Build GRCh38
rs576080556 Chr.10: 79382557-79382557 on Build GRCh38
rs768644098 Chr.10: 79382561-79382561 on Build GRCh38
rs1018173918 Chr.10: 79382580-79382580 on Build GRCh38
rs7085536 Chr.10: 79382292-79382292 on Build GRCh38
rs748375312 Chr.10: 79382543-79382543 on Build GRCh38
rs940146098 Chr.10: 79382405-79382405 on Build GRCh38
rs746687056 Chr.10: 79382531-79382531 on Build GRCh38
rs893327347 Chr.10: 79382322-79382322 on Build GRCh38
rs6480934 Chr.10: 79382453-79382453 on Build GRCh38
rs551198875 Chr.10: 79382669-79382669 on Build GRCh38
rs987906119 Chr.10: 79382267-79382267 on Build GRCh38
rs781179566 Chr.10: 79382253-79382253 on Build GRCh38
rs370261124 Chr.10: 79382506-79382506 on Build GRCh38
rs531067620 Chr.10: 79382647-79382647 on Build GRCh38
rs939589131 Chr.10: 79382368-79382368 on Build GRCh38
rs577488129 Chr.10: 79382483-79382483 on Build GRCh38
rs567762133 Chr.10: 79382366-79382366 on Build GRCh38
rs536341560 Chr.10: 79382375-79382375 on Build GRCh38
rs375410917 Chr.10: 79382566-79382566 on Build GRCh38
rs372562657 Chr.10: 79382451-79382451 on Build GRCh38
rs541141968 Chr.10: 79382490-79382490 on Build GRCh38
rs928598245 Chr.10: 79382648-79382648 on Build GRCh38
rs557647030 Chr.10: 79382458-79382458 on Build GRCh38
rs1043494687 Chr.10: 79382547-79382547 on Build GRCh38
rs556867867 Chr.10: 79382496-79382496 on Build GRCh38
rs1029571631 Chr.10: 79382411-79382411 on Build GRCh38
rs867759121 Chr.10: 79382467-79382467 on Build GRCh38
rs566509149 Chr.10: 79382439-79382439 on Build GRCh38
rs1038415310 Chr.10: 79382437-79382437 on Build GRCh38
rs918693643 Chr.10: 79382472-79382472 on Build GRCh38
rs771312204 Chr.10: 79382418-79382418 on Build GRCh38
rs573727030 Chr.10: 79382534-79382534 on Build GRCh38
rs899000313 Chr.10: 79382445-79382445 on Build GRCh38
rs139364160 Chr.10: 79382300-79382300 on Build GRCh38
rs547021731 Chr.10: 79382380-79382380 on Build GRCh38
rs781255204 Chr.10: 79382417-79382417 on Build GRCh38
rs7909640 Chr.10: 79382240-79382240 on Build GRCh38
rs1008805862 Chr.10: 79382477-79382477 on Build GRCh38
rs528820783 Chr.10: 79382258-79382258 on Build GRCh38
rs999290029 Chr.10: 79382556-79382556 on Build GRCh38
rs1018603518 Chr.10: 79382244-79382244 on Build GRCh38
rs531148528 Chr.10: 79382316-79382316 on Build GRCh38
rs932710032 Chr.10: 79382635-79382635 on Build GRCh38
rs1006730840 Chr.10: 79382576-79382576 on Build GRCh38
rs150798026 Chr.10: 79382224-79382224 on Build GRCh38
rs908544797 Chr.10: 79382384-79382384 on Build GRCh38
rs376437767 Chr.10: 79382527-79382527 on Build GRCh38
rs768178528 Chr.10: 79382546-79382546 on Build GRCh38
rs981569858 Chr.10: 79382634-79382634 on Build GRCh38
rs571911703 Chr.10: 79382415-79382415 on Build GRCh38
rs770007398 Chr.10: 79382523-79382523 on Build GRCh38
rs962499157 Chr.10: 79382581-79382581 on Build GRCh38
rs546458243 Chr.10: 79382667-79382667 on Build GRCh38
rs918428810 Chr.10: 79382611-79382611 on Build GRCh38
rs966349312 Chr.10: 79382379-79382379 on Build GRCh38

View Full Product Details