Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 87190750-87191277 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM35A
Entrez Gene ID
54537
Gene Name
family with sequence similarity 35 member A
Gene Aliases
FAM35A1, bA163M19.1
UniGene
Hs.500419
Gene Chromosome Location
Chr.10: 87094478-87191468 on Build GRCh38
Transcripts
NM_019054.2 XM_005269920.3 XM_006717903.3 XM_006717905.3 XM_011539876.2 XM_011539877.2 XM_011539878.2 XM_011539879.2 XM_011539881.2 XM_011539882.2 XM_011539883.2 XM_017016349.1 XM_017016350.1 XM_017016351.1 XM_017016352.1 XM_017016353.1

SNP Detail

SNP ID Chromosome Location
rs200686044 Chr.10: 87191171-87191172 on Build GRCh38
rs991950159 Chr.10: 87191138-87191138 on Build GRCh38
rs954068146 Chr.10: 87191035-87191035 on Build GRCh38
rs557503274 Chr.10: 87191061-87191061 on Build GRCh38
rs1017276195 Chr.10: 87191082-87191082 on Build GRCh38
rs1056149381 Chr.10: 87190845-87190845 on Build GRCh38
rs1044625595 Chr.10: 87191166-87191166 on Build GRCh38
rs371739003 Chr.10: 87191243-87191243 on Build GRCh38
rs535682905 Chr.10: 87191052-87191052 on Build GRCh38
rs938961713 Chr.10: 87190822-87190822 on Build GRCh38
rs8574 Chr.10: 87191252-87191252 on Build GRCh38
rs936147955 Chr.10: 87191173-87191173 on Build GRCh38
rs200967318 Chr.10: 87190797-87190797 on Build GRCh38
rs948949963 Chr.10: 87190855-87190855 on Build GRCh38
rs918306792 Chr.10: 87191133-87191133 on Build GRCh38
rs947741548 Chr.10: 87191162-87191162 on Build GRCh38
rs534695786 Chr.10: 87190981-87190981 on Build GRCh38
rs1000786011 Chr.10: 87190944-87190944 on Build GRCh38
rs11202380 Chr.10: 87191085-87191085 on Build GRCh38
rs1029613383 Chr.10: 87191013-87191013 on Build GRCh38
rs9553 Chr.10: 87191244-87191244 on Build GRCh38
rs985380954 Chr.10: 87191057-87191057 on Build GRCh38
rs563656967 Chr.10: 87190783-87190783 on Build GRCh38
rs531003181 Chr.10: 87190785-87190785 on Build GRCh38
rs1053556 Chr.10: 87191197-87191197 on Build GRCh38
rs3129402 Chr.10: 87190889-87190889 on Build GRCh38
rs539480101 Chr.10: 87191191-87191191 on Build GRCh38
rs972480782 Chr.10: 87190808-87190808 on Build GRCh38
rs1053686357 Chr.10: 87191208-87191208 on Build GRCh38
rs891888194 Chr.10: 87190988-87190988 on Build GRCh38
rs892000969 Chr.10: 87191224-87191224 on Build GRCh38
rs916234359 Chr.10: 87191145-87191145 on Build GRCh38
rs1045069713 Chr.10: 87190877-87190878 on Build GRCh38
rs894847851 Chr.10: 87190849-87190849 on Build GRCh38
rs577203551 Chr.10: 87190818-87190818 on Build GRCh38
rs546931474 Chr.10: 87190996-87190996 on Build GRCh38
rs568401686 Chr.10: 87191000-87191000 on Build GRCh38
rs918351327 Chr.10: 87190811-87190811 on Build GRCh38
rs939076228 Chr.10: 87191136-87191136 on Build GRCh38

View Full Product Details