Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 88284931-88285478 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNLS
Entrez Gene ID
55328
Gene Name
renalase, FAD dependent amine oxidase
Gene Aliases
C10orf59, RENALASE
UniGene
Hs.149849
Gene Chromosome Location
Chr.10: 88176458-88583860 on Build GRCh38
Transcripts
NM_001031709.2 NM_018363.3 XM_005269946.1 XM_005269947.1 XM_005269948.2 XM_005269950.3 XM_011539924.2 XM_017016380.1 XM_017016382.1 XM_017016384.1 XM_017016385.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7143813 c.971T>A p.F324Y Chr.10: 88285412-88285412 on Build GRCh38
1159052 c.966C>T p.S322S Chr.10: 88285417-88285417 on Build GRCh38
3441288 c.1017G>A p.K339K Chr.10: 88285366-88285366 on Build GRCh38
6262731 c.992C>A p.A331D Chr.10: 88285391-88285391 on Build GRCh38
1349648 c.1017G>T p.K339N Chr.10: 88285366-88285366 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs137945207 Chr.10: 88285435-88285435 on Build GRCh38
rs544985084 Chr.10: 88285379-88285379 on Build GRCh38
rs774458389 Chr.10: 88285356-88285356 on Build GRCh38
rs748468060 Chr.10: 88285322-88285322 on Build GRCh38
rs779914625 Chr.10: 88285314-88285314 on Build GRCh38
rs757192697 Chr.10: 88285394-88285394 on Build GRCh38
rs768725882 Chr.10: 88285232-88285234 on Build GRCh38
rs758122407 Chr.10: 88285307-88285307 on Build GRCh38
rs977191957 Chr.10: 88285399-88285399 on Build GRCh38
rs763903879 Chr.10: 88285387-88285387 on Build GRCh38
rs924511148 Chr.10: 88285208-88285208 on Build GRCh38
rs201859794 Chr.10: 88285293-88285295 on Build GRCh38
rs115186306 Chr.10: 88285351-88285351 on Build GRCh38
rs938214502 Chr.10: 88285060-88285060 on Build GRCh38
rs914283310 Chr.10: 88285097-88285097 on Build GRCh38
rs773553907 Chr.10: 88285328-88285328 on Build GRCh38
rs185037365 Chr.10: 88285221-88285221 on Build GRCh38
rs1054310803 Chr.10: 88285235-88285235 on Build GRCh38
rs949712206 Chr.10: 88285154-88285154 on Build GRCh38
rs947126094 Chr.10: 88285103-88285103 on Build GRCh38
rs1015532683 Chr.10: 88285315-88285315 on Build GRCh38
rs765158760 Chr.10: 88285411-88285411 on Build GRCh38
rs1025893657 Chr.10: 88285022-88285022 on Build GRCh38
rs998446769 Chr.10: 88284997-88284997 on Build GRCh38
rs1041374062 Chr.10: 88285106-88285106 on Build GRCh38
rs958498948 Chr.10: 88285092-88285092 on Build GRCh38
rs867845587 Chr.10: 88285451-88285451 on Build GRCh38
rs760393985 Chr.10: 88285386-88285386 on Build GRCh38
rs746515893 Chr.10: 88285249-88285249 on Build GRCh38
rs763385022 Chr.10: 88285370-88285370 on Build GRCh38
rs749939977 Chr.10: 88285420-88285420 on Build GRCh38
rs749655770 Chr.10: 88285446-88285446 on Build GRCh38
rs1034387750 Chr.10: 88285365-88285365 on Build GRCh38
rs544683742 Chr.10: 88285184-88285184 on Build GRCh38
rs910011821 Chr.10: 88285075-88285075 on Build GRCh38
rs756499748 Chr.10: 88285444-88285444 on Build GRCh38
rs1026784689 Chr.10: 88285241-88285241 on Build GRCh38
rs767711360 Chr.10: 88285372-88285372 on Build GRCh38
rs982798244 Chr.10: 88285043-88285043 on Build GRCh38
rs770289581 Chr.10: 88285324-88285324 on Build GRCh38
rs776035124 Chr.10: 88285160-88285160 on Build GRCh38
rs1018308538 Chr.10: 88284961-88284961 on Build GRCh38
rs534234746 Chr.10: 88285098-88285098 on Build GRCh38
rs558387003 Chr.10: 88284966-88284966 on Build GRCh38
rs977345984 Chr.10: 88285205-88285205 on Build GRCh38
rs749825886 Chr.10: 88285335-88285335 on Build GRCh38
rs36077330 Chr.10: 88285066-88285066 on Build GRCh38
rs926800097 Chr.10: 88285056-88285056 on Build GRCh38
rs745414571 Chr.10: 88285305-88285305 on Build GRCh38
rs746693474 Chr.10: 88285437-88285437 on Build GRCh38
rs757999439 Chr.10: 88285434-88285434 on Build GRCh38
rs555779146 Chr.10: 88285123-88285123 on Build GRCh38
rs1048956 Chr.10: 88285032-88285032 on Build GRCh38
rs286484 Chr.10: 88285146-88285146 on Build GRCh38
rs951672677 Chr.10: 88285026-88285026 on Build GRCh38

View Full Product Details