Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 88819815-88820314 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD22
Entrez Gene ID
118932
Gene Name
ankyrin repeat domain 22
Gene Aliases
-
UniGene
Hs.217484
Gene Chromosome Location
Chr.10: 88819899-88851975 on Build GRCh38
Transcripts
NM_144590.2 NM_001128215.1 XM_011539748.2 XM_011539751.2 XM_011539752.2

Gene Symbol
LIPM
Entrez Gene ID
340654
Gene Name
lipase family member M
Gene Aliases
LIPL3, bA304I5.1
UniGene
Hs.55118
Gene Chromosome Location
Chr.10: 88802730-88820857 on Build GRCh38
Transcripts
NM_144590.2 NM_001128215.1 XM_011539748.2 XM_011539751.2 XM_011539752.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5591654 c.908G>A p.R303K Chr.10: 88820257-88820257 on Build GRCh38
54368 c.1008T>C p.T336T Chr.10: 88820237-88820237 on Build GRCh38
5461881 c.1033A>G p.M345V Chr.10: 88820262-88820262 on Build GRCh38
5591653 c.1028G>A p.R343K Chr.10: 88820257-88820257 on Build GRCh38
5656637 c.926C>G p.T309R Chr.10: 88820275-88820275 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200146701 Chr.10: 88820216-88820216 on Build GRCh38
rs949616246 Chr.10: 88820093-88820093 on Build GRCh38
rs145438983 Chr.10: 88820016-88820016 on Build GRCh38
rs185586287 Chr.10: 88820134-88820134 on Build GRCh38
rs536545320 Chr.10: 88819969-88819969 on Build GRCh38
rs1012397473 Chr.10: 88820171-88820171 on Build GRCh38
rs1046959258 Chr.10: 88820118-88820118 on Build GRCh38
rs778782394 Chr.10: 88820120-88820120 on Build GRCh38
rs181144724 Chr.10: 88820077-88820077 on Build GRCh38
rs554155915 Chr.10: 88819882-88819882 on Build GRCh38
rs956711671 Chr.10: 88820221-88820221 on Build GRCh38
rs1003624317 Chr.10: 88820192-88820192 on Build GRCh38
rs963895410 Chr.10: 88820005-88820005 on Build GRCh38
rs935362679 Chr.10: 88820007-88820007 on Build GRCh38
rs749274045 Chr.10: 88820013-88820013 on Build GRCh38
rs61735049 Chr.10: 88820266-88820266 on Build GRCh38
rs1756775 Chr.10: 88819915-88819915 on Build GRCh38
rs867934920 Chr.10: 88820218-88820218 on Build GRCh38
rs1032308516 Chr.10: 88819840-88819840 on Build GRCh38
rs1021438664 Chr.10: 88819880-88819880 on Build GRCh38
rs3910679 Chr.10: 88819852-88819852 on Build GRCh38
rs117206902 Chr.10: 88820197-88820197 on Build GRCh38
rs899431204 Chr.10: 88820163-88820163 on Build GRCh38
rs987037244 Chr.10: 88819916-88819916 on Build GRCh38
rs547915695 Chr.10: 88820111-88820111 on Build GRCh38
rs12255753 Chr.10: 88820255-88820255 on Build GRCh38
rs938212292 Chr.10: 88820142-88820142 on Build GRCh38
rs771111880 Chr.10: 88819979-88819979 on Build GRCh38
rs540802940 Chr.10: 88820182-88820182 on Build GRCh38
rs970856283 Chr.10: 88819950-88819950 on Build GRCh38
rs757917704 Chr.10: 88820184-88820184 on Build GRCh38
rs1016514631 Chr.10: 88820148-88820148 on Build GRCh38
rs922467413 Chr.10: 88820281-88820281 on Build GRCh38
rs75374052 Chr.10: 88819869-88819869 on Build GRCh38
rs1047134161 Chr.10: 88820099-88820099 on Build GRCh38
rs370940208 Chr.10: 88820267-88820267 on Build GRCh38
rs886776340 Chr.10: 88820122-88820122 on Build GRCh38
rs1039523033 Chr.10: 88820062-88820062 on Build GRCh38
rs971187104 Chr.10: 88820186-88820186 on Build GRCh38
rs900893035 Chr.10: 88820078-88820078 on Build GRCh38
rs200206641 Chr.10: 88820236-88820236 on Build GRCh38
rs891081593 Chr.10: 88819859-88819859 on Build GRCh38
rs188143746 Chr.10: 88820158-88820158 on Build GRCh38
rs375499409 Chr.10: 88820084-88820084 on Build GRCh38
rs867895163 Chr.10: 88819848-88819848 on Build GRCh38
rs989353123 Chr.10: 88820012-88820012 on Build GRCh38
rs148834631 Chr.10: 88820046-88820046 on Build GRCh38
rs930964875 Chr.10: 88820069-88820069 on Build GRCh38

View Full Product Details