Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 91629539-91630049 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPP1R3C
Entrez Gene ID
5507
Gene Name
protein phosphatase 1 regulatory subunit 3C
Gene Aliases
PPP1R5
UniGene
Hs.303090
Gene Chromosome Location
Chr.10: 91628440-91633101 on Build GRCh38
Transcripts
NM_005398.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3868045 c.949C>T p.R317* Chr.10: 91629932-91629932 on Build GRCh38
276872 c.927A>C p.R309S Chr.10: 91629954-91629954 on Build GRCh38
921397 c.884G>T p.R295M Chr.10: 91629997-91629997 on Build GRCh38
3665948 c.870C>G p.I290M Chr.10: 91630011-91630011 on Build GRCh38
5084092 c.942C>T p.A314A Chr.10: 91629939-91629939 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs528278914 Chr.10: 91629781-91629781 on Build GRCh38
rs749682115 Chr.10: 91629960-91629968 on Build GRCh38
rs34247182 Chr.10: 91629742-91629743 on Build GRCh38
rs769588898 Chr.10: 91629933-91629933 on Build GRCh38
rs894667848 Chr.10: 91629585-91629585 on Build GRCh38
rs993176920 Chr.10: 91629643-91629643 on Build GRCh38
rs961756095 Chr.10: 91629642-91629642 on Build GRCh38
rs942221313 Chr.10: 91629863-91629863 on Build GRCh38
rs771662320 Chr.10: 91629957-91629957 on Build GRCh38
rs747930364 Chr.10: 91629932-91629932 on Build GRCh38
rs200679873 Chr.10: 91629919-91629919 on Build GRCh38
rs957896866 Chr.10: 91629780-91629780 on Build GRCh38
rs774066029 Chr.10: 91629949-91629949 on Build GRCh38
rs1030348541 Chr.10: 91629836-91629836 on Build GRCh38
rs942232119 Chr.10: 91629859-91629859 on Build GRCh38
rs561723994 Chr.10: 91629832-91629832 on Build GRCh38
rs188131084 Chr.10: 91629869-91629869 on Build GRCh38
rs770990729 Chr.10: 91629947-91629947 on Build GRCh38
rs1013232976 Chr.10: 91629758-91629758 on Build GRCh38
rs762207312 Chr.10: 91629884-91629884 on Build GRCh38
rs908166914 Chr.10: 91629857-91629857 on Build GRCh38
rs376921228 Chr.10: 91629984-91629984 on Build GRCh38
rs759770745 Chr.10: 91629846-91629846 on Build GRCh38
rs530862377 Chr.10: 91629841-91629841 on Build GRCh38
rs147736541 Chr.10: 91629915-91629915 on Build GRCh38
rs777780374 Chr.10: 91630014-91630014 on Build GRCh38
rs560174386 Chr.10: 91629596-91629596 on Build GRCh38
rs746776538 Chr.10: 91629756-91629756 on Build GRCh38
rs754644456 Chr.10: 91629674-91629674 on Build GRCh38
rs546719655 Chr.10: 91629963-91629963 on Build GRCh38
rs977481420 Chr.10: 91629830-91629830 on Build GRCh38
rs767661442 Chr.10: 91630000-91630000 on Build GRCh38
rs898252671 Chr.10: 91629615-91629615 on Build GRCh38
rs1021319376 Chr.10: 91629765-91629765 on Build GRCh38
rs150290304 Chr.10: 91629996-91629996 on Build GRCh38
rs755868278 Chr.10: 91630010-91630010 on Build GRCh38
rs769849532 Chr.10: 91629964-91629964 on Build GRCh38
rs542054154 Chr.10: 91629802-91629802 on Build GRCh38
rs954895956 Chr.10: 91629837-91629837 on Build GRCh38
rs373774243 Chr.10: 91629931-91629931 on Build GRCh38
rs12261209 Chr.10: 91629586-91629586 on Build GRCh38
rs773712201 Chr.10: 91629982-91629982 on Build GRCh38
rs1015991132 Chr.10: 91629634-91629634 on Build GRCh38
rs765695820 Chr.10: 91629887-91629887 on Build GRCh38
rs370552591 Chr.10: 91629990-91629990 on Build GRCh38
rs890152905 Chr.10: 91629948-91629948 on Build GRCh38
rs776822486 Chr.10: 91629814-91629814 on Build GRCh38
rs777872470 Chr.10: 91629934-91629934 on Build GRCh38
rs377761243 Chr.10: 91629893-91629893 on Build GRCh38
rs1030428147 Chr.10: 91629657-91629657 on Build GRCh38
rs758484064 Chr.10: 91629907-91629907 on Build GRCh38
rs966590986 Chr.10: 91629778-91629778 on Build GRCh38
rs36067525 Chr.10: 91629768-91629768 on Build GRCh38
rs769234517 Chr.10: 91630027-91630030 on Build GRCh38
rs867611820 Chr.10: 91629611-91629611 on Build GRCh38
rs745365980 Chr.10: 91629953-91629953 on Build GRCh38
rs927417147 Chr.10: 91629880-91629880 on Build GRCh38
rs34162307 Chr.10: 91629805-91629805 on Build GRCh38
rs777737907 Chr.10: 91629708-91629708 on Build GRCh38
rs989370506 Chr.10: 91629995-91629995 on Build GRCh38
rs914691558 Chr.10: 91629785-91629785 on Build GRCh38
rs138215894 Chr.10: 91630005-91630005 on Build GRCh38
rs765530523 Chr.10: 91629975-91629975 on Build GRCh38
rs749173506 Chr.10: 91629938-91629938 on Build GRCh38
rs928633932 Chr.10: 91629831-91629831 on Build GRCh38
rs1037846945 Chr.10: 91629870-91629870 on Build GRCh38
rs954364872 Chr.10: 91629666-91629666 on Build GRCh38
rs766858533 Chr.10: 91629989-91629989 on Build GRCh38

View Full Product Details