Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 92692502-92693000 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HHEX
Entrez Gene ID
3087
Gene Name
hematopoietically expressed homeobox
Gene Aliases
HEX, HMPH, HOX11L-PEN, PRH, PRHX
UniGene
Hs.118651
Gene Chromosome Location
Chr.10: 92689924-92695651 on Build GRCh38
Transcripts
NM_002729.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5698170 c.508C>A p.L170M Chr.10: 92692514-92692514 on Build GRCh38
325490 c.565C>T p.R189C Chr.10: 92692726-92692726 on Build GRCh38
320810 c.556C>T p.Q186* Chr.10: 92692717-92692717 on Build GRCh38
6130842 c.515A>T p.K172M Chr.10: 92692521-92692521 on Build GRCh38
1745971 c.496G>A p.E166K Chr.10: 92692502-92692502 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774776384 Chr.10: 92692594-92692594 on Build GRCh38
rs377275545 Chr.10: 92692668-92692668 on Build GRCh38
rs903707050 Chr.10: 92692848-92692848 on Build GRCh38
rs780820204 Chr.10: 92692746-92692746 on Build GRCh38
rs576535903 Chr.10: 92692956-92692956 on Build GRCh38
rs182124968 Chr.10: 92692618-92692618 on Build GRCh38
rs745436355 Chr.10: 92692556-92692556 on Build GRCh38
rs979370507 Chr.10: 92692538-92692538 on Build GRCh38
rs980960439 Chr.10: 92692622-92692622 on Build GRCh38
rs779766063 Chr.10: 92692557-92692557 on Build GRCh38
rs753656580 Chr.10: 92692539-92692539 on Build GRCh38
rs532429246 Chr.10: 92692836-92692836 on Build GRCh38
rs1030893427 Chr.10: 92692905-92692905 on Build GRCh38
rs2275729 Chr.10: 92692673-92692673 on Build GRCh38
rs778979179 Chr.10: 92692554-92692554 on Build GRCh38
rs191684511 Chr.10: 92692915-92692915 on Build GRCh38
rs755960214 Chr.10: 92692524-92692524 on Build GRCh38
rs746727301 Chr.10: 92692680-92692683 on Build GRCh38
rs757112503 Chr.10: 92692666-92692666 on Build GRCh38
rs563471029 Chr.10: 92692679-92692679 on Build GRCh38
rs370419063 Chr.10: 92692553-92692553 on Build GRCh38
rs1013554077 Chr.10: 92692916-92692916 on Build GRCh38
rs1011232541 Chr.10: 92692633-92692633 on Build GRCh38
rs996365384 Chr.10: 92692827-92692827 on Build GRCh38
rs984775651 Chr.10: 92692831-92692831 on Build GRCh38
rs373773105 Chr.10: 92692761-92692761 on Build GRCh38
rs767447009 Chr.10: 92692595-92692595 on Build GRCh38
rs917973891 Chr.10: 92692616-92692616 on Build GRCh38
rs750323865 Chr.10: 92692671-92692671 on Build GRCh38
rs770221901 Chr.10: 92692571-92692571 on Build GRCh38
rs199770552 Chr.10: 92692760-92692760 on Build GRCh38
rs71480833 Chr.10: 92692576-92692576 on Build GRCh38
rs746590738 Chr.10: 92692562-92692562 on Build GRCh38
rs773604024 Chr.10: 92692583-92692583 on Build GRCh38
rs764391242 Chr.10: 92692657-92692657 on Build GRCh38
rs768240433 Chr.10: 92692693-92692693 on Build GRCh38
rs2275730 Chr.10: 92692585-92692585 on Build GRCh38
rs574754629 Chr.10: 92692637-92692637 on Build GRCh38
rs748729216 Chr.10: 92692795-92692796 on Build GRCh38
rs775484791 Chr.10: 92692776-92692776 on Build GRCh38
rs771228365 Chr.10: 92692755-92692755 on Build GRCh38
rs938886206 Chr.10: 92692603-92692603 on Build GRCh38
rs753742389 Chr.10: 92692665-92692665 on Build GRCh38
rs752995067 Chr.10: 92692596-92692596 on Build GRCh38
rs779389753 Chr.10: 92692675-92692675 on Build GRCh38
rs938828817 Chr.10: 92692954-92692954 on Build GRCh38
rs555953607 Chr.10: 92692628-92692628 on Build GRCh38
rs777407704 Chr.10: 92692657-92692657 on Build GRCh38
rs749658938 Chr.10: 92692752-92692752 on Build GRCh38
rs760793981 Chr.10: 92692791-92692791 on Build GRCh38
rs892012563 Chr.10: 92692958-92692958 on Build GRCh38
rs140982034 Chr.10: 92692879-92692879 on Build GRCh38
rs376915003 Chr.10: 92692691-92692691 on Build GRCh38
rs370777153 Chr.10: 92692670-92692670 on Build GRCh38
rs909209740 Chr.10: 92692833-92692834 on Build GRCh38
rs748263982 Chr.10: 92692813-92692813 on Build GRCh38
rs771498064 Chr.10: 92692588-92692588 on Build GRCh38
rs777393392 Chr.10: 92692531-92692531 on Build GRCh38
rs528032641 Chr.10: 92692774-92692774 on Build GRCh38
rs972567173 Chr.10: 92692773-92692773 on Build GRCh38
rs186807208 Chr.10: 92692804-92692804 on Build GRCh38
rs928728149 Chr.10: 92692946-92692946 on Build GRCh38

View Full Product Details