Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 95067117-95067616 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP2C8
Entrez Gene ID
1558
Gene Name
cytochrome P450 family 2 subfamily C member 8
Gene Aliases
CPC8, CYPIIC8, MP-12/MP-20
UniGene
Hs.709188
Gene Chromosome Location
Chr.10: 95036772-95069512 on Build GRCh38
Transcripts
NM_000770.3 NM_001198853.1 NM_001198854.1 NM_001198855.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3442096 c.92G>A p.G31E Chr.10: 95067339-95067339 on Build GRCh38
5608212 c.120C>T p.F40F Chr.10: 95067311-95067311 on Build GRCh38
921757 c.314G>T p.R105I Chr.10: 95067546-95067546 on Build GRCh38
5608202 c.354G>A p.K118K Chr.10: 95067335-95067335 on Build GRCh38
5608207 c.442G>A p.E148K Chr.10: 95067247-95067247 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767877277 Chr.10: 95067433-95067433 on Build GRCh38
rs757341557 Chr.10: 95067461-95067461 on Build GRCh38
rs369552457 Chr.10: 95067344-95067344 on Build GRCh38
rs767098538 Chr.10: 95067229-95067229 on Build GRCh38
rs750338178 Chr.10: 95067539-95067539 on Build GRCh38
rs201739495 Chr.10: 95067321-95067321 on Build GRCh38
rs748104826 Chr.10: 95067389-95067389 on Build GRCh38
rs1050608014 Chr.10: 95067367-95067368 on Build GRCh38
rs772754152 Chr.10: 95067207-95067207 on Build GRCh38
rs937139929 Chr.10: 95067191-95067191 on Build GRCh38
rs370852349 Chr.10: 95067495-95067495 on Build GRCh38
rs780488900 Chr.10: 95067265-95067265 on Build GRCh38
rs11572076 Chr.10: 95067393-95067393 on Build GRCh38
rs369591911 Chr.10: 95067318-95067318 on Build GRCh38
rs11572079 Chr.10: 95067361-95067361 on Build GRCh38
rs768330125 Chr.10: 95067485-95067485 on Build GRCh38
rs780189252 Chr.10: 95067556-95067556 on Build GRCh38
rs780212136 Chr.10: 95067361-95067362 on Build GRCh38
rs11572082 Chr.10: 95067165-95067165 on Build GRCh38
rs11572078 Chr.10: 95067362-95067363 on Build GRCh38
rs759288662 Chr.10: 95067228-95067228 on Build GRCh38
rs75923676 Chr.10: 95067370-95067370 on Build GRCh38
rs751691701 Chr.10: 95067268-95067268 on Build GRCh38
rs77036916 Chr.10: 95067150-95067150 on Build GRCh38
rs751851689 Chr.10: 95067488-95067488 on Build GRCh38
rs767832069 Chr.10: 95067502-95067502 on Build GRCh38
rs398088095 Chr.10: 95067216-95067216 on Build GRCh38
rs757873008 Chr.10: 95067163-95067163 on Build GRCh38
rs746840813 Chr.10: 95067484-95067484 on Build GRCh38
rs758856245 Chr.10: 95067170-95067170 on Build GRCh38
rs776175402 Chr.10: 95067410-95067410 on Build GRCh38
rs746198469 Chr.10: 95067168-95067168 on Build GRCh38
rs201561213 Chr.10: 95067217-95067217 on Build GRCh38
rs771935485 Chr.10: 95067458-95067458 on Build GRCh38
rs755391351 Chr.10: 95067345-95067345 on Build GRCh38
rs188305680 Chr.10: 95067536-95067536 on Build GRCh38
rs529379413 Chr.10: 95067296-95067296 on Build GRCh38
rs367887998 Chr.10: 95067186-95067186 on Build GRCh38
rs778907051 Chr.10: 95067464-95067464 on Build GRCh38
rs754351169 Chr.10: 95067160-95067160 on Build GRCh38
rs781440953 Chr.10: 95067283-95067283 on Build GRCh38
rs745807384 Chr.10: 95067481-95067481 on Build GRCh38
rs770873636 Chr.10: 95067363-95067363 on Build GRCh38
rs11572081 Chr.10: 95067209-95067209 on Build GRCh38
rs397840174 Chr.10: 95067369-95067370 on Build GRCh38
rs11572075 Chr.10: 95067422-95067422 on Build GRCh38
rs200057634 Chr.10: 95067240-95067240 on Build GRCh38
rs371547491 Chr.10: 95067212-95067212 on Build GRCh38
rs376162394 Chr.10: 95067489-95067489 on Build GRCh38
rs757297073 Chr.10: 95067524-95067524 on Build GRCh38
rs781385756 Chr.10: 95067347-95067347 on Build GRCh38
rs578254206 Chr.10: 95067567-95067567 on Build GRCh38
rs398102371 Chr.10: 95067515-95067515 on Build GRCh38
rs769926169 Chr.10: 95067293-95067293 on Build GRCh38
rs760313151 Chr.10: 95067248-95067248 on Build GRCh38
rs758378725 Chr.10: 95067546-95067546 on Build GRCh38
rs775805017 Chr.10: 95067514-95067514 on Build GRCh38
rs771830853 Chr.10: 95067314-95067314 on Build GRCh38
rs61757320 Chr.10: 95067487-95067487 on Build GRCh38
rs145992929 Chr.10: 95067295-95067295 on Build GRCh38
rs767728082 Chr.10: 95067164-95067170 on Build GRCh38
rs756125116 Chr.10: 95067353-95067353 on Build GRCh38
rs371675563 Chr.10: 95067188-95067188 on Build GRCh38
rs775342549 Chr.10: 95067315-95067315 on Build GRCh38
rs746804534 Chr.10: 95067557-95067557 on Build GRCh38
rs72558196 Chr.10: 95067214-95067214 on Build GRCh38
rs745646379 Chr.10: 95067371-95067371 on Build GRCh38
rs376330415 Chr.10: 95067388-95067388 on Build GRCh38
rs757434823 Chr.10: 95067473-95067473 on Build GRCh38
rs763545622 Chr.10: 95067342-95067342 on Build GRCh38
rs183769701 Chr.10: 95067494-95067494 on Build GRCh38
rs188111115 Chr.10: 95067316-95067316 on Build GRCh38
rs752864377 Chr.10: 95067448-95067448 on Build GRCh38
rs2275622 Chr.10: 95067421-95067421 on Build GRCh38
rs879129614 Chr.10: 95067421-95067422 on Build GRCh38
rs752917521 Chr.10: 95067348-95067348 on Build GRCh38
rs1011421475 Chr.10: 95067336-95067336 on Build GRCh38
rs918830502 Chr.10: 95067457-95067457 on Build GRCh38
rs758210087 Chr.10: 95067482-95067482 on Build GRCh38
rs756393909 Chr.10: 95067450-95067450 on Build GRCh38
rs751649419 Chr.10: 95067458-95067458 on Build GRCh38
rs912588650 Chr.10: 95067518-95067518 on Build GRCh38
rs139650638 Chr.10: 95067300-95067300 on Build GRCh38
rs754076612 Chr.10: 95067523-95067523 on Build GRCh38
rs368299637 Chr.10: 95067203-95067203 on Build GRCh38
rs748264751 Chr.10: 95067196-95067196 on Build GRCh38
rs750846477 Chr.10: 95067338-95067338 on Build GRCh38
rs540288649 Chr.10: 95067259-95067259 on Build GRCh38
rs749143149 Chr.10: 95067362-95067362 on Build GRCh38
rs772709446 Chr.10: 95067528-95067528 on Build GRCh38
rs773255712 Chr.10: 95067204-95067204 on Build GRCh38
rs199931273 Chr.10: 95067527-95067527 on Build GRCh38
rs962133452 Chr.10: 95067207-95067207 on Build GRCh38
rs567203057 Chr.10: 95067432-95067432 on Build GRCh38
rs777501435 Chr.10: 95067294-95067294 on Build GRCh38
rs747766000 Chr.10: 95067584-95067584 on Build GRCh38
rs1012107644 Chr.10: 95067560-95067560 on Build GRCh38
rs780827802 Chr.10: 95067581-95067581 on Build GRCh38
rs1053507227 Chr.10: 95067358-95067358 on Build GRCh38
rs772061547 Chr.10: 95067374-95067374 on Build GRCh38
rs774778262 Chr.10: 95067492-95067492 on Build GRCh38
rs763144615 Chr.10: 95067417-95067417 on Build GRCh38
rs143096432 Chr.10: 95067369-95067369 on Build GRCh38
rs201770172 Chr.10: 95067491-95067491 on Build GRCh38
rs370055521 Chr.10: 95067159-95067159 on Build GRCh38
rs747268921 Chr.10: 95067187-95067187 on Build GRCh38
rs377386087 Chr.10: 95067319-95067319 on Build GRCh38
rs923633609 Chr.10: 95067299-95067299 on Build GRCh38
rs756367430 Chr.10: 95067360-95067361 on Build GRCh38
rs765827545 Chr.10: 95067258-95067258 on Build GRCh38
rs375369698 Chr.10: 95067189-95067189 on Build GRCh38
rs775217153 Chr.10: 95067386-95067386 on Build GRCh38

View Full Product Details