Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 95068277-95068782 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP2C8
Entrez Gene ID
1558
Gene Name
cytochrome P450 family 2 subfamily C member 8
Gene Aliases
CPC8, CYPIIC8, MP-12/MP-20
UniGene
Hs.709188
Gene Chromosome Location
Chr.10: 95036772-95069512 on Build GRCh38
Transcripts
NM_000770.3 NM_001198853.1 NM_001198854.1 NM_001198855.1

SNP Detail

SNP ID Chromosome Location
rs888746628 Chr.10: 95068706-95068706 on Build GRCh38
rs1934956 Chr.10: 95068403-95068403 on Build GRCh38
rs900022766 Chr.10: 95068740-95068740 on Build GRCh38
rs191121348 Chr.10: 95068321-95068321 on Build GRCh38
rs574229356 Chr.10: 95068382-95068382 on Build GRCh38
rs1047892441 Chr.10: 95068664-95068664 on Build GRCh38
rs915877667 Chr.10: 95068527-95068527 on Build GRCh38
rs753797273 Chr.10: 95068552-95068552 on Build GRCh38
rs1021549583 Chr.10: 95068484-95068484 on Build GRCh38
rs924716436 Chr.10: 95068502-95068502 on Build GRCh38
rs892333880 Chr.10: 95068655-95068655 on Build GRCh38
rs948737715 Chr.10: 95068534-95068534 on Build GRCh38
rs964243762 Chr.10: 95068442-95068442 on Build GRCh38
rs11572070 Chr.10: 95068387-95068387 on Build GRCh38
rs966207415 Chr.10: 95068384-95068384 on Build GRCh38
rs545050969 Chr.10: 95068509-95068509 on Build GRCh38
rs777401577 Chr.10: 95068581-95068581 on Build GRCh38
rs2071426 Chr.10: 95068566-95068566 on Build GRCh38
rs960433126 Chr.10: 95068519-95068519 on Build GRCh38
rs920225063 Chr.10: 95068638-95068638 on Build GRCh38
rs975654228 Chr.10: 95068458-95068458 on Build GRCh38
rs898851064 Chr.10: 95068331-95068331 on Build GRCh38
rs1019776631 Chr.10: 95068441-95068441 on Build GRCh38
rs1007648930 Chr.10: 95068737-95068737 on Build GRCh38
rs11572068 Chr.10: 95068524-95068524 on Build GRCh38
rs565032653 Chr.10: 95068558-95068558 on Build GRCh38
rs190031904 Chr.10: 95068642-95068642 on Build GRCh38
rs993626081 Chr.10: 95068352-95068352 on Build GRCh38
rs997018487 Chr.10: 95068741-95068741 on Build GRCh38
rs527519359 Chr.10: 95068559-95068559 on Build GRCh38
rs928731141 Chr.10: 95068577-95068577 on Build GRCh38
rs1037636118 Chr.10: 95068597-95068597 on Build GRCh38
rs928992722 Chr.10: 95068644-95068644 on Build GRCh38
rs890204619 Chr.10: 95068676-95068676 on Build GRCh38
rs10882526 Chr.10: 95068744-95068744 on Build GRCh38
rs752397723 Chr.10: 95068649-95068649 on Build GRCh38
rs756701385 Chr.10: 95068669-95068669 on Build GRCh38
rs934786692 Chr.10: 95068578-95068578 on Build GRCh38
rs547262589 Chr.10: 95068560-95068560 on Build GRCh38
rs1007269242 Chr.10: 95068364-95068364 on Build GRCh38
rs562851208 Chr.10: 95068325-95068325 on Build GRCh38
rs758239513 Chr.10: 95068711-95068711 on Build GRCh38
rs768492798 Chr.10: 95068605-95068605 on Build GRCh38
rs751843417 Chr.10: 95068320-95068320 on Build GRCh38
rs968753547 Chr.10: 95068487-95068487 on Build GRCh38
rs977869793 Chr.10: 95068494-95068494 on Build GRCh38
rs753496656 Chr.10: 95068665-95068665 on Build GRCh38
rs948596185 Chr.10: 95068571-95068571 on Build GRCh38
rs1041170592 Chr.10: 95068695-95068695 on Build GRCh38
rs184310913 Chr.10: 95068423-95068423 on Build GRCh38
rs371907206 Chr.10: 95068591-95068591 on Build GRCh38
rs529921040 Chr.10: 95068726-95068726 on Build GRCh38
rs1009093583 Chr.10: 95068681-95068681 on Build GRCh38
rs562507704 Chr.10: 95068400-95068400 on Build GRCh38

View Full Product Details