Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 101782157-101782651 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NPM3
Entrez Gene ID
10360
Gene Name
nucleophosmin/nucleoplasmin 3
Gene Aliases
PORMIN, TMEM123
UniGene
Hs.90691
Gene Chromosome Location
Chr.10: 101781325-101783413 on Build GRCh38
Transcripts
NM_006993.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6498254 c.207C>G p.L69L Chr.10: 101782595-101782595 on Build GRCh38
4998370 c.329G>T p.S110I Chr.10: 101782347-101782347 on Build GRCh38
426960 c.318A>G p.Q106Q Chr.10: 101782484-101782484 on Build GRCh38
2154163 c.310T>G p.S104A Chr.10: 101782492-101782492 on Build GRCh38
3367858 c.368G>T p.R123L Chr.10: 101782308-101782308 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778641797 Chr.10: 101782293-101782293 on Build GRCh38
rs140744459 Chr.10: 101782322-101782322 on Build GRCh38
rs373843260 Chr.10: 101782381-101782381 on Build GRCh38
rs969760962 Chr.10: 101782189-101782189 on Build GRCh38
rs781694665 Chr.10: 101782274-101782274 on Build GRCh38
rs888879096 Chr.10: 101782442-101782442 on Build GRCh38
rs142969681 Chr.10: 101782330-101782330 on Build GRCh38
rs769860842 Chr.10: 101782445-101782445 on Build GRCh38
rs747110225 Chr.10: 101782611-101782612 on Build GRCh38
rs749371700 Chr.10: 101782544-101782544 on Build GRCh38
rs778483639 Chr.10: 101782375-101782375 on Build GRCh38
rs3816354 Chr.10: 101782191-101782191 on Build GRCh38
rs754479783 Chr.10: 101782358-101782358 on Build GRCh38
rs777854702 Chr.10: 101782610-101782611 on Build GRCh38
rs992856666 Chr.10: 101782583-101782583 on Build GRCh38
rs781082503 Chr.10: 101782186-101782186 on Build GRCh38
rs779190979 Chr.10: 101782456-101782457 on Build GRCh38
rs763699677 Chr.10: 101782314-101782315 on Build GRCh38
rs759778942 Chr.10: 101782239-101782239 on Build GRCh38
rs768147586 Chr.10: 101782490-101782490 on Build GRCh38
rs769763325 Chr.10: 101782353-101782353 on Build GRCh38
rs2592835 Chr.10: 101782555-101782555 on Build GRCh38
rs1051927073 Chr.10: 101782439-101782439 on Build GRCh38
rs963160328 Chr.10: 101782333-101782333 on Build GRCh38
rs752387523 Chr.10: 101782376-101782376 on Build GRCh38
rs757032442 Chr.10: 101782562-101782562 on Build GRCh38
rs752340132 Chr.10: 101782532-101782532 on Build GRCh38
rs769914016 Chr.10: 101782586-101782586 on Build GRCh38
rs1035587510 Chr.10: 101782197-101782197 on Build GRCh38
rs2735420 Chr.10: 101782563-101782563 on Build GRCh38
rs555727936 Chr.10: 101782346-101782346 on Build GRCh38
rs770809218 Chr.10: 101782316-101782316 on Build GRCh38
rs774807313 Chr.10: 101782473-101782473 on Build GRCh38
rs537805213 Chr.10: 101782240-101782240 on Build GRCh38
rs746931269 Chr.10: 101782313-101782313 on Build GRCh38
rs368953964 Chr.10: 101782607-101782607 on Build GRCh38
rs745777576 Chr.10: 101782585-101782585 on Build GRCh38
rs771883465 Chr.10: 101782299-101782299 on Build GRCh38
rs953484901 Chr.10: 101782182-101782182 on Build GRCh38
rs775768455 Chr.10: 101782339-101782339 on Build GRCh38
rs369304032 Chr.10: 101782543-101782543 on Build GRCh38
rs747904834 Chr.10: 101782298-101782298 on Build GRCh38
rs748757895 Chr.10: 101782276-101782276 on Build GRCh38
rs377365267 Chr.10: 101782254-101782254 on Build GRCh38
rs776726192 Chr.10: 101782325-101782325 on Build GRCh38
rs764567878 Chr.10: 101782250-101782250 on Build GRCh38
rs781176675 Chr.10: 101782572-101782572 on Build GRCh38
rs915983073 Chr.10: 101782626-101782628 on Build GRCh38
rs561586403 Chr.10: 101782433-101782434 on Build GRCh38
rs151099007 Chr.10: 101782512-101782512 on Build GRCh38
rs755620891 Chr.10: 101782265-101782265 on Build GRCh38
rs529646776 Chr.10: 101782595-101782595 on Build GRCh38
rs768714568 Chr.10: 101782469-101782469 on Build GRCh38
rs748660840 Chr.10: 101782490-101782490 on Build GRCh38
rs950616064 Chr.10: 101782198-101782198 on Build GRCh38
rs755357002 Chr.10: 101782434-101782442 on Build GRCh38
rs775615848 Chr.10: 101782448-101782448 on Build GRCh38
rs371217356 Chr.10: 101782458-101782458 on Build GRCh38
rs374169540 Chr.10: 101782552-101782552 on Build GRCh38
rs762922272 Chr.10: 101782244-101782244 on Build GRCh38
rs758865059 Chr.10: 101782557-101782559 on Build GRCh38
rs1011324150 Chr.10: 101782183-101782183 on Build GRCh38
rs762198460 Chr.10: 101782482-101782482 on Build GRCh38
rs376839189 Chr.10: 101782223-101782223 on Build GRCh38
rs772560838 Chr.10: 101782615-101782615 on Build GRCh38
rs746744168 Chr.10: 101782394-101782394 on Build GRCh38
rs751369523 Chr.10: 101782553-101782553 on Build GRCh38
rs766439322 Chr.10: 101782214-101782214 on Build GRCh38
rs968954382 Chr.10: 101782451-101782451 on Build GRCh38
rs527921483 Chr.10: 101782521-101782521 on Build GRCh38
rs370768734 Chr.10: 101782342-101782342 on Build GRCh38
rs753012521 Chr.10: 101782242-101782242 on Build GRCh38
rs746048754 Chr.10: 101782281-101782281 on Build GRCh38
rs775197678 Chr.10: 101782520-101782520 on Build GRCh38
rs746009576 Chr.10: 101782398-101782398 on Build GRCh38
rs758735619 Chr.10: 101782246-101782246 on Build GRCh38
rs924642918 Chr.10: 101782230-101782230 on Build GRCh38
rs565668791 Chr.10: 101782602-101782602 on Build GRCh38
rs562305258 Chr.10: 101782345-101782345 on Build GRCh38
rs773087326 Chr.10: 101782310-101782310 on Build GRCh38
rs750952873 Chr.10: 101782507-101782507 on Build GRCh38
rs770162984 Chr.10: 101782337-101782337 on Build GRCh38
rs893306220 Chr.10: 101782565-101782565 on Build GRCh38
rs376410763 Chr.10: 101782386-101782386 on Build GRCh38
rs770862525 Chr.10: 101782396-101782396 on Build GRCh38
rs751072311 Chr.10: 101782434-101782436 on Build GRCh38
rs748576371 Chr.10: 101782612-101782612 on Build GRCh38
rs759549777 Chr.10: 101782329-101782329 on Build GRCh38
rs867616865 Chr.10: 101782601-101782601 on Build GRCh38

View Full Product Details