Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 102593845-102594343 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SUFU
Entrez Gene ID
51684
Gene Name
SUFU negative regulator of hedgehog signaling
Gene Aliases
PRO1280, SUFUH, SUFUXL
UniGene
Hs.404089
Gene Chromosome Location
Chr.10: 102502879-102633458 on Build GRCh38
Transcripts
NM_001178133.1 NM_016169.3 XM_011539858.2 XM_011539860.2 XM_011539861.2 XM_011539863.2 XM_011539864.2 XM_017016323.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6906248 c.703A>G p.I235V Chr.10: 102594012-102594012 on Build GRCh38
6965303 c.752T>G p.L251R Chr.10: 102594061-102594061 on Build GRCh38
6476904 c.756+1G>A p.? Chr.10: 102594066-102594066 on Build GRCh38
3966851 c.756+1G>T p.? Chr.10: 102594066-102594066 on Build GRCh38
2059395 c.741C>T p.I247I Chr.10: 102594050-102594050 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs928481146 Chr.10: 102593925-102593925 on Build GRCh38
rs776568361 Chr.10: 102593977-102593977 on Build GRCh38
rs969499162 Chr.10: 102594177-102594177 on Build GRCh38
rs544734275 Chr.10: 102594037-102594037 on Build GRCh38
rs750808785 Chr.10: 102594058-102594058 on Build GRCh38
rs998500184 Chr.10: 102593941-102593941 on Build GRCh38
rs928031252 Chr.10: 102594222-102594222 on Build GRCh38
rs540910508 Chr.10: 102593881-102593881 on Build GRCh38
rs78247976 Chr.10: 102594310-102594310 on Build GRCh38
rs908497372 Chr.10: 102594280-102594280 on Build GRCh38
rs374233754 Chr.10: 102593934-102593934 on Build GRCh38
rs187554337 Chr.10: 102594109-102594109 on Build GRCh38
rs746032665 Chr.10: 102593984-102593984 on Build GRCh38
rs773024733 Chr.10: 102593967-102593967 on Build GRCh38
rs990270248 Chr.10: 102593878-102593878 on Build GRCh38
rs769059712 Chr.10: 102594292-102594292 on Build GRCh38
rs775675385 Chr.10: 102593999-102593999 on Build GRCh38
rs377663654 Chr.10: 102593974-102593974 on Build GRCh38
rs981221751 Chr.10: 102594269-102594269 on Build GRCh38
rs941237962 Chr.10: 102594297-102594297 on Build GRCh38
rs967104848 Chr.10: 102594260-102594260 on Build GRCh38
rs559017159 Chr.10: 102593892-102593892 on Build GRCh38
rs981282262 Chr.10: 102594199-102594199 on Build GRCh38
rs956950883 Chr.10: 102594096-102594096 on Build GRCh38
rs200885602 Chr.10: 102594002-102594002 on Build GRCh38
rs139877584 Chr.10: 102594204-102594204 on Build GRCh38
rs1004947552 Chr.10: 102594145-102594145 on Build GRCh38
rs1022407537 Chr.10: 102594135-102594135 on Build GRCh38
rs1019058906 Chr.10: 102594147-102594147 on Build GRCh38
rs372426793 Chr.10: 102593869-102593869 on Build GRCh38
rs369465986 Chr.10: 102594024-102594024 on Build GRCh38
rs966218051 Chr.10: 102594180-102594180 on Build GRCh38
rs144666812 Chr.10: 102594012-102594012 on Build GRCh38
rs1038236397 Chr.10: 102594304-102594304 on Build GRCh38
rs937160454 Chr.10: 102594302-102594302 on Build GRCh38
rs895100144 Chr.10: 102593963-102593963 on Build GRCh38
rs996384285 Chr.10: 102594201-102594201 on Build GRCh38
rs374600088 Chr.10: 102594082-102594082 on Build GRCh38
rs754583077 Chr.10: 102594060-102594060 on Build GRCh38
rs982794314 Chr.10: 102594265-102594265 on Build GRCh38
rs762294261 Chr.10: 102594036-102594036 on Build GRCh38
rs1045571566 Chr.10: 102593923-102593923 on Build GRCh38
rs937233975 Chr.10: 102593942-102593942 on Build GRCh38
rs370938714 Chr.10: 102593960-102593960 on Build GRCh38
rs770841417 Chr.10: 102593972-102593972 on Build GRCh38
rs928449905 Chr.10: 102594294-102594294 on Build GRCh38
rs960997936 Chr.10: 102594229-102594229 on Build GRCh38
rs770167005 Chr.10: 102593987-102593987 on Build GRCh38
rs1029252844 Chr.10: 102594219-102594219 on Build GRCh38
rs890791327 Chr.10: 102593888-102593888 on Build GRCh38
rs945845242 Chr.10: 102593896-102593896 on Build GRCh38
rs749799131 Chr.10: 102594015-102594015 on Build GRCh38
rs541647541 Chr.10: 102594221-102594221 on Build GRCh38
rs764838079 Chr.10: 102594075-102594075 on Build GRCh38

View Full Product Details