Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 116676321-116676820 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSPA12A
Entrez Gene ID
259217
Gene Name
heat shock protein family A (Hsp70) member 12A
Gene Aliases
-
UniGene
Hs.648448
Gene Chromosome Location
Chr.10: 116671192-116851241 on Build GRCh38
Transcripts
NM_025015.2 XM_005269672.3 XM_005269673.4 XM_011539579.2 XM_011539580.2 XM_017016032.1

SNP Detail

SNP ID Chromosome Location
rs1004014290 Chr.10: 116676747-116676747 on Build GRCh38
rs782663497 Chr.10: 116676460-116676460 on Build GRCh38
rs575016908 Chr.10: 116676440-116676440 on Build GRCh38
rs782446909 Chr.10: 116676458-116676458 on Build GRCh38
rs782389106 Chr.10: 116676543-116676543 on Build GRCh38
rs1040140268 Chr.10: 116676575-116676575 on Build GRCh38
rs373264624 Chr.10: 116676501-116676501 on Build GRCh38
rs782193625 Chr.10: 116676411-116676411 on Build GRCh38
rs782368186 Chr.10: 116676465-116676465 on Build GRCh38
rs375658090 Chr.10: 116676506-116676506 on Build GRCh38
rs781884715 Chr.10: 116676549-116676549 on Build GRCh38
rs201730028 Chr.10: 116676423-116676423 on Build GRCh38
rs371358207 Chr.10: 116676464-116676464 on Build GRCh38
rs782096899 Chr.10: 116676422-116676422 on Build GRCh38
rs931819365 Chr.10: 116676585-116676585 on Build GRCh38
rs199760082 Chr.10: 116676420-116676420 on Build GRCh38
rs782216299 Chr.10: 116676394-116676394 on Build GRCh38
rs2121540 Chr.10: 116676528-116676528 on Build GRCh38
rs782727422 Chr.10: 116676521-116676521 on Build GRCh38
rs990982815 Chr.10: 116676783-116676783 on Build GRCh38
rs920835562 Chr.10: 116676548-116676548 on Build GRCh38
rs1027827141 Chr.10: 116676795-116676795 on Build GRCh38
rs2166634 Chr.10: 116676557-116676557 on Build GRCh38
rs782424256 Chr.10: 116676510-116676510 on Build GRCh38
rs1040577433 Chr.10: 116676362-116676362 on Build GRCh38
rs532284286 Chr.10: 116676454-116676454 on Build GRCh38
rs781844228 Chr.10: 116676538-116676538 on Build GRCh38
rs782222751 Chr.10: 116676568-116676568 on Build GRCh38
rs782777572 Chr.10: 116676428-116676428 on Build GRCh38
rs563208781 Chr.10: 116676425-116676425 on Build GRCh38
rs376394619 Chr.10: 116676442-116676442 on Build GRCh38
rs782401482 Chr.10: 116676413-116676413 on Build GRCh38
rs531091209 Chr.10: 116676751-116676751 on Build GRCh38
rs572101698 Chr.10: 116676371-116676371 on Build GRCh38
rs959750241 Chr.10: 116676767-116676767 on Build GRCh38
rs781991730 Chr.10: 116676514-116676514 on Build GRCh38
rs782665057 Chr.10: 116676392-116676392 on Build GRCh38
rs1010113598 Chr.10: 116676703-116676703 on Build GRCh38
rs76765419 Chr.10: 116676664-116676664 on Build GRCh38
rs540809005 Chr.10: 116676379-116676379 on Build GRCh38
rs368362365 Chr.10: 116676387-116676387 on Build GRCh38
rs902525895 Chr.10: 116676731-116676731 on Build GRCh38
rs930920417 Chr.10: 116676550-116676550 on Build GRCh38
rs959585602 Chr.10: 116676541-116676541 on Build GRCh38
rs782054387 Chr.10: 116676542-116676542 on Build GRCh38
rs528103638 Chr.10: 116676707-116676707 on Build GRCh38
rs782497457 Chr.10: 116676391-116676391 on Build GRCh38
rs900271159 Chr.10: 116676580-116676580 on Build GRCh38
rs782136682 Chr.10: 116676518-116676518 on Build GRCh38
rs1054249815 Chr.10: 116676605-116676605 on Build GRCh38
rs782136142 Chr.10: 116676369-116676369 on Build GRCh38
rs782107484 Chr.10: 116676418-116676418 on Build GRCh38
rs782151152 Chr.10: 116676508-116676508 on Build GRCh38
rs781955087 Chr.10: 116676421-116676421 on Build GRCh38
rs782734592 Chr.10: 116676546-116676546 on Build GRCh38
rs372317686 Chr.10: 116676396-116676396 on Build GRCh38
rs782529850 Chr.10: 116676372-116676372 on Build GRCh38
rs1020880243 Chr.10: 116676722-116676722 on Build GRCh38
rs782024633 Chr.10: 116676416-116676416 on Build GRCh38
rs371999275 Chr.10: 116676360-116676360 on Build GRCh38
rs781936173 Chr.10: 116676368-116676368 on Build GRCh38
rs181977065 Chr.10: 116676424-116676424 on Build GRCh38
rs782487879 Chr.10: 116676441-116676441 on Build GRCh38
rs184577824 Chr.10: 116676472-116676472 on Build GRCh38
rs781927735 Chr.10: 116676399-116676399 on Build GRCh38
rs781824167 Chr.10: 116676407-116676407 on Build GRCh38
rs782557265 Chr.10: 116676427-116676427 on Build GRCh38
rs188989524 Chr.10: 116676735-116676735 on Build GRCh38

View Full Product Details