Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 418796-419295 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANO9
Entrez Gene ID
338440
Gene Name
anoctamin 9
Gene Aliases
PIG5, TMEM16J, TP53I5
UniGene
Hs.501622
Gene Chromosome Location
Chr.11: 417930-442011 on Build GRCh38
Transcripts
NM_001012302.2 XM_011520053.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7163485 c.2008G>C p.E670Q Chr.11: 418916-418916 on Build GRCh38
926866 c.1969G>A p.V657I Chr.11: 418955-418955 on Build GRCh38
6601208 c.2051G>A p.R684H Chr.11: 418799-418799 on Build GRCh38
4662088 c.2029_2030delCT p.L677fs*33 Chr.11: 418894-418895 on Build GRCh38
4596761 c.2036G>T p.R679I Chr.11: 418888-418888 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs576751915 Chr.11: 418843-418843 on Build GRCh38
rs200394752 Chr.11: 418956-418956 on Build GRCh38
rs879371244 Chr.11: 419161-419161 on Build GRCh38
rs924933119 Chr.11: 419147-419147 on Build GRCh38
rs757856230 Chr.11: 418832-418832 on Build GRCh38
rs7482596 Chr.11: 419167-419167 on Build GRCh38
rs754169194 Chr.11: 418925-418925 on Build GRCh38
rs374660946 Chr.11: 418947-418947 on Build GRCh38
rs367921445 Chr.11: 418901-418901 on Build GRCh38
rs73402506 Chr.11: 419263-419263 on Build GRCh38
rs770726167 Chr.11: 418999-418999 on Build GRCh38
rs1037178792 Chr.11: 419221-419221 on Build GRCh38
rs533256091 Chr.11: 419230-419230 on Build GRCh38
rs773861579 Chr.11: 418865-418865 on Build GRCh38
rs762108633 Chr.11: 419035-419035 on Build GRCh38
rs542339850 Chr.11: 419124-419124 on Build GRCh38
rs549522312 Chr.11: 419255-419255 on Build GRCh38
rs897405278 Chr.11: 419222-419222 on Build GRCh38
rs917254911 Chr.11: 419204-419204 on Build GRCh38
rs759945469 Chr.11: 418830-418830 on Build GRCh38
rs888891150 Chr.11: 419113-419113 on Build GRCh38
rs757545958 Chr.11: 419053-419053 on Build GRCh38
rs1007611468 Chr.11: 419122-419122 on Build GRCh38
rs774775795 Chr.11: 419026-419026 on Build GRCh38
rs944216761 Chr.11: 419195-419195 on Build GRCh38
rs1018575232 Chr.11: 419125-419125 on Build GRCh38
rs1021306293 Chr.11: 419101-419101 on Build GRCh38
rs773192364 Chr.11: 418890-418890 on Build GRCh38
rs377197980 Chr.11: 419099-419099 on Build GRCh38
rs772566665 Chr.11: 418980-418980 on Build GRCh38
rs751696607 Chr.11: 418928-418928 on Build GRCh38
rs957770574 Chr.11: 419193-419193 on Build GRCh38
rs931533868 Chr.11: 419261-419261 on Build GRCh38
rs768300305 Chr.11: 418973-418973 on Build GRCh38
rs781413334 Chr.11: 418946-418946 on Build GRCh38
rs765892541 Chr.11: 418894-418895 on Build GRCh38
rs767732274 Chr.11: 419036-419036 on Build GRCh38
rs771565337 Chr.11: 418878-418878 on Build GRCh38
rs1033486306 Chr.11: 419175-419175 on Build GRCh38
rs932427673 Chr.11: 419159-419159 on Build GRCh38
rs757481877 Chr.11: 418935-418935 on Build GRCh38
rs796959622 Chr.11: 419133-419133 on Build GRCh38
rs979099195 Chr.11: 419134-419134 on Build GRCh38
rs960807394 Chr.11: 419070-419070 on Build GRCh38
rs371131273 Chr.11: 418856-418856 on Build GRCh38
rs368645509 Chr.11: 419011-419011 on Build GRCh38
rs991975253 Chr.11: 419073-419073 on Build GRCh38
rs746360214 Chr.11: 418838-418838 on Build GRCh38
rs764356229 Chr.11: 418923-418923 on Build GRCh38
rs547785181 Chr.11: 419219-419219 on Build GRCh38
rs376702614 Chr.11: 419014-419014 on Build GRCh38
rs771618673 Chr.11: 418977-418977 on Build GRCh38
rs778032728 Chr.11: 418817-418817 on Build GRCh38
rs747785580 Chr.11: 418975-418975 on Build GRCh38
rs781738375 Chr.11: 418837-418837 on Build GRCh38
rs374074944 Chr.11: 418869-418869 on Build GRCh38
rs760506226 Chr.11: 418897-418897 on Build GRCh38
rs371374296 Chr.11: 418902-418902 on Build GRCh38
rs763260469 Chr.11: 418908-418908 on Build GRCh38
rs562191228 Chr.11: 419156-419156 on Build GRCh38
rs950068408 Chr.11: 419213-419213 on Build GRCh38
rs776499926 Chr.11: 419010-419010 on Build GRCh38
rs564195317 Chr.11: 419229-419229 on Build GRCh38
rs368497607 Chr.11: 418848-418848 on Build GRCh38
rs759337830 Chr.11: 418828-418828 on Build GRCh38
rs778601535 Chr.11: 418974-418974 on Build GRCh38
rs912760436 Chr.11: 419188-419188 on Build GRCh38
rs760697784 Chr.11: 418992-418992 on Build GRCh38
rs750417054 Chr.11: 419041-419041 on Build GRCh38
rs780486405 Chr.11: 418955-418955 on Build GRCh38

View Full Product Details