Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 450385-450906 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTDSS2
Entrez Gene ID
81490
Gene Name
phosphatidylserine synthase 2
Gene Aliases
PSS2
UniGene
Hs.731700
Gene Chromosome Location
Chr.11: 448264-491399 on Build GRCh38
Transcripts
NM_030783.1 XM_005253153.1 XM_011520392.2 XM_011520393.2 XM_017018368.1 XM_017018369.1 XM_017018370.1 XM_017018371.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6374953 c.146C>A p.S49Y Chr.11: 450601-450601 on Build GRCh38
6397745 c.50C>G p.P17R Chr.11: 450505-450505 on Build GRCh38
1188103 c.31G>T p.G11C Chr.11: 450486-450486 on Build GRCh38
6042527 c.109G>A p.A37T Chr.11: 450564-450564 on Build GRCh38
4985473 c.8G>A p.R3K Chr.11: 450463-450463 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs890058192 Chr.11: 450757-450758 on Build GRCh38
rs751369515 Chr.11: 450462-450462 on Build GRCh38
rs926100254 Chr.11: 450700-450700 on Build GRCh38
rs117511007 Chr.11: 450769-450769 on Build GRCh38
rs1034255903 Chr.11: 450840-450840 on Build GRCh38
rs547193709 Chr.11: 450569-450569 on Build GRCh38
rs778888081 Chr.11: 450692-450692 on Build GRCh38
rs1040458969 Chr.11: 450617-450617 on Build GRCh38
rs539099548 Chr.11: 450662-450662 on Build GRCh38
rs1019692381 Chr.11: 450445-450445 on Build GRCh38
rs538248263 Chr.11: 450672-450672 on Build GRCh38
rs372615553 Chr.11: 450683-450683 on Build GRCh38
rs967495977 Chr.11: 450693-450693 on Build GRCh38
rs927240972 Chr.11: 450576-450576 on Build GRCh38
rs994022801 Chr.11: 450655-450655 on Build GRCh38
rs751140637 Chr.11: 450649-450649 on Build GRCh38
rs1050403988 Chr.11: 450527-450527 on Build GRCh38
rs186557747 Chr.11: 450599-450599 on Build GRCh38
rs936280231 Chr.11: 450709-450709 on Build GRCh38
rs531338729 Chr.11: 450463-450463 on Build GRCh38
rs910583327 Chr.11: 450737-450737 on Build GRCh38
rs980035063 Chr.11: 450545-450545 on Build GRCh38
rs182480820 Chr.11: 450485-450485 on Build GRCh38
rs530464406 Chr.11: 450512-450512 on Build GRCh38
rs1005507725 Chr.11: 450790-450790 on Build GRCh38
rs1046958052 Chr.11: 450785-450785 on Build GRCh38
rs929354594 Chr.11: 450776-450776 on Build GRCh38
rs1033168461 Chr.11: 450814-450814 on Build GRCh38
rs957533275 Chr.11: 450855-450855 on Build GRCh38
rs376660349 Chr.11: 450691-450691 on Build GRCh38
rs946232076 Chr.11: 450459-450459 on Build GRCh38
rs1024362152 Chr.11: 450799-450799 on Build GRCh38
rs541804773 Chr.11: 450887-450887 on Build GRCh38
rs553538102 Chr.11: 450750-450750 on Build GRCh38
rs967172816 Chr.11: 450808-450808 on Build GRCh38
rs550949084 Chr.11: 450471-450471 on Build GRCh38
rs931675967 Chr.11: 450708-450708 on Build GRCh38
rs567103161 Chr.11: 450661-450661 on Build GRCh38
rs1055715087 Chr.11: 450796-450796 on Build GRCh38
rs143829427 Chr.11: 450784-450784 on Build GRCh38
rs562610447 Chr.11: 450788-450788 on Build GRCh38
rs993024205 Chr.11: 450688-450688 on Build GRCh38
rs55768561 Chr.11: 450763-450763 on Build GRCh38
rs113547884 Chr.11: 450582-450582 on Build GRCh38
rs944728543 Chr.11: 450757-450757 on Build GRCh38
rs1013926733 Chr.11: 450798-450798 on Build GRCh38
rs889096423 Chr.11: 450550-450550 on Build GRCh38
rs960240413 Chr.11: 450841-450841 on Build GRCh38
rs940413150 Chr.11: 450860-450860 on Build GRCh38
rs569110004 Chr.11: 450671-450671 on Build GRCh38
rs969005751 Chr.11: 450502-450502 on Build GRCh38
rs907599035 Chr.11: 450657-450657 on Build GRCh38
rs998542439 Chr.11: 450812-450812 on Build GRCh38
rs771489950 Chr.11: 450761-450761 on Build GRCh38
rs574674313 Chr.11: 450728-450728 on Build GRCh38
rs1034046850 Chr.11: 450676-450676 on Build GRCh38
rs973120982 Chr.11: 450870-450870 on Build GRCh38
rs780965420 Chr.11: 450590-450590 on Build GRCh38
rs938508662 Chr.11: 450578-450578 on Build GRCh38
rs977571929 Chr.11: 450699-450699 on Build GRCh38
rs533867040 Chr.11: 450734-450734 on Build GRCh38
rs552662623 Chr.11: 450665-450665 on Build GRCh38
rs917437930 Chr.11: 450420-450420 on Build GRCh38
rs897865608 Chr.11: 450642-450642 on Build GRCh38
rs796092110 Chr.11: 450755-450755 on Build GRCh38
rs919362534 Chr.11: 450775-450775 on Build GRCh38
rs944821942 Chr.11: 450555-450555 on Build GRCh38
rs576095619 Chr.11: 450832-450832 on Build GRCh38
rs894342492 Chr.11: 450797-450797 on Build GRCh38
rs992594786 Chr.11: 450842-450842 on Build GRCh38

View Full Product Details