Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 837849-838346 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD151
Entrez Gene ID
977
Gene Name
CD151 molecule (Raph blood group)
Gene Aliases
GP27, MER2, PETA-3, RAPH, SFA1, TSPAN24
UniGene
Hs.654379
Gene Chromosome Location
Chr.11: 832952-838835 on Build GRCh38
Transcripts
NM_001039490.1 NM_004357.4 NM_139029.1 NM_139030.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7328069 c.729C>T p.C243C Chr.11: 838159-838159 on Build GRCh38
6134836 c.695G>T p.C232F Chr.11: 838021-838021 on Build GRCh38
3687819 c.684C>T p.I228I Chr.11: 838010-838010 on Build GRCh38
429973 c.645C>A p.F215L Chr.11: 837971-837971 on Build GRCh38
1746611 c.744C>G p.L248L Chr.11: 838174-838174 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767797783 Chr.11: 838019-838019 on Build GRCh38
rs140437381 Chr.11: 838250-838250 on Build GRCh38
rs981024956 Chr.11: 837873-837873 on Build GRCh38
rs773594481 Chr.11: 838090-838090 on Build GRCh38
rs760975473 Chr.11: 838187-838187 on Build GRCh38
rs912440689 Chr.11: 837922-837923 on Build GRCh38
rs781085595 Chr.11: 837903-837903 on Build GRCh38
rs749750091 Chr.11: 838071-838071 on Build GRCh38
rs771826222 Chr.11: 837983-837983 on Build GRCh38
rs780841918 Chr.11: 837944-837944 on Build GRCh38
rs559462734 Chr.11: 838283-838283 on Build GRCh38
rs771213389 Chr.11: 838239-838239 on Build GRCh38
rs1013583035 Chr.11: 838281-838281 on Build GRCh38
rs368757982 Chr.11: 837930-837930 on Build GRCh38
rs778215958 Chr.11: 838259-838259 on Build GRCh38
rs748823506 Chr.11: 837915-837915 on Build GRCh38
rs200997452 Chr.11: 837976-837976 on Build GRCh38
rs372451969 Chr.11: 838039-838039 on Build GRCh38
rs146967048 Chr.11: 838159-838159 on Build GRCh38
rs370569958 Chr.11: 838153-838153 on Build GRCh38
rs576870672 Chr.11: 837992-837992 on Build GRCh38
rs770245346 Chr.11: 837998-837998 on Build GRCh38
rs56125304 Chr.11: 837989-837989 on Build GRCh38
rs546483737 Chr.11: 838079-838079 on Build GRCh38
rs548707575 Chr.11: 837929-837929 on Build GRCh38
rs572689402 Chr.11: 838013-838013 on Build GRCh38
rs750178765 Chr.11: 838021-838021 on Build GRCh38
rs201597386 Chr.11: 838011-838011 on Build GRCh38
rs368998046 Chr.11: 838031-838031 on Build GRCh38
rs534240443 Chr.11: 837945-837945 on Build GRCh38
rs777986942 Chr.11: 838229-838229 on Build GRCh38
rs767809962 Chr.11: 838083-838083 on Build GRCh38
rs111724877 Chr.11: 837939-837939 on Build GRCh38
rs756061257 Chr.11: 837956-837956 on Build GRCh38
rs747100423 Chr.11: 838230-838230 on Build GRCh38
rs375649165 Chr.11: 838049-838049 on Build GRCh38
rs532677125 Chr.11: 837906-837906 on Build GRCh38
rs778535154 Chr.11: 837972-837972 on Build GRCh38
rs761052755 Chr.11: 837926-837926 on Build GRCh38
rs749673020 Chr.11: 838165-838165 on Build GRCh38
rs780824335 Chr.11: 838215-838215 on Build GRCh38
rs747752694 Chr.11: 838041-838041 on Build GRCh38
rs906848545 Chr.11: 838311-838311 on Build GRCh38
rs772260072 Chr.11: 838181-838181 on Build GRCh38
rs199919101 Chr.11: 837925-837925 on Build GRCh38
rs775870052 Chr.11: 838070-838070 on Build GRCh38
rs377744849 Chr.11: 837901-837901 on Build GRCh38
rs766768124 Chr.11: 838189-838189 on Build GRCh38
rs180832714 Chr.11: 837968-837968 on Build GRCh38
rs201420443 Chr.11: 838101-838101 on Build GRCh38
rs1011198232 Chr.11: 838267-838267 on Build GRCh38
rs769261930 Chr.11: 838076-838076 on Build GRCh38
rs943283562 Chr.11: 837933-837933 on Build GRCh38
rs753806157 Chr.11: 838032-838032 on Build GRCh38
rs879152631 Chr.11: 838128-838128 on Build GRCh38
rs991139142 Chr.11: 837877-837877 on Build GRCh38
rs924436516 Chr.11: 837922-837922 on Build GRCh38
rs35465458 Chr.11: 838115-838115 on Build GRCh38
rs1002331464 Chr.11: 838034-838034 on Build GRCh38
rs752107007 Chr.11: 838142-838142 on Build GRCh38
rs200873117 Chr.11: 838050-838050 on Build GRCh38
rs188674544 Chr.11: 838205-838205 on Build GRCh38
rs768292922 Chr.11: 837916-837916 on Build GRCh38
rs765745305 Chr.11: 837931-837931 on Build GRCh38
rs563094795 Chr.11: 837914-837914 on Build GRCh38
rs200052049 Chr.11: 837964-837964 on Build GRCh38
rs989850577 Chr.11: 837902-837902 on Build GRCh38
rs967035449 Chr.11: 838301-838302 on Build GRCh38
rs756966691 Chr.11: 838158-838158 on Build GRCh38
rs755395021 Chr.11: 838219-838219 on Build GRCh38
rs374971560 Chr.11: 838204-838204 on Build GRCh38
rs1025126989 Chr.11: 837868-837868 on Build GRCh38
rs568567149 Chr.11: 837938-837938 on Build GRCh38
rs764046037 Chr.11: 838010-838010 on Build GRCh38
rs373886552 Chr.11: 838056-838056 on Build GRCh38
rs748660774 Chr.11: 838225-838225 on Build GRCh38
rs746181850 Chr.11: 838059-838059 on Build GRCh38
rs5030780 Chr.11: 838110-838110 on Build GRCh38
rs879027656 Chr.11: 838129-838129 on Build GRCh38
rs752657569 Chr.11: 838203-838203 on Build GRCh38
rs751123997 Chr.11: 838206-838206 on Build GRCh38
rs200214987 Chr.11: 838069-838069 on Build GRCh38
rs35042031 Chr.11: 838222-838222 on Build GRCh38
rs765034001 Chr.11: 838196-838196 on Build GRCh38
rs759897271 Chr.11: 837994-837994 on Build GRCh38
rs562246419 Chr.11: 838292-838292 on Build GRCh38
rs564090455 Chr.11: 838127-838127 on Build GRCh38
rs372436486 Chr.11: 838218-838218 on Build GRCh38
rs774956479 Chr.11: 838172-838172 on Build GRCh38
rs563132103 Chr.11: 838209-838209 on Build GRCh38
rs771177871 Chr.11: 837927-837927 on Build GRCh38
rs570753524 Chr.11: 837975-837975 on Build GRCh38
rs775998930 Chr.11: 838007-838007 on Build GRCh38
rs527764114 Chr.11: 838299-838299 on Build GRCh38
rs773467335 Chr.11: 838183-838183 on Build GRCh38
rs374859485 Chr.11: 838025-838025 on Build GRCh38
rs777122361 Chr.11: 838164-838165 on Build GRCh38
rs143948122 Chr.11: 838118-838118 on Build GRCh38
rs926886125 Chr.11: 837875-837875 on Build GRCh38
rs748990514 Chr.11: 838276-838276 on Build GRCh38
rs374891770 Chr.11: 837937-837937 on Build GRCh38
rs894033100 Chr.11: 838266-838266 on Build GRCh38
rs772773681 Chr.11: 838226-838226 on Build GRCh38
rs768980499 Chr.11: 838170-838170 on Build GRCh38
rs563180215 Chr.11: 837984-837984 on Build GRCh38
rs945847747 Chr.11: 837909-837909 on Build GRCh38
rs878900628 Chr.11: 838126-838126 on Build GRCh38
rs1031447241 Chr.11: 838089-838089 on Build GRCh38
rs202108260 Chr.11: 838100-838100 on Build GRCh38
rs759405745 Chr.11: 838191-838191 on Build GRCh38
rs780146527 Chr.11: 837958-837958 on Build GRCh38
rs761960301 Chr.11: 838080-838080 on Build GRCh38
rs746778064 Chr.11: 837899-837899 on Build GRCh38
rs771634557 Chr.11: 837934-837934 on Build GRCh38
rs1021316795 Chr.11: 838060-838060 on Build GRCh38
rs372735477 Chr.11: 837932-837932 on Build GRCh38
rs887154171 Chr.11: 838163-838163 on Build GRCh38
rs35325640 Chr.11: 837913-837913 on Build GRCh38
rs35264386 Chr.11: 837871-837871 on Build GRCh38
rs777401956 Chr.11: 838081-838081 on Build GRCh38

View Full Product Details