Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 5697156-5697661 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRIM22
Entrez Gene ID
10346
Gene Name
tripartite motif containing 22
Gene Aliases
GPSTAF50, RNF94, STAF50
UniGene
Hs.501778
Gene Chromosome Location
Chr.11: 5689587-5710863 on Build GRCh38
Transcripts
NM_001199573.1 NM_006074.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4910999 c.519+1G>A p.? Chr.11: 5697344-5697344 on Build GRCh38
6014266 c.442C>T p.L148L Chr.11: 5697266-5697266 on Build GRCh38
1475564 c.495C>T p.I165I Chr.11: 5697319-5697319 on Build GRCh38
4548679 c.453G>A p.L151L Chr.11: 5697277-5697277 on Build GRCh38
5593572 c.424G>A p.E142K Chr.11: 5697248-5697248 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs188251473 Chr.11: 5697476-5697476 on Build GRCh38
rs767918987 Chr.11: 5697277-5697277 on Build GRCh38
rs749253174 Chr.11: 5697377-5697377 on Build GRCh38
rs771589887 Chr.11: 5697234-5697234 on Build GRCh38
rs746666286 Chr.11: 5697241-5697241 on Build GRCh38
rs184702467 Chr.11: 5697214-5697214 on Build GRCh38
rs896832332 Chr.11: 5697564-5697564 on Build GRCh38
rs771123565 Chr.11: 5697395-5697395 on Build GRCh38
rs979319753 Chr.11: 5697404-5697404 on Build GRCh38
rs556933383 Chr.11: 5697593-5697593 on Build GRCh38
rs144534727 Chr.11: 5697579-5697579 on Build GRCh38
rs745430542 Chr.11: 5697224-5697224 on Build GRCh38
rs1028843933 Chr.11: 5697615-5697615 on Build GRCh38
rs1023135857 Chr.11: 5697632-5697632 on Build GRCh38
rs867133296 Chr.11: 5697302-5697302 on Build GRCh38
rs1053032857 Chr.11: 5697499-5697499 on Build GRCh38
rs773217733 Chr.11: 5697374-5697374 on Build GRCh38
rs1029209244 Chr.11: 5697392-5697392 on Build GRCh38
rs145950369 Chr.11: 5697345-5697345 on Build GRCh38
rs547975419 Chr.11: 5697398-5697398 on Build GRCh38
rs139741647 Chr.11: 5697534-5697534 on Build GRCh38
rs559830362 Chr.11: 5697410-5697410 on Build GRCh38
rs990534502 Chr.11: 5697603-5697604 on Build GRCh38
rs934077384 Chr.11: 5697522-5697522 on Build GRCh38
rs769662561 Chr.11: 5697373-5697373 on Build GRCh38
rs781022253 Chr.11: 5697357-5697357 on Build GRCh38
rs570586516 Chr.11: 5697521-5697521 on Build GRCh38
rs768438040 Chr.11: 5697246-5697246 on Build GRCh38
rs750030346 Chr.11: 5697517-5697517 on Build GRCh38
rs200924168 Chr.11: 5697273-5697273 on Build GRCh38
rs1004503857 Chr.11: 5697554-5697554 on Build GRCh38
rs773308232 Chr.11: 5697256-5697256 on Build GRCh38
rs997361966 Chr.11: 5697355-5697355 on Build GRCh38
rs748003845 Chr.11: 5697192-5697192 on Build GRCh38
rs766572056 Chr.11: 5697267-5697267 on Build GRCh38
rs181280875 Chr.11: 5697417-5697417 on Build GRCh38
rs1036398211 Chr.11: 5697580-5697580 on Build GRCh38
rs772389956 Chr.11: 5697205-5697205 on Build GRCh38
rs765909592 Chr.11: 5697616-5697616 on Build GRCh38
rs375795798 Chr.11: 5697279-5697279 on Build GRCh38
rs7935929 Chr.11: 5697587-5697587 on Build GRCh38
rs553638122 Chr.11: 5697335-5697335 on Build GRCh38
rs747853537 Chr.11: 5697369-5697369 on Build GRCh38
rs753040398 Chr.11: 5697457-5697457 on Build GRCh38
rs753233760 Chr.11: 5697215-5697215 on Build GRCh38
rs779550205 Chr.11: 5697337-5697337 on Build GRCh38
rs779637948 Chr.11: 5697239-5697239 on Build GRCh38
rs1017241894 Chr.11: 5697547-5697547 on Build GRCh38
rs993863481 Chr.11: 5697575-5697575 on Build GRCh38
rs184273979 Chr.11: 5697430-5697430 on Build GRCh38
rs1005471993 Chr.11: 5697538-5697538 on Build GRCh38
rs751776081 Chr.11: 5697269-5697269 on Build GRCh38
rs761782306 Chr.11: 5697248-5697248 on Build GRCh38
rs916073958 Chr.11: 5697451-5697451 on Build GRCh38
rs1045252519 Chr.11: 5697477-5697477 on Build GRCh38
rs763087288 Chr.11: 5697266-5697266 on Build GRCh38
rs760088006 Chr.11: 5697351-5697351 on Build GRCh38
rs190022478 Chr.11: 5697334-5697334 on Build GRCh38
rs769673655 Chr.11: 5697251-5697251 on Build GRCh38
rs7935903 Chr.11: 5697512-5697512 on Build GRCh38
rs7935564 Chr.11: 5697287-5697287 on Build GRCh38
rs367599367 Chr.11: 5697221-5697221 on Build GRCh38
rs888927482 Chr.11: 5697630-5697630 on Build GRCh38
rs956677287 Chr.11: 5697420-5697420 on Build GRCh38
rs760826020 Chr.11: 5697217-5697217 on Build GRCh38
rs7936676 Chr.11: 5697482-5697482 on Build GRCh38

View Full Product Details